Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 66516201-66516713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf46
Entrez Gene ID
254778
Gene Name
chromosome 8 open reading frame 46
Gene Aliases
-
UniGene
Hs.268869
Gene Chromosome Location
Chr.8: 66493256-66518524 on Build GRCh38
Transcripts
NM_152765.3

SNP Detail

SNP ID Chromosome Location
rs995506507 Chr.8: 66516363-66516363 on Build GRCh38
rs1040400898 Chr.8: 66516629-66516629 on Build GRCh38
rs903599946 Chr.8: 66516247-66516247 on Build GRCh38
rs182252216 Chr.8: 66516387-66516387 on Build GRCh38
rs1049518735 Chr.8: 66516625-66516625 on Build GRCh38
rs995537606 Chr.8: 66516650-66516650 on Build GRCh38
rs575407553 Chr.8: 66516609-66516609 on Build GRCh38
rs775320901 Chr.8: 66516268-66516268 on Build GRCh38
rs572737763 Chr.8: 66516429-66516429 on Build GRCh38
rs898504684 Chr.8: 66516310-66516310 on Build GRCh38
rs1051686827 Chr.8: 66516248-66516248 on Build GRCh38
rs1019633845 Chr.8: 66516345-66516345 on Build GRCh38
rs1007570652 Chr.8: 66516272-66516272 on Build GRCh38
rs1043784710 Chr.8: 66516546-66516546 on Build GRCh38
rs544597175 Chr.8: 66516680-66516680 on Build GRCh38
rs557438697 Chr.8: 66516295-66516295 on Build GRCh38
rs1009482564 Chr.8: 66516676-66516676 on Build GRCh38
rs1014101578 Chr.8: 66516687-66516687 on Build GRCh38
rs370804349 Chr.8: 66516319-66516320 on Build GRCh38
rs927175095 Chr.8: 66516489-66516489 on Build GRCh38
rs989765917 Chr.8: 66516521-66516521 on Build GRCh38
rs757535780 Chr.8: 66516479-66516479 on Build GRCh38
rs555648680 Chr.8: 66516599-66516599 on Build GRCh38
rs1009123135 Chr.8: 66516315-66516316 on Build GRCh38
rs753748678 Chr.8: 66516434-66516434 on Build GRCh38
rs980434174 Chr.8: 66516482-66516482 on Build GRCh38
rs552642677 Chr.8: 66516409-66516409 on Build GRCh38
rs913011182 Chr.8: 66516543-66516543 on Build GRCh38
rs951431581 Chr.8: 66516397-66516397 on Build GRCh38
rs935708073 Chr.8: 66516506-66516506 on Build GRCh38
rs942847523 Chr.8: 66516621-66516621 on Build GRCh38
rs1025633537 Chr.8: 66516661-66516661 on Build GRCh38
rs537521974 Chr.8: 66516292-66516292 on Build GRCh38
rs550269778 Chr.8: 66516279-66516279 on Build GRCh38
rs898743381 Chr.8: 66516639-66516639 on Build GRCh38
rs891399942 Chr.8: 66516256-66516256 on Build GRCh38
rs945037388 Chr.8: 66516234-66516234 on Build GRCh38
rs779216337 Chr.8: 66516536-66516536 on Build GRCh38
rs535243208 Chr.8: 66516545-66516545 on Build GRCh38
rs903546173 Chr.8: 66516589-66516589 on Build GRCh38
rs972482916 Chr.8: 66516380-66516380 on Build GRCh38
rs946251023 Chr.8: 66516236-66516236 on Build GRCh38
rs889265469 Chr.8: 66516638-66516638 on Build GRCh38
rs1050840551 Chr.8: 66516581-66516581 on Build GRCh38
rs891299070 Chr.8: 66516583-66516583 on Build GRCh38
rs939204458 Chr.8: 66516462-66516462 on Build GRCh38
rs187922417 Chr.8: 66516406-66516406 on Build GRCh38
rs887084559 Chr.8: 66516686-66516686 on Build GRCh38

View Full Product Details