Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 67175135-67175680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFGEF1
Entrez Gene ID
10565
Gene Name
ADP ribosylation factor guanine nucleotide exchange factor 1
Gene Aliases
ARFGEP1, BIG1, P200
UniGene
Hs.656902
Gene Chromosome Location
Chr.8: 67174298-67343927 on Build GRCh38
Transcripts
XM_005251136.3 NM_001291339.1 NM_024790.6 XM_005251305.3 XM_006716474.2 XM_006716477.2 XM_011517598.1 XM_011517599.1 XM_011517600.1 XM_011517601.1 XM_011517602.1 XM_011517603.1 XM_011517607.1 XM_011517608.1 XM_011517609.1 XM_011517611.2 XM_017013847.1 XM_017013848.1 XM_017013849.1 XM_017013850.1 XM_017013851.1 XM_017013852.1 XM_017013853.1 XM_017013854.1 XM_017013855.1 XM_017013856.1 XM_017013857.1 XM_017013858.1 XM_017013859.1

Gene Symbol
CSPP1
Entrez Gene ID
79848
Gene Name
centrosome and spindle pole associated protein 1
Gene Aliases
CSPP, JBTS21
UniGene
Hs.370147
Gene Chromosome Location
Chr.8: 67062417-67196614 on Build GRCh38
Transcripts
XM_005251136.3 NM_001291339.1 NM_024790.6 XM_005251305.3 XM_006716474.2 XM_006716477.2 XM_011517598.1 XM_011517599.1 XM_011517600.1 XM_011517601.1 XM_011517602.1 XM_011517603.1 XM_011517607.1 XM_011517608.1 XM_011517609.1 XM_011517611.2 XM_017013847.1 XM_017013848.1 XM_017013849.1 XM_017013850.1 XM_017013851.1 XM_017013852.1 XM_017013853.1 XM_017013854.1 XM_017013855.1 XM_017013856.1 XM_017013857.1 XM_017013858.1 XM_017013859.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
454771 c.3004G>C p.D1002H Chr.8: 67175346-67175346 on Build GRCh38
6636909 c.2989C>A p.L997M Chr.8: 67175331-67175331 on Build GRCh38
1101195 c.3019G>T p.E1007* Chr.8: 67175361-67175361 on Build GRCh38
750922 c.3046G>C p.E1016Q Chr.8: 67175388-67175388 on Build GRCh38
6636900 c.2966G>A p.R989Q Chr.8: 67175308-67175308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922371394 Chr.8: 67175203-67175203 on Build GRCh38
rs560702910 Chr.8: 67175623-67175623 on Build GRCh38
rs149297949 Chr.8: 67175472-67175472 on Build GRCh38
rs766403684 Chr.8: 67175617-67175617 on Build GRCh38
rs537349039 Chr.8: 67175209-67175209 on Build GRCh38
rs556839762 Chr.8: 67175275-67175275 on Build GRCh38
rs775097330 Chr.8: 67175298-67175298 on Build GRCh38
rs75428500 Chr.8: 67175183-67175183 on Build GRCh38
rs1014781544 Chr.8: 67175384-67175384 on Build GRCh38
rs774687205 Chr.8: 67175439-67175439 on Build GRCh38
rs750306493 Chr.8: 67175463-67175463 on Build GRCh38
rs1014337056 Chr.8: 67175643-67175643 on Build GRCh38
rs756807936 Chr.8: 67175331-67175331 on Build GRCh38
rs769354548 Chr.8: 67175556-67175556 on Build GRCh38
rs993509179 Chr.8: 67175338-67175338 on Build GRCh38
rs749389567 Chr.8: 67175260-67175260 on Build GRCh38
rs780045401 Chr.8: 67175413-67175413 on Build GRCh38
rs746830683 Chr.8: 67175428-67175428 on Build GRCh38
rs201723047 Chr.8: 67175530-67175530 on Build GRCh38
rs555730971 Chr.8: 67175467-67175467 on Build GRCh38
rs62513125 Chr.8: 67175475-67175475 on Build GRCh38
rs1023123048 Chr.8: 67175389-67175389 on Build GRCh38
rs765074640 Chr.8: 67175334-67175334 on Build GRCh38
rs367949127 Chr.8: 67175308-67175308 on Build GRCh38
rs1049563230 Chr.8: 67175223-67175223 on Build GRCh38
rs759812666 Chr.8: 67175283-67175283 on Build GRCh38
rs769189572 Chr.8: 67175326-67175331 on Build GRCh38
rs778067206 Chr.8: 67175432-67175432 on Build GRCh38
rs374717800 Chr.8: 67175367-67175367 on Build GRCh38
rs191548161 Chr.8: 67175464-67175464 on Build GRCh38
rs767082092 Chr.8: 67175508-67175509 on Build GRCh38
rs969480103 Chr.8: 67175656-67175656 on Build GRCh38
rs960856836 Chr.8: 67175629-67175630 on Build GRCh38
rs759558950 Chr.8: 67175533-67175533 on Build GRCh38
rs139214760 Chr.8: 67175566-67175566 on Build GRCh38
rs762942654 Chr.8: 67175466-67175466 on Build GRCh38
rs762472103 Chr.8: 67175488-67175488 on Build GRCh38
rs926218821 Chr.8: 67175437-67175437 on Build GRCh38
rs772779980 Chr.8: 67175561-67175561 on Build GRCh38
rs775794851 Chr.8: 67175444-67175444 on Build GRCh38
rs369984993 Chr.8: 67175200-67175200 on Build GRCh38
rs780554630 Chr.8: 67175522-67175522 on Build GRCh38
rs771283905 Chr.8: 67175262-67175262 on Build GRCh38
rs372291198 Chr.8: 67175440-67175440 on Build GRCh38
rs772505024 Chr.8: 67175515-67175515 on Build GRCh38
rs751340189 Chr.8: 67175470-67175470 on Build GRCh38
rs146234559 Chr.8: 67175487-67175487 on Build GRCh38
rs960742996 Chr.8: 67175333-67175333 on Build GRCh38
rs770162804 Chr.8: 67175252-67175252 on Build GRCh38
rs749633996 Chr.8: 67175282-67175283 on Build GRCh38
rs767905675 Chr.8: 67175288-67175288 on Build GRCh38
rs774977183 Chr.8: 67175266-67175266 on Build GRCh38
rs761576017 Chr.8: 67175552-67175552 on Build GRCh38
rs933803709 Chr.8: 67175204-67175204 on Build GRCh38
rs376619870 Chr.8: 67175450-67175450 on Build GRCh38
rs760275528 Chr.8: 67175307-67175307 on Build GRCh38
rs750443075 Chr.8: 67175356-67175356 on Build GRCh38
rs549393576 Chr.8: 67175640-67175640 on Build GRCh38
rs754127361 Chr.8: 67175476-67175476 on Build GRCh38
rs772586492 Chr.8: 67175441-67175441 on Build GRCh38
rs548949213 Chr.8: 67175509-67175509 on Build GRCh38
rs922296361 Chr.8: 67175473-67175473 on Build GRCh38
rs895851308 Chr.8: 67175641-67175641 on Build GRCh38
rs563868874 Chr.8: 67175211-67175211 on Build GRCh38
rs756338126 Chr.8: 67175430-67175430 on Build GRCh38
rs200546493 Chr.8: 67175436-67175436 on Build GRCh38
rs139599638 Chr.8: 67175454-67175454 on Build GRCh38
rs774996607 Chr.8: 67175346-67175347 on Build GRCh38
rs889647243 Chr.8: 67175235-67175235 on Build GRCh38
rs74996372 Chr.8: 67175648-67175648 on Build GRCh38
rs371720718 Chr.8: 67175314-67175314 on Build GRCh38
rs1047746686 Chr.8: 67175578-67175578 on Build GRCh38
rs773124318 Chr.8: 67175259-67175259 on Build GRCh38
rs1033841418 Chr.8: 67175572-67175572 on Build GRCh38
rs373555747 Chr.8: 67175456-67175456 on Build GRCh38
rs767330628 Chr.8: 67175540-67175540 on Build GRCh38
rs369060106 Chr.8: 67175433-67175433 on Build GRCh38
rs994180172 Chr.8: 67175179-67175179 on Build GRCh38
rs757258532 Chr.8: 67175486-67175486 on Build GRCh38
rs764843751 Chr.8: 67175461-67175461 on Build GRCh38
rs934332794 Chr.8: 67175159-67175159 on Build GRCh38
rs776405087 Chr.8: 67175455-67175455 on Build GRCh38
rs772607300 Chr.8: 67175555-67175555 on Build GRCh38

View Full Product Details