Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 67195285-67195832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFGEF1
Entrez Gene ID
10565
Gene Name
ADP ribosylation factor guanine nucleotide exchange factor 1
Gene Aliases
ARFGEP1, BIG1, P200
UniGene
Hs.656902
Gene Chromosome Location
Chr.8: 67174298-67343927 on Build GRCh38
Transcripts
XM_005251136.3 NM_001291339.1 NM_024790.6 XM_005251305.3 XM_006716474.2 XM_006716477.2 XM_011517598.1 XM_011517599.1 XM_011517600.1 XM_011517601.1 XM_011517602.1 XM_011517603.1 XM_011517607.1 XM_011517608.1 XM_011517609.1 XM_011517611.2 XM_017013847.1 XM_017013848.1 XM_017013849.1 XM_017013850.1 XM_017013851.1 XM_017013852.1 XM_017013853.1 XM_017013854.1 XM_017013855.1 XM_017013856.1 XM_017013857.1 XM_017013858.1 XM_017013859.1

Gene Symbol
CSPP1
Entrez Gene ID
79848
Gene Name
centrosome and spindle pole associated protein 1
Gene Aliases
CSPP, JBTS21
UniGene
Hs.370147
Gene Chromosome Location
Chr.8: 67062417-67196614 on Build GRCh38
Transcripts
XM_005251136.3 NM_001291339.1 NM_024790.6 XM_005251305.3 XM_006716474.2 XM_006716477.2 XM_011517598.1 XM_011517599.1 XM_011517600.1 XM_011517601.1 XM_011517602.1 XM_011517603.1 XM_011517607.1 XM_011517608.1 XM_011517609.1 XM_011517611.2 XM_017013847.1 XM_017013848.1 XM_017013849.1 XM_017013850.1 XM_017013851.1 XM_017013852.1 XM_017013853.1 XM_017013854.1 XM_017013855.1 XM_017013856.1 XM_017013857.1 XM_017013858.1 XM_017013859.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7187015 c.3637T>C p.W1213R Chr.8: 67195564-67195564 on Build GRCh38
6636904 c.3610A>G p.I1204V Chr.8: 67195537-67195537 on Build GRCh38
239468 c.3563C>T p.T1188M Chr.8: 67195490-67195490 on Build GRCh38
5976910 c.3556_3557insCAGAAAC p.L1189fs*>36 Chr.8: 67195483-67195484 on Build GRCh38
5414930 c.3515C>G p.S1172* Chr.8: 67195442-67195442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778807238 Chr.8: 67195449-67195449 on Build GRCh38
rs755185914 Chr.8: 67195453-67195453 on Build GRCh38
rs560993203 Chr.8: 67195321-67195321 on Build GRCh38
rs577603880 Chr.8: 67195749-67195752 on Build GRCh38
rs536196752 Chr.8: 67195370-67195370 on Build GRCh38
rs527426163 Chr.8: 67195550-67195550 on Build GRCh38
rs749332092 Chr.8: 67195557-67195557 on Build GRCh38
rs191829129 Chr.8: 67195759-67195759 on Build GRCh38
rs753937898 Chr.8: 67195608-67195608 on Build GRCh38
rs774627165 Chr.8: 67195497-67195497 on Build GRCh38
rs267601974 Chr.8: 67195439-67195439 on Build GRCh38
rs200079718 Chr.8: 67195438-67195438 on Build GRCh38
rs781579090 Chr.8: 67195471-67195471 on Build GRCh38
rs780395454 Chr.8: 67195348-67195348 on Build GRCh38
rs1040715506 Chr.8: 67195584-67195584 on Build GRCh38
rs759903681 Chr.8: 67195498-67195498 on Build GRCh38
rs772729206 Chr.8: 67195381-67195383 on Build GRCh38
rs766814639 Chr.8: 67195390-67195390 on Build GRCh38
rs747288364 Chr.8: 67195357-67195357 on Build GRCh38
rs760288295 Chr.8: 67195437-67195437 on Build GRCh38
rs969965795 Chr.8: 67195613-67195613 on Build GRCh38
rs200161440 Chr.8: 67195405-67195405 on Build GRCh38
rs776145236 Chr.8: 67195419-67195420 on Build GRCh38
rs770143622 Chr.8: 67195364-67195364 on Build GRCh38
rs923782575 Chr.8: 67195795-67195795 on Build GRCh38
rs374262758 Chr.8: 67195441-67195441 on Build GRCh38
rs779358572 Chr.8: 67195627-67195627 on Build GRCh38
rs148896413 Chr.8: 67195362-67195362 on Build GRCh38
rs769025909 Chr.8: 67195361-67195361 on Build GRCh38
rs770516051 Chr.8: 67195394-67195422 on Build GRCh38
rs980768057 Chr.8: 67195636-67195636 on Build GRCh38
rs758328481 Chr.8: 67195535-67195535 on Build GRCh38
rs762818148 Chr.8: 67195589-67195589 on Build GRCh38
rs776007873 Chr.8: 67195444-67195444 on Build GRCh38
rs142626075 Chr.8: 67195378-67195378 on Build GRCh38
rs188257548 Chr.8: 67195716-67195716 on Build GRCh38
rs534825175 Chr.8: 67195796-67195798 on Build GRCh38
rs772491058 Chr.8: 67195371-67195372 on Build GRCh38
rs764952763 Chr.8: 67195592-67195593 on Build GRCh38
rs761243317 Chr.8: 67195578-67195578 on Build GRCh38
rs147611370 Chr.8: 67195418-67195418 on Build GRCh38
rs1026132903 Chr.8: 67195532-67195532 on Build GRCh38
rs768228167 Chr.8: 67195577-67195577 on Build GRCh38
rs748165041 Chr.8: 67195472-67195472 on Build GRCh38
rs552763349 Chr.8: 67195491-67195491 on Build GRCh38
rs775187286 Chr.8: 67195511-67195511 on Build GRCh38
rs760969187 Chr.8: 67195488-67195488 on Build GRCh38
rs760082779 Chr.8: 67195383-67195384 on Build GRCh38
rs567275883 Chr.8: 67195648-67195648 on Build GRCh38
rs759243151 Chr.8: 67195540-67195540 on Build GRCh38
rs773828395 Chr.8: 67195366-67195366 on Build GRCh38
rs769955793 Chr.8: 67195475-67195475 on Build GRCh38
rs756293613 Chr.8: 67195601-67195601 on Build GRCh38
rs753315035 Chr.8: 67195524-67195524 on Build GRCh38
rs368550470 Chr.8: 67195567-67195567 on Build GRCh38
rs950791497 Chr.8: 67195314-67195314 on Build GRCh38
rs936435444 Chr.8: 67195531-67195531 on Build GRCh38
rs145624133 Chr.8: 67195322-67195322 on Build GRCh38
rs766225860 Chr.8: 67195590-67195590 on Build GRCh38
rs762646043 Chr.8: 67195678-67195678 on Build GRCh38
rs777867972 Chr.8: 67195551-67195551 on Build GRCh38
rs377160929 Chr.8: 67195565-67195565 on Build GRCh38
rs1006314875 Chr.8: 67195700-67195701 on Build GRCh38
rs760329183 Chr.8: 67195574-67195574 on Build GRCh38
rs372265885 Chr.8: 67195599-67195599 on Build GRCh38
rs780534337 Chr.8: 67195367-67195367 on Build GRCh38
rs746426693 Chr.8: 67195637-67195637 on Build GRCh38
rs752140903 Chr.8: 67195432-67195432 on Build GRCh38
rs767157409 Chr.8: 67195425-67195425 on Build GRCh38
rs943662063 Chr.8: 67195766-67195766 on Build GRCh38
rs760074883 Chr.8: 67195512-67195512 on Build GRCh38
rs936793531 Chr.8: 67195340-67195340 on Build GRCh38
rs925310954 Chr.8: 67195668-67195668 on Build GRCh38
rs771696486 Chr.8: 67195503-67195503 on Build GRCh38
rs933454236 Chr.8: 67195738-67195738 on Build GRCh38
rs201972175 Chr.8: 67195525-67195525 on Build GRCh38
rs886568987 Chr.8: 67195677-67195677 on Build GRCh38
rs772464232 Chr.8: 67195572-67195572 on Build GRCh38
rs959484549 Chr.8: 67195722-67195722 on Build GRCh38
rs376255546 Chr.8: 67195587-67195587 on Build GRCh38
rs914006889 Chr.8: 67195730-67195730 on Build GRCh38
rs750278770 Chr.8: 67195450-67195450 on Build GRCh38
rs988656315 Chr.8: 67195717-67195717 on Build GRCh38
rs778119559 Chr.8: 67195486-67195486 on Build GRCh38
rs781562670 Chr.8: 67195545-67195545 on Build GRCh38
rs764232514 Chr.8: 67195605-67195605 on Build GRCh38
rs765253659 Chr.8: 67195586-67195586 on Build GRCh38
rs541017968 Chr.8: 67195309-67195309 on Build GRCh38
rs553068500 Chr.8: 67195796-67195797 on Build GRCh38
rs750186693 Chr.8: 67195527-67195527 on Build GRCh38
rs757391546 Chr.8: 67195609-67195609 on Build GRCh38
rs201142840 Chr.8: 67195490-67195490 on Build GRCh38
rs549823805 Chr.8: 67195747-67195747 on Build GRCh38
rs752492612 Chr.8: 67195639-67195640 on Build GRCh38
rs771365440 Chr.8: 67195559-67195559 on Build GRCh38
rs763195750 Chr.8: 67195369-67195369 on Build GRCh38

View Full Product Details