Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 67198336-67198853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFGEF1
Entrez Gene ID
10565
Gene Name
ADP ribosylation factor guanine nucleotide exchange factor 1
Gene Aliases
ARFGEP1, BIG1, P200
UniGene
Hs.656902
Gene Chromosome Location
Chr.8: 67174298-67343927 on Build GRCh38
Transcripts
NM_006421.4 XM_005251134.4 XM_005251135.3 XM_005251136.3 XM_011517442.2

SNP Detail

SNP ID Chromosome Location
rs1042334693 Chr.8: 67198660-67198660 on Build GRCh38
rs947599424 Chr.8: 67198651-67198651 on Build GRCh38
rs962009389 Chr.8: 67198366-67198366 on Build GRCh38
rs573163778 Chr.8: 67198553-67198553 on Build GRCh38
rs777059843 Chr.8: 67198425-67198425 on Build GRCh38
rs541130204 Chr.8: 67198506-67198506 on Build GRCh38
rs947125324 Chr.8: 67198491-67198491 on Build GRCh38
rs553438682 Chr.8: 67198805-67198805 on Build GRCh38
rs138961233 Chr.8: 67198667-67198667 on Build GRCh38
rs185469115 Chr.8: 67198597-67198597 on Build GRCh38
rs539822467 Chr.8: 67198768-67198768 on Build GRCh38
rs908472649 Chr.8: 67198360-67198360 on Build GRCh38
rs979962604 Chr.8: 67198367-67198367 on Build GRCh38
rs1007811991 Chr.8: 67198668-67198668 on Build GRCh38
rs551071279 Chr.8: 67198689-67198689 on Build GRCh38
rs981448365 Chr.8: 67198394-67198394 on Build GRCh38
rs989945234 Chr.8: 67198638-67198638 on Build GRCh38
rs1027416927 Chr.8: 67198705-67198705 on Build GRCh38
rs867401881 Chr.8: 67198713-67198713 on Build GRCh38
rs914750118 Chr.8: 67198650-67198650 on Build GRCh38
rs1043119259 Chr.8: 67198504-67198504 on Build GRCh38
rs891364987 Chr.8: 67198611-67198611 on Build GRCh38
rs928664417 Chr.8: 67198422-67198422 on Build GRCh38
rs367961662 Chr.8: 67198688-67198688 on Build GRCh38
rs748530236 Chr.8: 67198682-67198682 on Build GRCh38
rs913007671 Chr.8: 67198456-67198456 on Build GRCh38
rs73256687 Chr.8: 67198730-67198730 on Build GRCh38
rs900146003 Chr.8: 67198519-67198519 on Build GRCh38
rs988661458 Chr.8: 67198446-67198447 on Build GRCh38
rs181218449 Chr.8: 67198373-67198373 on Build GRCh38
rs938544177 Chr.8: 67198416-67198420 on Build GRCh38
rs937443448 Chr.8: 67198487-67198487 on Build GRCh38

View Full Product Details