Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 23430456-23430978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD4
Entrez Gene ID
9583
Gene Name
ectonucleoside triphosphate diphosphohydrolase 4
Gene Aliases
LALP70, LAP70, LYSAL1, NTPDase-4, UDPase
UniGene
Hs.444389
Gene Chromosome Location
Chr.8: 23429152-23457731 on Build GRCh38
Transcripts
NM_001128930.2 NM_004901.4

SNP Detail

SNP ID Chromosome Location
rs553174368 Chr.8: 23430725-23430725 on Build GRCh38
rs908313141 Chr.8: 23430750-23430750 on Build GRCh38
rs1015324290 Chr.8: 23430888-23430888 on Build GRCh38
rs932101918 Chr.8: 23430693-23430693 on Build GRCh38
rs555621143 Chr.8: 23430582-23430582 on Build GRCh38
rs115273243 Chr.8: 23430585-23430585 on Build GRCh38
rs970187985 Chr.8: 23430680-23430680 on Build GRCh38
rs952393111 Chr.8: 23430719-23430719 on Build GRCh38
rs928592074 Chr.8: 23430754-23430754 on Build GRCh38
rs747171316 Chr.8: 23430741-23430741 on Build GRCh38
rs148740800 Chr.8: 23430891-23430891 on Build GRCh38
rs894315339 Chr.8: 23430853-23430853 on Build GRCh38
rs541195291 Chr.8: 23430640-23430640 on Build GRCh38
rs989900105 Chr.8: 23430701-23430701 on Build GRCh38
rs966040606 Chr.8: 23430890-23430890 on Build GRCh38
rs561132103 Chr.8: 23430943-23430943 on Build GRCh38
rs1015212308 Chr.8: 23430528-23430528 on Build GRCh38
rs386723571 Chr.8: 23430857-23430866 on Build GRCh38
rs113850166 Chr.8: 23430870-23430870 on Build GRCh38
rs1044454051 Chr.8: 23430903-23430903 on Build GRCh38
rs998810724 Chr.8: 23430553-23430553 on Build GRCh38
rs895794963 Chr.8: 23430894-23430894 on Build GRCh38
rs867012143 Chr.8: 23430866-23430866 on Build GRCh38
rs61745334 Chr.8: 23430904-23430904 on Build GRCh38
rs542476860 Chr.8: 23430863-23430866 on Build GRCh38
rs774117678 Chr.8: 23430942-23430942 on Build GRCh38
rs938615454 Chr.8: 23430763-23430763 on Build GRCh38
rs1053242673 Chr.8: 23430559-23430559 on Build GRCh38
rs945727153 Chr.8: 23430718-23430718 on Build GRCh38
rs114374719 Chr.8: 23430503-23430503 on Build GRCh38
rs914350080 Chr.8: 23430948-23430948 on Build GRCh38
rs758222074 Chr.8: 23430487-23430487 on Build GRCh38
rs748188562 Chr.8: 23430861-23430861 on Build GRCh38
rs914320606 Chr.8: 23430716-23430716 on Build GRCh38
rs976503946 Chr.8: 23430775-23430775 on Build GRCh38
rs1004907008 Chr.8: 23430872-23430872 on Build GRCh38
rs528687506 Chr.8: 23430856-23430860 on Build GRCh38
rs780606863 Chr.8: 23430919-23430919 on Build GRCh38
rs773660994 Chr.8: 23430889-23430889 on Build GRCh38
rs780475570 Chr.8: 23430726-23430726 on Build GRCh38
rs1027940209 Chr.8: 23430955-23430955 on Build GRCh38
rs966683926 Chr.8: 23430576-23430576 on Build GRCh38
rs144302827 Chr.8: 23430855-23430855 on Build GRCh38
rs142296309 Chr.8: 23430898-23430898 on Build GRCh38
rs764817379 Chr.8: 23430622-23430624 on Build GRCh38
rs932587968 Chr.8: 23430483-23430483 on Build GRCh38
rs994998886 Chr.8: 23430941-23430941 on Build GRCh38
rs757073532 Chr.8: 23430612-23430612 on Build GRCh38
rs188759094 Chr.8: 23430507-23430507 on Build GRCh38
rs995097176 Chr.8: 23430704-23430704 on Build GRCh38
rs1042925782 Chr.8: 23430486-23430486 on Build GRCh38
rs61744075 Chr.8: 23430833-23430833 on Build GRCh38
rs544765869 Chr.8: 23430849-23430849 on Build GRCh38
rs1014448616 Chr.8: 23430594-23430594 on Build GRCh38

View Full Product Details