Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 23432397-23432908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD4
Entrez Gene ID
9583
Gene Name
ectonucleoside triphosphate diphosphohydrolase 4
Gene Aliases
LALP70, LAP70, LYSAL1, NTPDase-4, UDPase
UniGene
Hs.444389
Gene Chromosome Location
Chr.8: 23429152-23457731 on Build GRCh38
Transcripts
NM_001128930.2 NM_004901.4

SNP Detail

SNP ID Chromosome Location
rs574269454 Chr.8: 23432656-23432656 on Build GRCh38
rs1024279348 Chr.8: 23432749-23432749 on Build GRCh38
rs1050736891 Chr.8: 23432680-23432680 on Build GRCh38
rs530301042 Chr.8: 23432821-23432821 on Build GRCh38
rs534610800 Chr.8: 23432468-23432468 on Build GRCh38
rs937086794 Chr.8: 23432697-23432697 on Build GRCh38
rs541653918 Chr.8: 23432684-23432684 on Build GRCh38
rs532843241 Chr.8: 23432499-23432499 on Build GRCh38
rs370603221 Chr.8: 23432537-23432537 on Build GRCh38
rs185704643 Chr.8: 23432650-23432650 on Build GRCh38
rs969993940 Chr.8: 23432750-23432750 on Build GRCh38
rs398073241 Chr.8: 23432517-23432518 on Build GRCh38
rs1022179859 Chr.8: 23432734-23432734 on Build GRCh38
rs60497420 Chr.8: 23432791-23432791 on Build GRCh38
rs575808542 Chr.8: 23432794-23432794 on Build GRCh38
rs981501804 Chr.8: 23432691-23432691 on Build GRCh38
rs189128330 Chr.8: 23432611-23432611 on Build GRCh38
rs889456576 Chr.8: 23432838-23432838 on Build GRCh38
rs144973738 Chr.8: 23432740-23432740 on Build GRCh38
rs548817345 Chr.8: 23432878-23432878 on Build GRCh38
rs111851628 Chr.8: 23432568-23432568 on Build GRCh38
rs540215421 Chr.8: 23432741-23432741 on Build GRCh38
rs937186783 Chr.8: 23432487-23432487 on Build GRCh38
rs561009712 Chr.8: 23432725-23432725 on Build GRCh38
rs958320126 Chr.8: 23432810-23432810 on Build GRCh38
rs61390254 Chr.8: 23432790-23432790 on Build GRCh38
rs928262203 Chr.8: 23432692-23432692 on Build GRCh38
rs981453592 Chr.8: 23432738-23432738 on Build GRCh38
rs906944120 Chr.8: 23432480-23432480 on Build GRCh38
rs1032136635 Chr.8: 23432530-23432530 on Build GRCh38
rs896895719 Chr.8: 23432644-23432644 on Build GRCh38
rs34507802 Chr.8: 23432516-23432517 on Build GRCh38
rs550854645 Chr.8: 23432524-23432524 on Build GRCh38
rs113957461 Chr.8: 23432484-23432484 on Build GRCh38
rs569480085 Chr.8: 23432800-23432800 on Build GRCh38
rs369303343 Chr.8: 23432517-23432517 on Build GRCh38
rs958167915 Chr.8: 23432630-23432630 on Build GRCh38
rs889425339 Chr.8: 23432720-23432720 on Build GRCh38
rs988771783 Chr.8: 23432439-23432439 on Build GRCh38
rs4872135 Chr.8: 23432640-23432640 on Build GRCh38
rs368575254 Chr.8: 23432744-23432744 on Build GRCh38
rs917530309 Chr.8: 23432564-23432564 on Build GRCh38
rs565289231 Chr.8: 23432788-23432788 on Build GRCh38
rs769692133 Chr.8: 23432860-23432861 on Build GRCh38
rs33928899 Chr.8: 23432511-23432512 on Build GRCh38
rs1050807690 Chr.8: 23432830-23432830 on Build GRCh38
rs887581121 Chr.8: 23432498-23432498 on Build GRCh38
rs1138445 Chr.8: 23432567-23432567 on Build GRCh38
rs958240075 Chr.8: 23432789-23432789 on Build GRCh38
rs903690643 Chr.8: 23432739-23432739 on Build GRCh38
rs978870643 Chr.8: 23432503-23432503 on Build GRCh38
rs933694965 Chr.8: 23432665-23432665 on Build GRCh38
rs1055518476 Chr.8: 23432706-23432706 on Build GRCh38
rs866205253 Chr.8: 23432612-23432612 on Build GRCh38
rs1013256827 Chr.8: 23432551-23432551 on Build GRCh38
rs980330144 Chr.8: 23432753-23432753 on Build GRCh38
rs577019991 Chr.8: 23432639-23432639 on Build GRCh38
rs1055654513 Chr.8: 23432497-23432497 on Build GRCh38
rs933748231 Chr.8: 23432809-23432809 on Build GRCh38
rs11403204 Chr.8: 23432504-23432505 on Build GRCh38
rs971837745 Chr.8: 23432729-23432729 on Build GRCh38
rs1006756497 Chr.8: 23432467-23432467 on Build GRCh38
rs1030443146 Chr.8: 23432610-23432610 on Build GRCh38
rs942307847 Chr.8: 23432866-23432866 on Build GRCh38
rs867954708 Chr.8: 23432495-23432495 on Build GRCh38
rs55894164 Chr.8: 23432538-23432538 on Build GRCh38
rs558547175 Chr.8: 23432619-23432619 on Build GRCh38
rs908953004 Chr.8: 23432868-23432868 on Build GRCh38
rs992846355 Chr.8: 23432745-23432745 on Build GRCh38

View Full Product Details