Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 24400054-24400551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMDEC1
Entrez Gene ID
27299
Gene Name
ADAM like decysin 1
Gene Aliases
M12.219
UniGene
Hs.521459
Gene Chromosome Location
Chr.8: 24384285-24406013 on Build GRCh38
Transcripts
NM_001145271.1 NM_001145272.1 NM_014479.3 NR_125808.1

Gene Symbol
LOC101929294
Entrez Gene ID
101929294
Gene Name
uncharacterized LOC101929294
Gene Aliases
-
UniGene
Hs.637673
Gene Chromosome Location
Chr.8: 24295814-24548618 on Build GRCh38
Transcripts
NM_001145271.1 NM_001145272.1 NM_014479.3 NR_125808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4413385 c.1121G>A p.C374Y Chr.8: 24400279-24400279 on Build GRCh38
5874422 c.1072C>T p.L358F Chr.8: 24400230-24400230 on Build GRCh38
1699842 c.1066C>T p.H356Y Chr.8: 24400224-24400224 on Build GRCh38
1699843 c.1108C>T p.P370S Chr.8: 24400266-24400266 on Build GRCh38
3647508 c.1062G>C p.L354L Chr.8: 24400220-24400220 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74769414 Chr.8: 24400495-24400495 on Build GRCh38
rs766276874 Chr.8: 24400227-24400227 on Build GRCh38
rs756613291 Chr.8: 24400145-24400145 on Build GRCh38
rs1023474066 Chr.8: 24400433-24400433 on Build GRCh38
rs757694534 Chr.8: 24400314-24400314 on Build GRCh38
rs569750220 Chr.8: 24400106-24400106 on Build GRCh38
rs76855193 Chr.8: 24400422-24400422 on Build GRCh38
rs752770055 Chr.8: 24400136-24400136 on Build GRCh38
rs756461025 Chr.8: 24400259-24400260 on Build GRCh38
rs896334628 Chr.8: 24400354-24400354 on Build GRCh38
rs757797434 Chr.8: 24400172-24400173 on Build GRCh38
rs764070825 Chr.8: 24400292-24400292 on Build GRCh38
rs746670007 Chr.8: 24400170-24400170 on Build GRCh38
rs1019711019 Chr.8: 24400509-24400509 on Build GRCh38
rs113981898 Chr.8: 24400157-24400157 on Build GRCh38
rs774441890 Chr.8: 24400345-24400345 on Build GRCh38
rs773340785 Chr.8: 24400223-24400223 on Build GRCh38
rs779245918 Chr.8: 24400165-24400165 on Build GRCh38
rs368913512 Chr.8: 24400192-24400192 on Build GRCh38
rs139397711 Chr.8: 24400196-24400196 on Build GRCh38
rs756269376 Chr.8: 24400276-24400276 on Build GRCh38
rs781693183 Chr.8: 24400173-24400173 on Build GRCh38
rs772777693 Chr.8: 24400343-24400343 on Build GRCh38
rs902292869 Chr.8: 24400088-24400088 on Build GRCh38
rs976231690 Chr.8: 24400180-24400180 on Build GRCh38
rs576052612 Chr.8: 24400351-24400351 on Build GRCh38
rs76067849 Chr.8: 24400418-24400418 on Build GRCh38
rs770718644 Chr.8: 24400217-24400217 on Build GRCh38
rs201372881 Chr.8: 24400344-24400345 on Build GRCh38
rs896298316 Chr.8: 24400362-24400362 on Build GRCh38
rs975870011 Chr.8: 24400518-24400518 on Build GRCh38
rs185404726 Chr.8: 24400507-24400507 on Build GRCh38
rs370138921 Chr.8: 24400320-24400320 on Build GRCh38
rs530585014 Chr.8: 24400115-24400115 on Build GRCh38
rs779264150 Chr.8: 24400316-24400316 on Build GRCh38
rs759857391 Chr.8: 24400253-24400253 on Build GRCh38
rs752025567 Chr.8: 24400171-24400172 on Build GRCh38
rs73217092 Chr.8: 24400463-24400463 on Build GRCh38
rs143506329 Chr.8: 24400335-24400335 on Build GRCh38
rs112171746 Chr.8: 24400454-24400454 on Build GRCh38
rs995214181 Chr.8: 24400469-24400469 on Build GRCh38
rs896393209 Chr.8: 24400114-24400114 on Build GRCh38
rs780137361 Chr.8: 24400305-24400310 on Build GRCh38
rs1025847361 Chr.8: 24400477-24400478 on Build GRCh38
rs73223116 Chr.8: 24400466-24400466 on Build GRCh38
rs555841858 Chr.8: 24400383-24400383 on Build GRCh38
rs75758866 Chr.8: 24400496-24400496 on Build GRCh38
rs375768004 Chr.8: 24400166-24400166 on Build GRCh38
rs754412982 Chr.8: 24400307-24400307 on Build GRCh38
rs749428341 Chr.8: 24400311-24400313 on Build GRCh38
rs78290629 Chr.8: 24400431-24400431 on Build GRCh38
rs144434383 Chr.8: 24400205-24400205 on Build GRCh38
rs374876945 Chr.8: 24400160-24400160 on Build GRCh38
rs372619362 Chr.8: 24400201-24400201 on Build GRCh38
rs962422739 Chr.8: 24400137-24400137 on Build GRCh38
rs140087766 Chr.8: 24400255-24400255 on Build GRCh38
rs1015001597 Chr.8: 24400163-24400163 on Build GRCh38
rs796737888 Chr.8: 24400414-24400414 on Build GRCh38
rs566061544 Chr.8: 24400332-24400332 on Build GRCh38
rs758658062 Chr.8: 24400325-24400325 on Build GRCh38
rs902241865 Chr.8: 24400328-24400328 on Build GRCh38
rs764707283 Chr.8: 24400142-24400145 on Build GRCh38
rs138026723 Chr.8: 24400156-24400156 on Build GRCh38
rs1034839578 Chr.8: 24400113-24400113 on Build GRCh38
rs568514008 Chr.8: 24400159-24400159 on Build GRCh38
rs746174250 Chr.8: 24400173-24400176 on Build GRCh38
rs749247332 Chr.8: 24400346-24400346 on Build GRCh38
rs762978844 Chr.8: 24400225-24400225 on Build GRCh38
rs768299279 Chr.8: 24400177-24400177 on Build GRCh38
rs543506887 Chr.8: 24400366-24400366 on Build GRCh38
rs1006219710 Chr.8: 24400494-24400494 on Build GRCh38
rs146111288 Chr.8: 24400243-24400243 on Build GRCh38
rs781142368 Chr.8: 24400119-24400119 on Build GRCh38
rs952041944 Chr.8: 24400126-24400126 on Build GRCh38
rs898119230 Chr.8: 24400398-24400398 on Build GRCh38
rs914832557 Chr.8: 24400264-24400264 on Build GRCh38
rs116311320 Chr.8: 24400339-24400339 on Build GRCh38
rs964198293 Chr.8: 24400512-24400512 on Build GRCh38

View Full Product Details