Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144373710-144374212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCK5
Entrez Gene ID
203054
Gene Name
aarF domain containing kinase 5
Gene Aliases
-
UniGene
Hs.283374
Gene Chromosome Location
Chr.8: 144373762-144393242 on Build GRCh38
Transcripts
NM_174922.4 XM_006716527.2 XM_011516910.1 XM_017013174.1

SNP Detail

SNP ID Chromosome Location
rs558653477 Chr.8: 144373831-144373831 on Build GRCh38
rs552201418 Chr.8: 144374051-144374051 on Build GRCh38
rs575992251 Chr.8: 144373813-144373813 on Build GRCh38
rs542074411 Chr.8: 144373954-144373954 on Build GRCh38
rs923387638 Chr.8: 144374167-144374167 on Build GRCh38
rs782667107 Chr.8: 144373933-144373933 on Build GRCh38
rs782066399 Chr.8: 144373744-144373744 on Build GRCh38
rs961715096 Chr.8: 144374025-144374025 on Build GRCh38
rs946049328 Chr.8: 144374141-144374141 on Build GRCh38
rs189058493 Chr.8: 144373794-144373794 on Build GRCh38
rs535601480 Chr.8: 144373796-144373796 on Build GRCh38
rs781886405 Chr.8: 144374123-144374123 on Build GRCh38
rs782494221 Chr.8: 144373836-144373836 on Build GRCh38
rs891890643 Chr.8: 144373739-144373739 on Build GRCh38
rs201116950 Chr.8: 144374091-144374091 on Build GRCh38
rs782581749 Chr.8: 144373840-144373840 on Build GRCh38
rs1040506357 Chr.8: 144373917-144373917 on Build GRCh38
rs377556188 Chr.8: 144374052-144374052 on Build GRCh38
rs961766082 Chr.8: 144373819-144373819 on Build GRCh38
rs782287073 Chr.8: 144374053-144374053 on Build GRCh38
rs374055237 Chr.8: 144374092-144374092 on Build GRCh38
rs782818551 Chr.8: 144374116-144374116 on Build GRCh38
rs779225941 Chr.8: 144374082-144374082 on Build GRCh38
rs114819450 Chr.8: 144373871-144373871 on Build GRCh38
rs996477940 Chr.8: 144373782-144373782 on Build GRCh38
rs374533538 Chr.8: 144374083-144374083 on Build GRCh38
rs370185857 Chr.8: 144374057-144374057 on Build GRCh38
rs946101622 Chr.8: 144373911-144373911 on Build GRCh38
rs780733556 Chr.8: 144373932-144373932 on Build GRCh38
rs781861549 Chr.8: 144373829-144373829 on Build GRCh38
rs7836258 Chr.8: 144373902-144373902 on Build GRCh38
rs901706769 Chr.8: 144373762-144373762 on Build GRCh38
rs1006132262 Chr.8: 144373957-144373957 on Build GRCh38
rs927578465 Chr.8: 144373865-144373865 on Build GRCh38
rs549494748 Chr.8: 144373820-144373820 on Build GRCh38
rs200435576 Chr.8: 144374104-144374104 on Build GRCh38
rs111335406 Chr.8: 144373907-144373907 on Build GRCh38
rs1014587427 Chr.8: 144373811-144373811 on Build GRCh38
rs971940695 Chr.8: 144374079-144374079 on Build GRCh38
rs1014535148 Chr.8: 144374019-144374019 on Build GRCh38
rs781902194 Chr.8: 144373758-144373758 on Build GRCh38
rs1054512636 Chr.8: 144373736-144373736 on Build GRCh38
rs980735137 Chr.8: 144373848-144373848 on Build GRCh38
rs563560822 Chr.8: 144373967-144373967 on Build GRCh38
rs536902419 Chr.8: 144373793-144373793 on Build GRCh38
rs782505446 Chr.8: 144374142-144374142 on Build GRCh38
rs782708891 Chr.8: 144373752-144373752 on Build GRCh38
rs566317690 Chr.8: 144373928-144373928 on Build GRCh38
rs991902325 Chr.8: 144373822-144373822 on Build GRCh38
rs931768980 Chr.8: 144374189-144374189 on Build GRCh38
rs1024742969 Chr.8: 144374059-144374059 on Build GRCh38
rs782512920 Chr.8: 144374152-144374152 on Build GRCh38
rs184057021 Chr.8: 144374085-144374085 on Build GRCh38
rs530552169 Chr.8: 144373990-144373990 on Build GRCh38
rs7845652 Chr.8: 144373823-144373823 on Build GRCh38
rs574667270 Chr.8: 144373930-144373930 on Build GRCh38

View Full Product Details