Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144379295-144379797 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCK5
Entrez Gene ID
203054
Gene Name
aarF domain containing kinase 5
Gene Aliases
-
UniGene
Hs.283374
Gene Chromosome Location
Chr.8: 144373762-144393242 on Build GRCh38
Transcripts
NM_174922.4 XM_006716527.2 XM_011516907.2 XM_011516909.1 XM_011516910.1 XM_017013174.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5874199 c.16C>T p.Q6* Chr.8: 144379390-144379390 on Build GRCh38
7303458 c.19C>G p.L7V Chr.8: 144379393-144379393 on Build GRCh38
4659850 c.68C>A p.P23Q Chr.8: 144379442-144379442 on Build GRCh38
5455652 c.20T>G p.L7R Chr.8: 144379394-144379394 on Build GRCh38
1623772 c.18G>A p.Q6Q Chr.8: 144379392-144379392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782143364 Chr.8: 144379383-144379383 on Build GRCh38
rs373379221 Chr.8: 144379510-144379510 on Build GRCh38
rs149682358 Chr.8: 144379471-144379471 on Build GRCh38
rs782754087 Chr.8: 144379496-144379496 on Build GRCh38
rs782028470 Chr.8: 144379369-144379369 on Build GRCh38
rs368263421 Chr.8: 144379340-144379340 on Build GRCh38
rs782813225 Chr.8: 144379435-144379435 on Build GRCh38
rs961634883 Chr.8: 144379469-144379469 on Build GRCh38
rs563557453 Chr.8: 144379569-144379569 on Build GRCh38
rs559274510 Chr.8: 144379641-144379641 on Build GRCh38
rs6599528 Chr.8: 144379425-144379425 on Build GRCh38
rs782608476 Chr.8: 144379516-144379516 on Build GRCh38
rs558045909 Chr.8: 144379359-144379359 on Build GRCh38
rs782658587 Chr.8: 144379353-144379353 on Build GRCh38
rs561033526 Chr.8: 144379500-144379500 on Build GRCh38
rs782456416 Chr.8: 144379450-144379450 on Build GRCh38
rs548332299 Chr.8: 144379666-144379666 on Build GRCh38
rs377009134 Chr.8: 144379498-144379498 on Build GRCh38
rs547082110 Chr.8: 144379636-144379636 on Build GRCh38
rs1028657021 Chr.8: 144379744-144379744 on Build GRCh38
rs782723994 Chr.8: 144379423-144379423 on Build GRCh38
rs543211958 Chr.8: 144379588-144379588 on Build GRCh38
rs782303335 Chr.8: 144379363-144379363 on Build GRCh38
rs532396240 Chr.8: 144379605-144379605 on Build GRCh38
rs145681525 Chr.8: 144379424-144379424 on Build GRCh38
rs1010599370 Chr.8: 144379338-144379338 on Build GRCh38
rs375809099 Chr.8: 144379342-144379342 on Build GRCh38
rs782564884 Chr.8: 144379452-144379452 on Build GRCh38
rs373989215 Chr.8: 144379495-144379495 on Build GRCh38
rs781890167 Chr.8: 144379662-144379662 on Build GRCh38
rs138199884 Chr.8: 144379649-144379649 on Build GRCh38
rs565075940 Chr.8: 144379594-144379594 on Build GRCh38
rs140712091 Chr.8: 144379470-144379470 on Build GRCh38
rs782515775 Chr.8: 144379773-144379773 on Build GRCh38
rs910072985 Chr.8: 144379445-144379445 on Build GRCh38
rs781952279 Chr.8: 144379478-144379478 on Build GRCh38
rs782548791 Chr.8: 144379506-144379506 on Build GRCh38
rs372533252 Chr.8: 144379341-144379341 on Build GRCh38
rs782392323 Chr.8: 144379387-144379387 on Build GRCh38
rs369986202 Chr.8: 144379501-144379501 on Build GRCh38
rs781981788 Chr.8: 144379391-144379391 on Build GRCh38
rs782756308 Chr.8: 144379490-144379490 on Build GRCh38
rs782208653 Chr.8: 144379524-144379524 on Build GRCh38
rs780512982 Chr.8: 144379591-144379591 on Build GRCh38
rs782728021 Chr.8: 144379552-144379552 on Build GRCh38
rs369601211 Chr.8: 144379477-144379477 on Build GRCh38
rs901485121 Chr.8: 144379724-144379724 on Build GRCh38
rs782287690 Chr.8: 144379465-144379465 on Build GRCh38
rs554086624 Chr.8: 144379446-144379446 on Build GRCh38
rs782362808 Chr.8: 144379358-144379358 on Build GRCh38
rs755768626 Chr.8: 144379364-144379364 on Build GRCh38
rs551600122 Chr.8: 144379442-144379442 on Build GRCh38
rs116317547 Chr.8: 144379479-144379479 on Build GRCh38
rs782789464 Chr.8: 144379443-144379443 on Build GRCh38
rs782535591 Chr.8: 144379350-144379350 on Build GRCh38
rs782790319 Chr.8: 144379507-144379507 on Build GRCh38
rs782426516 Chr.8: 144379480-144379480 on Build GRCh38
rs891705051 Chr.8: 144379333-144379333 on Build GRCh38
rs7007386 Chr.8: 144379543-144379543 on Build GRCh38
rs375853047 Chr.8: 144379608-144379608 on Build GRCh38
rs569763960 Chr.8: 144379675-144379675 on Build GRCh38
rs936109646 Chr.8: 144379596-144379596 on Build GRCh38
rs556942945 Chr.8: 144379400-144379400 on Build GRCh38
rs538480806 Chr.8: 144379697-144379697 on Build GRCh38
rs550306761 Chr.8: 144379733-144379733 on Build GRCh38
rs782029764 Chr.8: 144379514-144379515 on Build GRCh38
rs538420037 Chr.8: 144379355-144379355 on Build GRCh38
rs782400800 Chr.8: 144379468-144379468 on Build GRCh38

View Full Product Details