Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144390581-144391071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCK5
Entrez Gene ID
203054
Gene Name
aarF domain containing kinase 5
Gene Aliases
-
UniGene
Hs.283374
Gene Chromosome Location
Chr.8: 144373762-144393242 on Build GRCh38
Transcripts
NM_174922.4 XM_006716527.2 XM_011516907.2 XM_011516909.1 XM_011516910.1 XM_017013174.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5874200 c.280G>A p.G94S Chr.8: 144390684-144390684 on Build GRCh38
3646575 c.293C>T p.S98F Chr.8: 144390697-144390697 on Build GRCh38
1313857 c.452G>A p.G151E Chr.8: 144390965-144390965 on Build GRCh38
4427754 c.360G>A p.L120L Chr.8: 144390873-144390873 on Build GRCh38
6594822 c.281G>A p.G94D Chr.8: 144390685-144390685 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781960393 Chr.8: 144390654-144390654 on Build GRCh38
rs782794261 Chr.8: 144390689-144390689 on Build GRCh38
rs111509485 Chr.8: 144390623-144390623 on Build GRCh38
rs782186573 Chr.8: 144390844-144390844 on Build GRCh38
rs782690978 Chr.8: 144390719-144390719 on Build GRCh38
rs372634320 Chr.8: 144390625-144390625 on Build GRCh38
rs782554498 Chr.8: 144390936-144390937 on Build GRCh38
rs782554130 Chr.8: 144390948-144390948 on Build GRCh38
rs782276754 Chr.8: 144390859-144390859 on Build GRCh38
rs375370922 Chr.8: 144390621-144390621 on Build GRCh38
rs782609549 Chr.8: 144390698-144390698 on Build GRCh38
rs567194058 Chr.8: 144390786-144390786 on Build GRCh38
rs782044294 Chr.8: 144390744-144390744 on Build GRCh38
rs560973535 Chr.8: 144390684-144390684 on Build GRCh38
rs782285425 Chr.8: 144390727-144390727 on Build GRCh38
rs200877552 Chr.8: 144390767-144390767 on Build GRCh38
rs782813437 Chr.8: 144390887-144390887 on Build GRCh38
rs141834717 Chr.8: 144390695-144390695 on Build GRCh38
rs577667370 Chr.8: 144391015-144391015 on Build GRCh38
rs143148314 Chr.8: 144391018-144391018 on Build GRCh38
rs549893722 Chr.8: 144390722-144390722 on Build GRCh38
rs782811188 Chr.8: 144390746-144390746 on Build GRCh38
rs782590743 Chr.8: 144391010-144391010 on Build GRCh38
rs772772352 Chr.8: 144390741-144390741 on Build GRCh38
rs148075370 Chr.8: 144390691-144390691 on Build GRCh38
rs370821655 Chr.8: 144390997-144390997 on Build GRCh38
rs544823046 Chr.8: 144391016-144391016 on Build GRCh38
rs782166065 Chr.8: 144391035-144391035 on Build GRCh38
rs782698075 Chr.8: 144390787-144390787 on Build GRCh38
rs374040823 Chr.8: 144390846-144390846 on Build GRCh38
rs782022775 Chr.8: 144391012-144391012 on Build GRCh38
rs782771261 Chr.8: 144390645-144390645 on Build GRCh38
rs782447125 Chr.8: 144390718-144390718 on Build GRCh38
rs377092408 Chr.8: 144390641-144390641 on Build GRCh38
rs781910730 Chr.8: 144390789-144390789 on Build GRCh38
rs782088343 Chr.8: 144390863-144390863 on Build GRCh38
rs782137436 Chr.8: 144390996-144390996 on Build GRCh38
rs537850893 Chr.8: 144391006-144391006 on Build GRCh38
rs782669473 Chr.8: 144390990-144390991 on Build GRCh38
rs201895535 Chr.8: 144390629-144390629 on Build GRCh38
rs199662782 Chr.8: 144390659-144390659 on Build GRCh38
rs782449120 Chr.8: 144390928-144390928 on Build GRCh38
rs781969865 Chr.8: 144390862-144390862 on Build GRCh38
rs782122488 Chr.8: 144390902-144390902 on Build GRCh38
rs202165926 Chr.8: 144390927-144390927 on Build GRCh38
rs782030500 Chr.8: 144390850-144390850 on Build GRCh38
rs76866583 Chr.8: 144391004-144391004 on Build GRCh38
rs375841571 Chr.8: 144390903-144390903 on Build GRCh38
rs782655815 Chr.8: 144390807-144390807 on Build GRCh38
rs1056897366 Chr.8: 144390802-144390802 on Build GRCh38
rs896552414 Chr.8: 144390803-144390803 on Build GRCh38
rs1039196082 Chr.8: 144390665-144390665 on Build GRCh38
rs930798308 Chr.8: 144391038-144391038 on Build GRCh38
rs782715520 Chr.8: 144390667-144390667 on Build GRCh38
rs922058100 Chr.8: 144391034-144391034 on Build GRCh38
rs781824311 Chr.8: 144390773-144390773 on Build GRCh38
rs782398356 Chr.8: 144390732-144390732 on Build GRCh38
rs908107075 Chr.8: 144391052-144391052 on Build GRCh38
rs782222826 Chr.8: 144390739-144390739 on Build GRCh38
rs142025248 Chr.8: 144391042-144391042 on Build GRCh38
rs782014013 Chr.8: 144390753-144390753 on Build GRCh38
rs556660351 Chr.8: 144391007-144391007 on Build GRCh38
rs782541590 Chr.8: 144390791-144390791 on Build GRCh38
rs781860670 Chr.8: 144390639-144390639 on Build GRCh38
rs199992416 Chr.8: 144390622-144390622 on Build GRCh38
rs782388246 Chr.8: 144390860-144390860 on Build GRCh38
rs782131015 Chr.8: 144390761-144390761 on Build GRCh38
rs781784683 Chr.8: 144391033-144391033 on Build GRCh38
rs782787009 Chr.8: 144390981-144390981 on Build GRCh38
rs375059392 Chr.8: 144391017-144391017 on Build GRCh38
rs781873553 Chr.8: 144390888-144390888 on Build GRCh38
rs56192724 Chr.8: 144390740-144390740 on Build GRCh38
rs62530407 Chr.8: 144390870-144390870 on Build GRCh38
rs543742812 Chr.8: 144390839-144390839 on Build GRCh38
rs577463762 Chr.8: 144390610-144390610 on Build GRCh38
rs571243432 Chr.8: 144390736-144390736 on Build GRCh38
rs782193011 Chr.8: 144390715-144390715 on Build GRCh38
rs782074577 Chr.8: 144390666-144390666 on Build GRCh38
rs781903388 Chr.8: 144390980-144390980 on Build GRCh38
rs781790317 Chr.8: 144390671-144390671 on Build GRCh38
rs557916587 Chr.8: 144390617-144390617 on Build GRCh38
rs781865438 Chr.8: 144391043-144391043 on Build GRCh38

View Full Product Details