Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144391498-144392003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCK5
Entrez Gene ID
203054
Gene Name
aarF domain containing kinase 5
Gene Aliases
-
UniGene
Hs.283374
Gene Chromosome Location
Chr.8: 144373762-144393242 on Build GRCh38
Transcripts
NM_174922.4 XM_006716527.2 XM_011516907.2 XM_011516909.1 XM_011516910.1 XM_017013174.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1455619 c.809G>T p.G270V Chr.8: 144391590-144391590 on Build GRCh38
4594338 c.905G>A p.R302H Chr.8: 144391686-144391686 on Build GRCh38
5441796 c.900G>A p.V300V Chr.8: 144391681-144391681 on Build GRCh38
99007 c.971A>G p.N324S Chr.8: 144391823-144391823 on Build GRCh38
5824669 c.1016T>C p.I339T Chr.8: 144391942-144391942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782356500 Chr.8: 144391898-144391898 on Build GRCh38
rs2928372 Chr.8: 144391895-144391895 on Build GRCh38
rs199991427 Chr.8: 144391823-144391823 on Build GRCh38
rs782698053 Chr.8: 144391566-144391566 on Build GRCh38
rs144396697 Chr.8: 144391648-144391648 on Build GRCh38
rs782072777 Chr.8: 144391564-144391564 on Build GRCh38
rs992370425 Chr.8: 144391615-144391615 on Build GRCh38
rs567968893 Chr.8: 144391952-144391952 on Build GRCh38
rs201576165 Chr.8: 144391889-144391889 on Build GRCh38
rs782477867 Chr.8: 144391785-144391785 on Build GRCh38
rs782584097 Chr.8: 144391892-144391892 on Build GRCh38
rs782404445 Chr.8: 144391875-144391875 on Build GRCh38
rs532129022 Chr.8: 144391883-144391883 on Build GRCh38
rs782077212 Chr.8: 144391874-144391875 on Build GRCh38
rs782100032 Chr.8: 144391881-144391881 on Build GRCh38
rs980714753 Chr.8: 144391683-144391683 on Build GRCh38
rs782683304 Chr.8: 144391941-144391941 on Build GRCh38
rs537802011 Chr.8: 144391765-144391765 on Build GRCh38
rs781853926 Chr.8: 144391574-144391574 on Build GRCh38
rs782081313 Chr.8: 144391856-144391856 on Build GRCh38
rs1026624607 Chr.8: 144391837-144391837 on Build GRCh38
rs554856977 Chr.8: 144391751-144391751 on Build GRCh38
rs781800832 Chr.8: 144391930-144391930 on Build GRCh38
rs367748430 Chr.8: 144391563-144391563 on Build GRCh38
rs542447834 Chr.8: 144391749-144391749 on Build GRCh38
rs782182631 Chr.8: 144391797-144391797 on Build GRCh38
rs549271923 Chr.8: 144391904-144391904 on Build GRCh38
rs959843937 Chr.8: 144391906-144391906 on Build GRCh38
rs369711044 Chr.8: 144391766-144391766 on Build GRCh38
rs782636582 Chr.8: 144391758-144391758 on Build GRCh38
rs781926584 Chr.8: 144391670-144391670 on Build GRCh38
rs782491985 Chr.8: 144391917-144391917 on Build GRCh38
rs782607023 Chr.8: 144391602-144391602 on Build GRCh38
rs782627190 Chr.8: 144391973-144391973 on Build GRCh38
rs782379034 Chr.8: 144391641-144391641 on Build GRCh38
rs782120255 Chr.8: 144391553-144391553 on Build GRCh38
rs781823605 Chr.8: 144391735-144391735 on Build GRCh38
rs1052952798 Chr.8: 144391747-144391747 on Build GRCh38
rs201908526 Chr.8: 144391597-144391597 on Build GRCh38
rs201284257 Chr.8: 144391890-144391890 on Build GRCh38
rs782252525 Chr.8: 144391811-144391811 on Build GRCh38
rs564840942 Chr.8: 144391847-144391847 on Build GRCh38
rs782000085 Chr.8: 144391894-144391894 on Build GRCh38
rs888111567 Chr.8: 144391852-144391852 on Build GRCh38
rs782801751 Chr.8: 144391682-144391682 on Build GRCh38
rs782191036 Chr.8: 144391624-144391624 on Build GRCh38
rs201101725 Chr.8: 144391629-144391629 on Build GRCh38
rs782766955 Chr.8: 144391562-144391562 on Build GRCh38
rs782266725 Chr.8: 144391640-144391640 on Build GRCh38
rs1040928358 Chr.8: 144391777-144391777 on Build GRCh38
rs199613537 Chr.8: 144391547-144391547 on Build GRCh38
rs1008107901 Chr.8: 144391518-144391518 on Build GRCh38
rs782280570 Chr.8: 144391945-144391945 on Build GRCh38
rs370557280 Chr.8: 144391899-144391899 on Build GRCh38
rs372024693 Chr.8: 144391796-144391796 on Build GRCh38
rs781846962 Chr.8: 144391915-144391915 on Build GRCh38
rs781920182 Chr.8: 144391861-144391861 on Build GRCh38
rs962517778 Chr.8: 144391601-144391601 on Build GRCh38
rs200670250 Chr.8: 144391687-144391687 on Build GRCh38
rs782326417 Chr.8: 144391653-144391653 on Build GRCh38
rs782419291 Chr.8: 144391893-144391893 on Build GRCh38
rs782477111 Chr.8: 144391591-144391591 on Build GRCh38
rs925248579 Chr.8: 144391951-144391951 on Build GRCh38
rs541157890 Chr.8: 144391888-144391888 on Build GRCh38
rs1003920382 Chr.8: 144391867-144391867 on Build GRCh38
rs925299108 Chr.8: 144391686-144391686 on Build GRCh38
rs782407702 Chr.8: 144391530-144391530 on Build GRCh38
rs543428878 Chr.8: 144391844-144391844 on Build GRCh38
rs782770887 Chr.8: 144391914-144391914 on Build GRCh38
rs571420747 Chr.8: 144391572-144391572 on Build GRCh38
rs782107766 Chr.8: 144391893-144391894 on Build GRCh38
rs781813814 Chr.8: 144391882-144391882 on Build GRCh38
rs781855058 Chr.8: 144391876-144391876 on Build GRCh38
rs782650117 Chr.8: 144391800-144391800 on Build GRCh38
rs782716786 Chr.8: 144391918-144391918 on Build GRCh38
rs781975682 Chr.8: 144391647-144391647 on Build GRCh38
rs781901091 Chr.8: 144391567-144391567 on Build GRCh38
rs188530740 Chr.8: 144391976-144391976 on Build GRCh38
rs782504254 Chr.8: 144391961-144391961 on Build GRCh38
rs782690182 Chr.8: 144391746-144391746 on Build GRCh38
rs782688623 Chr.8: 144391885-144391885 on Build GRCh38
rs782065481 Chr.8: 144391745-144391745 on Build GRCh38
rs899784860 Chr.8: 144391533-144391533 on Build GRCh38
rs918153788 Chr.8: 144391672-144391672 on Build GRCh38
rs781931033 Chr.8: 144391876-144391877 on Build GRCh38
rs782617934 Chr.8: 144391626-144391626 on Build GRCh38
rs782160430 Chr.8: 144391874-144391874 on Build GRCh38
rs531875170 Chr.8: 144391977-144391977 on Build GRCh38
rs571955758 Chr.8: 144391717-144391717 on Build GRCh38
rs373166971 Chr.8: 144391909-144391909 on Build GRCh38
rs782043580 Chr.8: 144391907-144391907 on Build GRCh38
rs782112833 Chr.8: 144391693-144391693 on Build GRCh38
rs782793602 Chr.8: 144391875-144391875 on Build GRCh38
rs782528655 Chr.8: 144391568-144391568 on Build GRCh38
rs980997052 Chr.8: 144391942-144391942 on Build GRCh38

View Full Product Details