Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144392001-144392476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCK5
Entrez Gene ID
203054
Gene Name
aarF domain containing kinase 5
Gene Aliases
-
UniGene
Hs.283374
Gene Chromosome Location
Chr.8: 144373762-144393242 on Build GRCh38
Transcripts
NM_174922.4 XM_006716527.2 XM_011516907.2 XM_011516909.1 XM_011516910.1 XM_017013174.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6372821 c.1142T>G p.L381R Chr.8: 144392137-144392137 on Build GRCh38
6371690 c.1252G>A p.A418T Chr.8: 144392330-144392330 on Build GRCh38
2871238 c.1077G>A p.S359S Chr.8: 144392003-144392003 on Build GRCh38
1283288 c.1131G>A p.E377E Chr.8: 144392126-144392126 on Build GRCh38
4162844 c.1095C>A p.N365K Chr.8: 144392021-144392021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576302152 Chr.8: 144392353-144392353 on Build GRCh38
rs781890231 Chr.8: 144392261-144392261 on Build GRCh38
rs913923794 Chr.8: 144392283-144392283 on Build GRCh38
rs1040878217 Chr.8: 144392060-144392060 on Build GRCh38
rs782211102 Chr.8: 144392069-144392069 on Build GRCh38
rs782181616 Chr.8: 144392061-144392061 on Build GRCh38
rs375545962 Chr.8: 144392066-144392066 on Build GRCh38
rs574419228 Chr.8: 144392425-144392425 on Build GRCh38
rs782259124 Chr.8: 144392190-144392190 on Build GRCh38
rs782058643 Chr.8: 144392022-144392022 on Build GRCh38
rs200173122 Chr.8: 144392111-144392111 on Build GRCh38
rs782103303 Chr.8: 144392239-144392239 on Build GRCh38
rs770576203 Chr.8: 144392244-144392244 on Build GRCh38
rs782726471 Chr.8: 144392437-144392437 on Build GRCh38
rs373269841 Chr.8: 144392234-144392234 on Build GRCh38
rs782463385 Chr.8: 144392434-144392434 on Build GRCh38
rs536979930 Chr.8: 144392370-144392370 on Build GRCh38
rs781848331 Chr.8: 144392237-144392237 on Build GRCh38
rs782676765 Chr.8: 144392184-144392184 on Build GRCh38
rs782292939 Chr.8: 144392308-144392308 on Build GRCh38
rs781843786 Chr.8: 144392048-144392048 on Build GRCh38
rs781891761 Chr.8: 144392135-144392135 on Build GRCh38
rs541020614 Chr.8: 144392403-144392403 on Build GRCh38
rs782000623 Chr.8: 144392100-144392100 on Build GRCh38
rs1004270977 Chr.8: 144392180-144392180 on Build GRCh38
rs782418809 Chr.8: 144392183-144392183 on Build GRCh38
rs2977837 Chr.8: 144392053-144392053 on Build GRCh38
rs921386378 Chr.8: 144392319-144392319 on Build GRCh38
rs377410012 Chr.8: 144392052-144392052 on Build GRCh38
rs782780780 Chr.8: 144392431-144392431 on Build GRCh38
rs988528660 Chr.8: 144392260-144392260 on Build GRCh38
rs558873265 Chr.8: 144392384-144392384 on Build GRCh38
rs782531911 Chr.8: 144392282-144392282 on Build GRCh38
rs782208902 Chr.8: 144392304-144392304 on Build GRCh38
rs782686342 Chr.8: 144392259-144392259 on Build GRCh38
rs781949362 Chr.8: 144392202-144392202 on Build GRCh38
rs372812691 Chr.8: 144392040-144392040 on Build GRCh38
rs370373406 Chr.8: 144392079-144392079 on Build GRCh38
rs932463619 Chr.8: 144392335-144392335 on Build GRCh38
rs782650161 Chr.8: 144392434-144392435 on Build GRCh38
rs782139226 Chr.8: 144392225-144392225 on Build GRCh38
rs148808398 Chr.8: 144392115-144392115 on Build GRCh38
rs781900039 Chr.8: 144392028-144392028 on Build GRCh38
rs782704643 Chr.8: 144392027-144392027 on Build GRCh38
rs782187418 Chr.8: 144392179-144392179 on Build GRCh38
rs782320085 Chr.8: 144392306-144392306 on Build GRCh38
rs376579859 Chr.8: 144392241-144392241 on Build GRCh38
rs782308307 Chr.8: 144392216-144392216 on Build GRCh38
rs782536544 Chr.8: 144392037-144392037 on Build GRCh38
rs782479798 Chr.8: 144392167-144392167 on Build GRCh38
rs782072521 Chr.8: 144392422-144392422 on Build GRCh38
rs969529995 Chr.8: 144392238-144392238 on Build GRCh38
rs368403183 Chr.8: 144392077-144392077 on Build GRCh38
rs782400904 Chr.8: 144392099-144392099 on Build GRCh38
rs782152916 Chr.8: 144392430-144392430 on Build GRCh38
rs782102304 Chr.8: 144392110-144392110 on Build GRCh38
rs1036720989 Chr.8: 144392206-144392206 on Build GRCh38
rs377651967 Chr.8: 144392358-144392358 on Build GRCh38
rs576962347 Chr.8: 144392389-144392389 on Build GRCh38
rs782590320 Chr.8: 144392178-144392178 on Build GRCh38
rs563415390 Chr.8: 144392338-144392352 on Build GRCh38
rs201295286 Chr.8: 144392307-144392307 on Build GRCh38
rs782066898 Chr.8: 144392212-144392212 on Build GRCh38
rs782550004 Chr.8: 144392062-144392062 on Build GRCh38
rs905655975 Chr.8: 144392396-144392396 on Build GRCh38
rs781966061 Chr.8: 144392068-144392068 on Build GRCh38
rs781852794 Chr.8: 144392414-144392415 on Build GRCh38
rs201745279 Chr.8: 144392123-144392123 on Build GRCh38
rs565794754 Chr.8: 144392165-144392165 on Build GRCh38
rs782357047 Chr.8: 144392192-144392192 on Build GRCh38
rs782031152 Chr.8: 144392217-144392217 on Build GRCh38
rs781916323 Chr.8: 144392078-144392078 on Build GRCh38
rs781787734 Chr.8: 144392446-144392446 on Build GRCh38
rs939971532 Chr.8: 144392357-144392357 on Build GRCh38
rs1044123730 Chr.8: 144392395-144392395 on Build GRCh38
rs932523960 Chr.8: 144392126-144392126 on Build GRCh38
rs782071356 Chr.8: 144392334-144392335 on Build GRCh38
rs782626192 Chr.8: 144392297-144392297 on Build GRCh38
rs190563627 Chr.8: 144392021-144392021 on Build GRCh38
rs782234244 Chr.8: 144392355-144392355 on Build GRCh38
rs559501595 Chr.8: 144392417-144392417 on Build GRCh38
rs561354112 Chr.8: 144392433-144392433 on Build GRCh38
rs370954240 Chr.8: 144392320-144392320 on Build GRCh38
rs7813146 Chr.8: 144392426-144392426 on Build GRCh38
rs895531783 Chr.8: 144392214-144392214 on Build GRCh38
rs782680934 Chr.8: 144392064-144392064 on Build GRCh38
rs781845522 Chr.8: 144392166-144392166 on Build GRCh38
rs781815159 Chr.8: 144392243-144392243 on Build GRCh38

View Full Product Details