Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144504976-144505481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPT
Entrez Gene ID
2875
Gene Name
glutamic--pyruvic transaminase
Gene Aliases
AAT1, ALT1, GPT1
UniGene
Hs.103502
Gene Chromosome Location
Chr.8: 144502217-144507174 on Build GRCh38
Transcripts
NM_005309.2 XM_011516993.2

Gene Symbol
LOC101928953
Entrez Gene ID
101928953
Gene Name
uncharacterized LOC101928953
Gene Aliases
-
UniGene
Hs.521937
Gene Chromosome Location
Chr.8: 144497357-144505474 on Build GRCh38
Transcripts
NM_005309.2 XM_011516993.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5345552 c.497C>T p.T166M Chr.8: 144505247-144505247 on Build GRCh38
6227482 c.496-6C>T p.? Chr.8: 144505240-144505240 on Build GRCh38
3374826 c.645G>A p.A215A Chr.8: 144505395-144505395 on Build GRCh38
4884576 c.406G>A p.V136M Chr.8: 144505042-144505042 on Build GRCh38
1097274 c.433G>A p.G145R Chr.8: 144505069-144505069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758875108 Chr.8: 144505225-144505225 on Build GRCh38
rs767936601 Chr.8: 144505335-144505335 on Build GRCh38
rs922141711 Chr.8: 144505005-144505005 on Build GRCh38
rs748547625 Chr.8: 144505063-144505063 on Build GRCh38
rs763375784 Chr.8: 144505172-144505172 on Build GRCh38
rs141571140 Chr.8: 144505061-144505061 on Build GRCh38
rs371565005 Chr.8: 144505383-144505383 on Build GRCh38
rs771282203 Chr.8: 144505434-144505434 on Build GRCh38
rs754527405 Chr.8: 144505360-144505360 on Build GRCh38
rs774645693 Chr.8: 144505329-144505329 on Build GRCh38
rs370098669 Chr.8: 144505179-144505179 on Build GRCh38
rs775213760 Chr.8: 144505073-144505073 on Build GRCh38
rs765923514 Chr.8: 144505276-144505276 on Build GRCh38
rs747843263 Chr.8: 144505287-144505287 on Build GRCh38
rs763130402 Chr.8: 144505240-144505240 on Build GRCh38
rs377045644 Chr.8: 144505152-144505152 on Build GRCh38
rs930407725 Chr.8: 144505104-144505104 on Build GRCh38
rs755633945 Chr.8: 144505195-144505195 on Build GRCh38
rs866709568 Chr.8: 144505268-144505268 on Build GRCh38
rs765501508 Chr.8: 144505164-144505164 on Build GRCh38
rs777764097 Chr.8: 144505380-144505380 on Build GRCh38
rs746987943 Chr.8: 144505310-144505310 on Build GRCh38
rs1020196606 Chr.8: 144505091-144505091 on Build GRCh38
rs576608308 Chr.8: 144505169-144505169 on Build GRCh38
rs769036858 Chr.8: 144505214-144505214 on Build GRCh38
rs748008493 Chr.8: 144505205-144505205 on Build GRCh38
rs764780268 Chr.8: 144505373-144505373 on Build GRCh38
rs530505425 Chr.8: 144505342-144505342 on Build GRCh38
rs765080403 Chr.8: 144505040-144505040 on Build GRCh38
rs752068526 Chr.8: 144505102-144505102 on Build GRCh38
rs879595120 Chr.8: 144505351-144505351 on Build GRCh38
rs758328894 Chr.8: 144505290-144505290 on Build GRCh38
rs752457447 Chr.8: 144505374-144505374 on Build GRCh38
rs146211713 Chr.8: 144505064-144505064 on Build GRCh38
rs149505691 Chr.8: 144505069-144505069 on Build GRCh38
rs777251291 Chr.8: 144505447-144505447 on Build GRCh38
rs751336078 Chr.8: 144505459-144505459 on Build GRCh38
rs756756813 Chr.8: 144505109-144505109 on Build GRCh38
rs758962078 Chr.8: 144505448-144505448 on Build GRCh38
rs765105086 Chr.8: 144504999-144504999 on Build GRCh38
rs762385509 Chr.8: 144505377-144505377 on Build GRCh38
rs138531202 Chr.8: 144505049-144505049 on Build GRCh38
rs371677109 Chr.8: 144505454-144505454 on Build GRCh38
rs754091629 Chr.8: 144505088-144505088 on Build GRCh38
rs764402392 Chr.8: 144505174-144505174 on Build GRCh38
rs374426053 Chr.8: 144505198-144505198 on Build GRCh38
rs373632268 Chr.8: 144505407-144505407 on Build GRCh38
rs768692147 Chr.8: 144505320-144505320 on Build GRCh38
rs375120964 Chr.8: 144505056-144505056 on Build GRCh38
rs748409747 Chr.8: 144505418-144505418 on Build GRCh38
rs917483341 Chr.8: 144505375-144505375 on Build GRCh38
rs973687000 Chr.8: 144505363-144505363 on Build GRCh38
rs778524553 Chr.8: 144505202-144505202 on Build GRCh38
rs951810643 Chr.8: 144505303-144505303 on Build GRCh38
rs745556641 Chr.8: 144505071-144505071 on Build GRCh38
rs780170301 Chr.8: 144505013-144505013 on Build GRCh38
rs781196445 Chr.8: 144505384-144505384 on Build GRCh38
rs753219034 Chr.8: 144505358-144505358 on Build GRCh38
rs148683067 Chr.8: 144505284-144505284 on Build GRCh38
rs755615955 Chr.8: 144505271-144505271 on Build GRCh38
rs752080977 Chr.8: 144505175-144505175 on Build GRCh38
rs777852066 Chr.8: 144505309-144505309 on Build GRCh38
rs144588980 Chr.8: 144505017-144505017 on Build GRCh38
rs759695712 Chr.8: 144505153-144505153 on Build GRCh38
rs1010077183 Chr.8: 144505254-144505254 on Build GRCh38
rs144520970 Chr.8: 144505317-144505317 on Build GRCh38
rs374035225 Chr.8: 144505144-144505144 on Build GRCh38
rs759784704 Chr.8: 144505095-144505095 on Build GRCh38
rs746826518 Chr.8: 144505382-144505382 on Build GRCh38
rs752686497 Chr.8: 144505004-144505004 on Build GRCh38
rs765542863 Chr.8: 144505025-144505026 on Build GRCh38
rs774169083 Chr.8: 144505080-144505080 on Build GRCh38
rs376363949 Chr.8: 144505334-144505334 on Build GRCh38
rs369853958 Chr.8: 144505139-144505139 on Build GRCh38
rs1018777294 Chr.8: 144505257-144505257 on Build GRCh38
rs759335306 Chr.8: 144504996-144504996 on Build GRCh38
rs559557046 Chr.8: 144505270-144505270 on Build GRCh38
rs367801266 Chr.8: 144505427-144505427 on Build GRCh38
rs138709427 Chr.8: 144505350-144505350 on Build GRCh38
rs776165677 Chr.8: 144505038-144505038 on Build GRCh38
rs770554695 Chr.8: 144505034-144505034 on Build GRCh38
rs754006020 Chr.8: 144504999-144505017 on Build GRCh38
rs376198243 Chr.8: 144505458-144505458 on Build GRCh38
rs745932486 Chr.8: 144505215-144505215 on Build GRCh38
rs779685262 Chr.8: 144505128-144505128 on Build GRCh38
rs372141763 Chr.8: 144505047-144505047 on Build GRCh38
rs148163358 Chr.8: 144505008-144505008 on Build GRCh38
rs143080454 Chr.8: 144505009-144505009 on Build GRCh38
rs1015116883 Chr.8: 144505336-144505336 on Build GRCh38
rs547778127 Chr.8: 144505285-144505285 on Build GRCh38
rs910044306 Chr.8: 144505328-144505328 on Build GRCh38
rs768393831 Chr.8: 144505132-144505132 on Build GRCh38
rs769967846 Chr.8: 144505311-144505311 on Build GRCh38
rs202140398 Chr.8: 144505286-144505286 on Build GRCh38
rs564918488 Chr.8: 144505184-144505184 on Build GRCh38
rs538093303 Chr.8: 144505046-144505046 on Build GRCh38
rs760804575 Chr.8: 144505082-144505082 on Build GRCh38
rs770070762 Chr.8: 144505221-144505221 on Build GRCh38
rs200088103 Chr.8: 144505033-144505033 on Build GRCh38
rs866078339 Chr.8: 144505224-144505224 on Build GRCh38
rs80198511 Chr.8: 144505119-144505119 on Build GRCh38
rs778310245 Chr.8: 144505290-144505295 on Build GRCh38
rs749597350 Chr.8: 144505016-144505016 on Build GRCh38
rs748706977 Chr.8: 144505020-144505020 on Build GRCh38
rs753222214 Chr.8: 144505174-144505174 on Build GRCh38
rs999848363 Chr.8: 144505101-144505101 on Build GRCh38
rs1000709364 Chr.8: 144505225-144505225 on Build GRCh38
rs762168599 Chr.8: 144505248-144505248 on Build GRCh38
rs756151404 Chr.8: 144505048-144505048 on Build GRCh38
rs748933417 Chr.8: 144505201-144505201 on Build GRCh38
rs144054398 Chr.8: 144505078-144505078 on Build GRCh38
rs1032661716 Chr.8: 144505120-144505120 on Build GRCh38
rs773478792 Chr.8: 144505451-144505451 on Build GRCh38
rs769394527 Chr.8: 144505125-144505125 on Build GRCh38
rs764395897 Chr.8: 144505241-144505241 on Build GRCh38
rs759156616 Chr.8: 144505453-144505453 on Build GRCh38
rs569438469 Chr.8: 144505312-144505312 on Build GRCh38
rs866440873 Chr.8: 144505087-144505087 on Build GRCh38
rs144666868 Chr.8: 144505398-144505398 on Build GRCh38
rs775371734 Chr.8: 144505041-144505041 on Build GRCh38
rs762766505 Chr.8: 144505074-144505074 on Build GRCh38
rs374307577 Chr.8: 144505247-144505247 on Build GRCh38
rs769552512 Chr.8: 144505072-144505072 on Build GRCh38
rs1010460386 Chr.8: 144505431-144505431 on Build GRCh38
rs762649725 Chr.8: 144505457-144505457 on Build GRCh38
rs772159817 Chr.8: 144505150-144505150 on Build GRCh38
rs139107415 Chr.8: 144505068-144505068 on Build GRCh38
rs753411777 Chr.8: 144505279-144505279 on Build GRCh38
rs761121667 Chr.8: 144505347-144505347 on Build GRCh38
rs561409231 Chr.8: 144505154-144505154 on Build GRCh38
rs762924568 Chr.8: 144505042-144505042 on Build GRCh38
rs759308893 Chr.8: 144505039-144505039 on Build GRCh38
rs940284019 Chr.8: 144505157-144505157 on Build GRCh38
rs529679428 Chr.8: 144505273-144505273 on Build GRCh38
rs140704858 Chr.8: 144505392-144505392 on Build GRCh38
rs747452460 Chr.8: 144505186-144505186 on Build GRCh38
rs772070124 Chr.8: 144505211-144505211 on Build GRCh38
rs755336318 Chr.8: 144505018-144505018 on Build GRCh38
rs200128241 Chr.8: 144505180-144505180 on Build GRCh38
rs757320127 Chr.8: 144505007-144505007 on Build GRCh38
rs200479839 Chr.8: 144505018-144505018 on Build GRCh38
rs927315588 Chr.8: 144505410-144505410 on Build GRCh38
rs936090537 Chr.8: 144505411-144505411 on Build GRCh38
rs769944889 Chr.8: 144505429-144505429 on Build GRCh38
rs753047103 Chr.8: 144505098-144505098 on Build GRCh38
rs141187317 Chr.8: 144505426-144505426 on Build GRCh38
rs572474138 Chr.8: 144505099-144505099 on Build GRCh38
rs780726435 Chr.8: 144505111-144505111 on Build GRCh38
rs200304696 Chr.8: 144505222-144505222 on Build GRCh38
rs541927226 Chr.8: 144505401-144505401 on Build GRCh38
rs201065145 Chr.8: 144505096-144505096 on Build GRCh38

View Full Product Details