Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144532198-144532698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP39
Entrez Gene ID
80728
Gene Name
Rho GTPase activating protein 39
Gene Aliases
CrGAP, Vilse
UniGene
Hs.148988
Gene Chromosome Location
Chr.8: 144529179-144700520 on Build GRCh38
Transcripts
NM_001308207.1 NM_001308208.1 NM_025251.2 XM_011517308.1 XM_011517309.1 XM_011517312.2 XM_017013870.1 XM_017013871.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3834304 c.2973C>T p.H991H Chr.8: 144532312-144532312 on Build GRCh38
7235823 c.2980+9C>G p.? Chr.8: 144532296-144532296 on Build GRCh38
7259309 c.2938C>A p.Q980K Chr.8: 144532347-144532347 on Build GRCh38
2872019 c.2908G>A p.E970K Chr.8: 144532377-144532377 on Build GRCh38
389688 c.2949G>T p.V983V Chr.8: 144532336-144532336 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767320961 Chr.8: 144532299-144532299 on Build GRCh38
rs1033403544 Chr.8: 144532526-144532526 on Build GRCh38
rs977858557 Chr.8: 144532547-144532547 on Build GRCh38
rs749635962 Chr.8: 144532274-144532274 on Build GRCh38
rs777679326 Chr.8: 144532348-144532348 on Build GRCh38
rs760520687 Chr.8: 144532441-144532441 on Build GRCh38
rs774841380 Chr.8: 144532444-144532444 on Build GRCh38
rs768030114 Chr.8: 144532288-144532288 on Build GRCh38
rs750159928 Chr.8: 144532302-144532302 on Build GRCh38
rs780353737 Chr.8: 144532288-144532289 on Build GRCh38
rs35236830 Chr.8: 144532563-144532563 on Build GRCh38
rs766369631 Chr.8: 144532311-144532311 on Build GRCh38
rs549587390 Chr.8: 144532478-144532478 on Build GRCh38
rs952752701 Chr.8: 144532234-144532234 on Build GRCh38
rs769490353 Chr.8: 144532564-144532564 on Build GRCh38
rs140238043 Chr.8: 144532392-144532392 on Build GRCh38
rs775556558 Chr.8: 144532491-144532491 on Build GRCh38
rs1013076169 Chr.8: 144532530-144532530 on Build GRCh38
rs111775706 Chr.8: 144532242-144532242 on Build GRCh38
rs571074241 Chr.8: 144532492-144532492 on Build GRCh38
rs775859080 Chr.8: 144532275-144532275 on Build GRCh38
rs200403169 Chr.8: 144532287-144532287 on Build GRCh38
rs760608723 Chr.8: 144532309-144532309 on Build GRCh38
rs921693226 Chr.8: 144532537-144532537 on Build GRCh38
rs895766587 Chr.8: 144532589-144532589 on Build GRCh38
rs111825321 Chr.8: 144532639-144532639 on Build GRCh38
rs369438966 Chr.8: 144532272-144532272 on Build GRCh38
rs771645411 Chr.8: 144532426-144532426 on Build GRCh38
rs944473425 Chr.8: 144532532-144532532 on Build GRCh38
rs758403004 Chr.8: 144532259-144532259 on Build GRCh38
rs932510832 Chr.8: 144532595-144532595 on Build GRCh38
rs898243856 Chr.8: 144532306-144532306 on Build GRCh38
rs370900954 Chr.8: 144532440-144532440 on Build GRCh38
rs888683034 Chr.8: 144532466-144532466 on Build GRCh38
rs749729532 Chr.8: 144532402-144532402 on Build GRCh38
rs774095048 Chr.8: 144532292-144532292 on Build GRCh38
rs747929755 Chr.8: 144532524-144532524 on Build GRCh38
rs888715543 Chr.8: 144532552-144532552 on Build GRCh38
rs757500213 Chr.8: 144532357-144532357 on Build GRCh38
rs751834444 Chr.8: 144532353-144532353 on Build GRCh38
rs200665709 Chr.8: 144532408-144532408 on Build GRCh38
rs995039915 Chr.8: 144532347-144532347 on Build GRCh38
rs781002265 Chr.8: 144532474-144532474 on Build GRCh38
rs768244228 Chr.8: 144532407-144532407 on Build GRCh38
rs955922712 Chr.8: 144532521-144532521 on Build GRCh38
rs1000054779 Chr.8: 144532662-144532662 on Build GRCh38
rs770273101 Chr.8: 144532387-144532387 on Build GRCh38
rs1025000306 Chr.8: 144532543-144532543 on Build GRCh38
rs986649708 Chr.8: 144532599-144532599 on Build GRCh38
rs550596902 Chr.8: 144532600-144532600 on Build GRCh38
rs1048658640 Chr.8: 144532461-144532461 on Build GRCh38
rs565585303 Chr.8: 144532545-144532545 on Build GRCh38
rs982737388 Chr.8: 144532241-144532241 on Build GRCh38
rs958820315 Chr.8: 144532391-144532391 on Build GRCh38
rs915703847 Chr.8: 144532250-144532250 on Build GRCh38
rs773585138 Chr.8: 144532240-144532240 on Build GRCh38
rs758551180 Chr.8: 144532319-144532319 on Build GRCh38
rs578206787 Chr.8: 144532676-144532676 on Build GRCh38
rs992695154 Chr.8: 144532247-144532247 on Build GRCh38
rs1048523860 Chr.8: 144532548-144532548 on Build GRCh38
rs752753658 Chr.8: 144532256-144532256 on Build GRCh38
rs781696294 Chr.8: 144532376-144532376 on Build GRCh38
rs201288884 Chr.8: 144532410-144532410 on Build GRCh38
rs971831147 Chr.8: 144532414-144532414 on Build GRCh38
rs781653120 Chr.8: 144532273-144532273 on Build GRCh38
rs531660285 Chr.8: 144532473-144532473 on Build GRCh38
rs778111969 Chr.8: 144532266-144532266 on Build GRCh38
rs890801024 Chr.8: 144532267-144532267 on Build GRCh38
rs1039233896 Chr.8: 144532535-144532535 on Build GRCh38
rs536232700 Chr.8: 144532551-144532551 on Build GRCh38
rs934350002 Chr.8: 144532384-144532384 on Build GRCh38
rs201397901 Chr.8: 144532286-144532286 on Build GRCh38
rs761333246 Chr.8: 144532296-144532296 on Build GRCh38
rs899794362 Chr.8: 144532248-144532248 on Build GRCh38
rs898131784 Chr.8: 144532508-144532508 on Build GRCh38
rs569611920 Chr.8: 144532456-144532456 on Build GRCh38
rs117567558 Chr.8: 144532459-144532459 on Build GRCh38
rs1004395488 Chr.8: 144532560-144532560 on Build GRCh38
rs919608230 Chr.8: 144532648-144532648 on Build GRCh38
rs921335659 Chr.8: 144532578-144532578 on Build GRCh38
rs116655978 Chr.8: 144532529-144532529 on Build GRCh38
rs753702967 Chr.8: 144532312-144532312 on Build GRCh38
rs556750937 Chr.8: 144532246-144532246 on Build GRCh38
rs554228688 Chr.8: 144532596-144532596 on Build GRCh38
rs939963381 Chr.8: 144532626-144532626 on Build GRCh38
rs891028614 Chr.8: 144532480-144532480 on Build GRCh38
rs978584875 Chr.8: 144532514-144532514 on Build GRCh38
rs192971226 Chr.8: 144532378-144532378 on Build GRCh38
rs1004190831 Chr.8: 144532468-144532468 on Build GRCh38
rs532305355 Chr.8: 144532516-144532516 on Build GRCh38
rs925888681 Chr.8: 144532515-144532515 on Build GRCh38
rs1026944420 Chr.8: 144532233-144532233 on Build GRCh38
rs1041225505 Chr.8: 144532455-144532455 on Build GRCh38
rs574195740 Chr.8: 144532237-144532237 on Build GRCh38
rs1014722425 Chr.8: 144532475-144532475 on Build GRCh38
rs537118152 Chr.8: 144532625-144532625 on Build GRCh38
rs924728337 Chr.8: 144532431-144532431 on Build GRCh38

View Full Product Details