Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 3898560-3899061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLIS3
Entrez Gene ID
169792
Gene Name
GLIS family zinc finger 3
Gene Aliases
NDH, ZNF515
UniGene
Hs.162125
Gene Chromosome Location
Chr.9: 3824127-4310694 on Build GRCh38
Transcripts
NM_001042413.1 NM_152629.3 XM_005251386.4 XM_005251387.4 XM_005251388.4 XM_011517763.2 XM_011517764.2 XM_011517765.2 XM_011517766.2 XM_011517767.2 XM_017014361.1 NR_026663.1

Gene Symbol
GLIS3-AS1
Entrez Gene ID
84850
Gene Name
GLIS3 antisense RNA 1
Gene Aliases
C9orf70
UniGene
Hs.676113
Gene Chromosome Location
Chr.9: 3898646-3901248 on Build GRCh38
Transcripts
NM_001042413.1 NM_152629.3 XM_005251386.4 XM_005251387.4 XM_005251388.4 XM_011517763.2 XM_011517764.2 XM_011517765.2 XM_011517766.2 XM_011517767.2 XM_017014361.1 NR_026663.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3930071 c.2035G>A p.V679M Chr.9: 3898784-3898784 on Build GRCh38
753409 c.2021C>A p.T674K Chr.9: 3898798-3898798 on Build GRCh38
7332405 c.1593T>A p.T531T Chr.9: 3898761-3898761 on Build GRCh38
1109153 c.2034C>T p.T678T Chr.9: 3898785-3898785 on Build GRCh38
4847993 c.1661C>T p.S554L Chr.9: 3898693-3898693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778931847 Chr.9: 3898807-3898807 on Build GRCh38
rs149840771 Chr.9: 3898723-3898723 on Build GRCh38
rs767982176 Chr.9: 3898859-3898859 on Build GRCh38
rs1043024204 Chr.9: 3899009-3899009 on Build GRCh38
rs983698069 Chr.9: 3898726-3898726 on Build GRCh38
rs766143053 Chr.9: 3898863-3898863 on Build GRCh38
rs752655699 Chr.9: 3898875-3898875 on Build GRCh38
rs775354523 Chr.9: 3898591-3898591 on Build GRCh38
rs768709952 Chr.9: 3898739-3898739 on Build GRCh38
rs149983395 Chr.9: 3898792-3898792 on Build GRCh38
rs761011670 Chr.9: 3898614-3898614 on Build GRCh38
rs965961382 Chr.9: 3898914-3898914 on Build GRCh38
rs750887891 Chr.9: 3898860-3898860 on Build GRCh38
rs757250015 Chr.9: 3898946-3898946 on Build GRCh38
rs762263594 Chr.9: 3898857-3898857 on Build GRCh38
rs900219690 Chr.9: 3898740-3898740 on Build GRCh38
rs746811228 Chr.9: 3898883-3898883 on Build GRCh38
rs143998067 Chr.9: 3898731-3898731 on Build GRCh38
rs751808508 Chr.9: 3898776-3898776 on Build GRCh38
rs777760361 Chr.9: 3898917-3898917 on Build GRCh38
rs886063953 Chr.9: 3898709-3898709 on Build GRCh38
rs770069148 Chr.9: 3898830-3898830 on Build GRCh38
rs766359946 Chr.9: 3898615-3898615 on Build GRCh38
rs758719598 Chr.9: 3898637-3898637 on Build GRCh38
rs1046037850 Chr.9: 3898603-3898603 on Build GRCh38
rs749355262 Chr.9: 3898918-3898918 on Build GRCh38
rs370792971 Chr.9: 3898710-3898710 on Build GRCh38
rs753170924 Chr.9: 3898706-3898706 on Build GRCh38
rs568262538 Chr.9: 3898742-3898742 on Build GRCh38
rs1009840380 Chr.9: 3898597-3898597 on Build GRCh38
rs892881743 Chr.9: 3898635-3898635 on Build GRCh38
rs542599450 Chr.9: 3898768-3898768 on Build GRCh38
rs765113162 Chr.9: 3898610-3898610 on Build GRCh38
rs1032521326 Chr.9: 3898593-3898593 on Build GRCh38
rs963267736 Chr.9: 3898953-3898953 on Build GRCh38
rs753397371 Chr.9: 3898798-3898798 on Build GRCh38
rs995973547 Chr.9: 3898958-3898958 on Build GRCh38
rs950606396 Chr.9: 3898897-3898897 on Build GRCh38
rs756790400 Chr.9: 3898713-3898713 on Build GRCh38
rs374704177 Chr.9: 3898645-3898645 on Build GRCh38
rs945603237 Chr.9: 3899008-3899008 on Build GRCh38
rs1033764529 Chr.9: 3898647-3898648 on Build GRCh38
rs997367444 Chr.9: 3898719-3898719 on Build GRCh38
rs765694369 Chr.9: 3898700-3898700 on Build GRCh38
rs761918869 Chr.9: 3898844-3898844 on Build GRCh38
rs374929970 Chr.9: 3898759-3898759 on Build GRCh38
rs765749277 Chr.9: 3898632-3898632 on Build GRCh38
rs935738652 Chr.9: 3898745-3898745 on Build GRCh38
rs746619651 Chr.9: 3898643-3898643 on Build GRCh38
rs776871579 Chr.9: 3898767-3898767 on Build GRCh38
rs767726512 Chr.9: 3898596-3898596 on Build GRCh38
rs1004839994 Chr.9: 3898938-3898938 on Build GRCh38
rs747988294 Chr.9: 3898747-3898747 on Build GRCh38
rs750080742 Chr.9: 3898785-3898785 on Build GRCh38
rs779710501 Chr.9: 3898795-3898795 on Build GRCh38
rs61201518 Chr.9: 3898896-3898896 on Build GRCh38
rs140882668 Chr.9: 3898819-3898819 on Build GRCh38
rs780841378 Chr.9: 3898654-3898654 on Build GRCh38
rs762852840 Chr.9: 3898592-3898592 on Build GRCh38
rs368500557 Chr.9: 3898655-3898655 on Build GRCh38
rs928661053 Chr.9: 3899035-3899035 on Build GRCh38
rs144720690 Chr.9: 3898831-3898831 on Build GRCh38
rs753011920 Chr.9: 3898633-3898633 on Build GRCh38
rs934831344 Chr.9: 3898628-3898628 on Build GRCh38
rs10122183 Chr.9: 3898987-3898987 on Build GRCh38
rs762258189 Chr.9: 3898783-3898783 on Build GRCh38
rs145670205 Chr.9: 3898851-3898851 on Build GRCh38
rs753239534 Chr.9: 3898866-3898866 on Build GRCh38
rs983771962 Chr.9: 3898902-3898902 on Build GRCh38
rs145867412 Chr.9: 3898724-3898724 on Build GRCh38
rs775604544 Chr.9: 3898773-3898773 on Build GRCh38
rs201556131 Chr.9: 3898784-3898784 on Build GRCh38
rs911849390 Chr.9: 3898907-3898907 on Build GRCh38
rs764353179 Chr.9: 3898711-3898711 on Build GRCh38
rs200705602 Chr.9: 3898712-3898712 on Build GRCh38
rs1023069615 Chr.9: 3898973-3898973 on Build GRCh38
rs754814654 Chr.9: 3898802-3898802 on Build GRCh38
rs934809482 Chr.9: 3898832-3898832 on Build GRCh38
rs760341925 Chr.9: 3898862-3898862 on Build GRCh38
rs547125358 Chr.9: 3898959-3898959 on Build GRCh38
rs759637587 Chr.9: 3898686-3898686 on Build GRCh38
rs367590913 Chr.9: 3898809-3898809 on Build GRCh38
rs763102598 Chr.9: 3898842-3898842 on Build GRCh38
rs377343842 Chr.9: 3898660-3898660 on Build GRCh38
rs774264365 Chr.9: 3898668-3898668 on Build GRCh38
rs773277946 Chr.9: 3898604-3898604 on Build GRCh38
rs139466536 Chr.9: 3898627-3898627 on Build GRCh38
rs201490424 Chr.9: 3898876-3898876 on Build GRCh38
rs778328383 Chr.9: 3898874-3898874 on Build GRCh38
rs754549738 Chr.9: 3898871-3898871 on Build GRCh38
rs751672476 Chr.9: 3898747-3898748 on Build GRCh38
rs764663528 Chr.9: 3898775-3898775 on Build GRCh38
rs749281737 Chr.9: 3898664-3898664 on Build GRCh38
rs749172684 Chr.9: 3898734-3898734 on Build GRCh38
rs974215468 Chr.9: 3898761-3898761 on Build GRCh38
rs777640894 Chr.9: 3898890-3898890 on Build GRCh38
rs766629040 Chr.9: 3898684-3898684 on Build GRCh38
rs148816140 Chr.9: 3898730-3898730 on Build GRCh38
rs578155771 Chr.9: 3898671-3898671 on Build GRCh38
rs558256107 Chr.9: 3898666-3898666 on Build GRCh38
rs529580636 Chr.9: 3899000-3899000 on Build GRCh38
rs758874232 Chr.9: 3898729-3898729 on Build GRCh38
rs375970704 Chr.9: 3898850-3898850 on Build GRCh38
rs755350906 Chr.9: 3898824-3898824 on Build GRCh38
rs199817194 Chr.9: 3898825-3898825 on Build GRCh38
rs959189410 Chr.9: 3898676-3898676 on Build GRCh38

View Full Product Details