Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 4706290-4706790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC37L1
Entrez Gene ID
55664
Gene Name
cell division cycle 37 like 1
Gene Aliases
CDC37B, HARC
UniGene
Hs.666439
Gene Chromosome Location
Chr.9: 4679553-4708399 on Build GRCh38
Transcripts
NM_017913.3

SNP Detail

SNP ID Chromosome Location
rs1004642883 Chr.9: 4706695-4706695 on Build GRCh38
rs3174298 Chr.9: 4706366-4706366 on Build GRCh38
rs934792127 Chr.9: 4706717-4706717 on Build GRCh38
rs549477192 Chr.9: 4706590-4706590 on Build GRCh38
rs565074108 Chr.9: 4706452-4706452 on Build GRCh38
rs1040587579 Chr.9: 4706318-4706318 on Build GRCh38
rs976319737 Chr.9: 4706669-4706669 on Build GRCh38
rs189256250 Chr.9: 4706722-4706722 on Build GRCh38
rs545358882 Chr.9: 4706384-4706384 on Build GRCh38
rs970656375 Chr.9: 4706531-4706531 on Build GRCh38
rs893494467 Chr.9: 4706408-4706408 on Build GRCh38
rs753578502 Chr.9: 4706538-4706538 on Build GRCh38
rs910542348 Chr.9: 4706572-4706572 on Build GRCh38
rs947384710 Chr.9: 4706633-4706633 on Build GRCh38
rs765975391 Chr.9: 4706439-4706439 on Build GRCh38
rs900403377 Chr.9: 4706679-4706679 on Build GRCh38
rs997384590 Chr.9: 4706680-4706680 on Build GRCh38
rs886234974 Chr.9: 4706688-4706688 on Build GRCh38
rs1044774924 Chr.9: 4706748-4706748 on Build GRCh38
rs986111213 Chr.9: 4706567-4706567 on Build GRCh38
rs1051595376 Chr.9: 4706684-4706684 on Build GRCh38
rs185756287 Chr.9: 4706708-4706708 on Build GRCh38
rs755740888 Chr.9: 4706422-4706422 on Build GRCh38
rs924797207 Chr.9: 4706698-4706698 on Build GRCh38
rs372254206 Chr.9: 4706549-4706549 on Build GRCh38
rs1004590339 Chr.9: 4706377-4706377 on Build GRCh38
rs74660776 Chr.9: 4706562-4706562 on Build GRCh38
rs1052165866 Chr.9: 4706407-4706407 on Build GRCh38
rs763517846 Chr.9: 4706336-4706336 on Build GRCh38
rs946376423 Chr.9: 4706745-4706745 on Build GRCh38
rs386731955 Chr.9: 4706484-4706485 on Build GRCh38
rs188682303 Chr.9: 4706484-4706484 on Build GRCh38
rs914568731 Chr.9: 4706612-4706612 on Build GRCh38
rs1053097384 Chr.9: 4706582-4706582 on Build GRCh38
rs1034847919 Chr.9: 4706498-4706498 on Build GRCh38
rs1052195596 Chr.9: 4706721-4706721 on Build GRCh38
rs376830552 Chr.9: 4706592-4706592 on Build GRCh38
rs1003488922 Chr.9: 4706755-4706755 on Build GRCh38
rs531622067 Chr.9: 4706376-4706376 on Build GRCh38
rs945011765 Chr.9: 4706639-4706640 on Build GRCh38
rs961836517 Chr.9: 4706524-4706524 on Build GRCh38
rs893521831 Chr.9: 4706729-4706729 on Build GRCh38
rs775201034 Chr.9: 4706546-4706546 on Build GRCh38
rs904846960 Chr.9: 4706754-4706754 on Build GRCh38
rs565112709 Chr.9: 4706481-4706481 on Build GRCh38
rs754718438 Chr.9: 4706557-4706557 on Build GRCh38
rs192852130 Chr.9: 4706485-4706485 on Build GRCh38
rs532213892 Chr.9: 4706568-4706568 on Build GRCh38
rs1023981270 Chr.9: 4706464-4706464 on Build GRCh38
rs1039386352 Chr.9: 4706670-4706670 on Build GRCh38

View Full Product Details