Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 4709465-4709971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK3
Entrez Gene ID
50808
Gene Name
adenylate kinase 3
Gene Aliases
AK3L1, AK6, AKL3L, AKL3L1, FIX
UniGene
Hs.732022
Gene Chromosome Location
Chr.9: 4709556-4742043 on Build GRCh38
Transcripts
NM_001199852.1 NM_001199853.1 NM_001199855.1 NM_001199856.1 NM_016282.3

SNP Detail

SNP ID Chromosome Location
rs1006096730 Chr.9: 4709916-4709916 on Build GRCh38
rs146817880 Chr.9: 4709774-4709774 on Build GRCh38
rs757382044 Chr.9: 4709612-4709614 on Build GRCh38
rs892809670 Chr.9: 4709693-4709693 on Build GRCh38
rs895442052 Chr.9: 4709721-4709731 on Build GRCh38
rs1034154300 Chr.9: 4709897-4709897 on Build GRCh38
rs971156394 Chr.9: 4709567-4709567 on Build GRCh38
rs948302349 Chr.9: 4709721-4709721 on Build GRCh38
rs551050409 Chr.9: 4709769-4709769 on Build GRCh38
rs301492 Chr.9: 4709861-4709861 on Build GRCh38
rs774209284 Chr.9: 4709508-4709508 on Build GRCh38
rs933495501 Chr.9: 4709651-4709651 on Build GRCh38
rs748578576 Chr.9: 4709792-4709792 on Build GRCh38
rs902498526 Chr.9: 4709521-4709521 on Build GRCh38
rs201527873 Chr.9: 4709833-4709834 on Build GRCh38
rs953977693 Chr.9: 4709810-4709810 on Build GRCh38
rs1032609000 Chr.9: 4709677-4709677 on Build GRCh38
rs1050591464 Chr.9: 4709797-4709797 on Build GRCh38
rs1015700429 Chr.9: 4709571-4709571 on Build GRCh38
rs1048020 Chr.9: 4709784-4709784 on Build GRCh38
rs570710592 Chr.9: 4709642-4709644 on Build GRCh38
rs761603084 Chr.9: 4709586-4709586 on Build GRCh38
rs147921235 Chr.9: 4709944-4709944 on Build GRCh38
rs1002692640 Chr.9: 4709725-4709725 on Build GRCh38
rs159878 Chr.9: 4709615-4709615 on Build GRCh38
rs936652613 Chr.9: 4709698-4709698 on Build GRCh38
rs1041800265 Chr.9: 4709875-4709875 on Build GRCh38
rs892757283 Chr.9: 4709858-4709858 on Build GRCh38
rs971585595 Chr.9: 4709724-4709724 on Build GRCh38
rs984766525 Chr.9: 4709542-4709542 on Build GRCh38
rs767517791 Chr.9: 4709614-4709614 on Build GRCh38
rs1046609552 Chr.9: 4709911-4709912 on Build GRCh38
rs548347895 Chr.9: 4709729-4709729 on Build GRCh38
rs768219334 Chr.9: 4709503-4709503 on Build GRCh38
rs1054449166 Chr.9: 4709701-4709701 on Build GRCh38
rs1025099080 Chr.9: 4709563-4709563 on Build GRCh38
rs570760410 Chr.9: 4709770-4709770 on Build GRCh38
rs149199157 Chr.9: 4709536-4709536 on Build GRCh38
rs575389380 Chr.9: 4709880-4709880 on Build GRCh38
rs183331999 Chr.9: 4709804-4709804 on Build GRCh38
rs142439656 Chr.9: 4709658-4709658 on Build GRCh38
rs924010047 Chr.9: 4709854-4709854 on Build GRCh38
rs747578811 Chr.9: 4709892-4709892 on Build GRCh38
rs779102648 Chr.9: 4709788-4709788 on Build GRCh38
rs909225919 Chr.9: 4709547-4709547 on Build GRCh38
rs919818125 Chr.9: 4709612-4709612 on Build GRCh38
rs377008347 Chr.9: 4709920-4709920 on Build GRCh38
rs936887475 Chr.9: 4709825-4709825 on Build GRCh38
rs528678312 Chr.9: 4709728-4709728 on Build GRCh38
rs909173321 Chr.9: 4709749-4709749 on Build GRCh38
rs1002566948 Chr.9: 4709568-4709568 on Build GRCh38
rs1019481161 Chr.9: 4709826-4709826 on Build GRCh38
rs974407574 Chr.9: 4709605-4709605 on Build GRCh38
rs972507625 Chr.9: 4709777-4709777 on Build GRCh38
rs1001088521 Chr.9: 4709673-4709673 on Build GRCh38
rs530946941 Chr.9: 4709762-4709762 on Build GRCh38
rs985360824 Chr.9: 4709812-4709817 on Build GRCh38
rs937280727 Chr.9: 4709537-4709537 on Build GRCh38
rs948331994 Chr.9: 4709901-4709901 on Build GRCh38
rs1053962525 Chr.9: 4709827-4709827 on Build GRCh38
rs560066613 Chr.9: 4709911-4709911 on Build GRCh38
rs193190747 Chr.9: 4709708-4709708 on Build GRCh38
rs961046262 Chr.9: 4709577-4709577 on Build GRCh38
rs1012330951 Chr.9: 4709561-4709561 on Build GRCh38
rs1009900289 Chr.9: 4709704-4709704 on Build GRCh38
rs776550637 Chr.9: 4709863-4709863 on Build GRCh38
rs984714003 Chr.9: 4709740-4709740 on Build GRCh38
rs916569964 Chr.9: 4709888-4709888 on Build GRCh38
rs962036606 Chr.9: 4709751-4709751 on Build GRCh38
rs187933208 Chr.9: 4709807-4709807 on Build GRCh38
rs1051060087 Chr.9: 4709653-4709653 on Build GRCh38
rs561929578 Chr.9: 4709739-4709739 on Build GRCh38
rs563968751 Chr.9: 4709581-4709581 on Build GRCh38
rs763729018 Chr.9: 4709507-4709507 on Build GRCh38
rs766515435 Chr.9: 4709710-4709710 on Build GRCh38

View Full Product Details