Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 4712790-4713295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK3
Entrez Gene ID
50808
Gene Name
adenylate kinase 3
Gene Aliases
AK3L1, AK6, AKL3L, AKL3L1, FIX
UniGene
Hs.732022
Gene Chromosome Location
Chr.9: 4709556-4742043 on Build GRCh38
Transcripts
NM_001199852.1 NM_001199853.1 NM_001199855.1 NM_001199856.1 NM_016282.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
324684 c.633C>A p.F211L Chr.9: 4713027-4713027 on Build GRCh38
6039198 c.629C>T p.A210V Chr.9: 4713031-4713031 on Build GRCh38
3907503 c.683G>C p.*228S Chr.9: 4712977-4712977 on Build GRCh38
3907504 c.612T>G p.I204M Chr.9: 4713048-4713048 on Build GRCh38
3848608 c.593G>A p.G198E Chr.9: 4713067-4713067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891352754 Chr.9: 4713109-4713109 on Build GRCh38
rs374202443 Chr.9: 4713096-4713096 on Build GRCh38
rs1023039243 Chr.9: 4713252-4713252 on Build GRCh38
rs201868095 Chr.9: 4713033-4713033 on Build GRCh38
rs779235021 Chr.9: 4713117-4713120 on Build GRCh38
rs142894703 Chr.9: 4713087-4713087 on Build GRCh38
rs1012618485 Chr.9: 4713251-4713251 on Build GRCh38
rs375251919 Chr.9: 4712977-4712977 on Build GRCh38
rs750272249 Chr.9: 4712946-4712946 on Build GRCh38
rs767490182 Chr.9: 4713019-4713019 on Build GRCh38
rs769541213 Chr.9: 4713009-4713009 on Build GRCh38
rs374481496 Chr.9: 4713159-4713159 on Build GRCh38
rs780281634 Chr.9: 4713133-4713133 on Build GRCh38
rs371329382 Chr.9: 4713180-4713180 on Build GRCh38
rs372383858 Chr.9: 4713068-4713068 on Build GRCh38
rs1045782637 Chr.9: 4712847-4712847 on Build GRCh38
rs755185342 Chr.9: 4713109-4713110 on Build GRCh38
rs376111896 Chr.9: 4713160-4713160 on Build GRCh38
rs762691609 Chr.9: 4713011-4713011 on Build GRCh38
rs76198439 Chr.9: 4712993-4712993 on Build GRCh38
rs763692807 Chr.9: 4712932-4712932 on Build GRCh38
rs570640083 Chr.9: 4712948-4712949 on Build GRCh38
rs1004256840 Chr.9: 4712860-4712862 on Build GRCh38
rs770238321 Chr.9: 4713050-4713050 on Build GRCh38
rs773920260 Chr.9: 4713017-4713017 on Build GRCh38
rs372184729 Chr.9: 4713145-4713145 on Build GRCh38
rs296827 Chr.9: 4713213-4713213 on Build GRCh38
rs755861699 Chr.9: 4713027-4713027 on Build GRCh38
rs530394621 Chr.9: 4713167-4713167 on Build GRCh38
rs916541050 Chr.9: 4713249-4713249 on Build GRCh38
rs374392990 Chr.9: 4712950-4712950 on Build GRCh38
rs974127508 Chr.9: 4713157-4713157 on Build GRCh38
rs759459731 Chr.9: 4712856-4712856 on Build GRCh38
rs368373795 Chr.9: 4712999-4712999 on Build GRCh38
rs902667437 Chr.9: 4713220-4713220 on Build GRCh38
rs749707630 Chr.9: 4713138-4713138 on Build GRCh38
rs768409645 Chr.9: 4713069-4713069 on Build GRCh38
rs777921982 Chr.9: 4712963-4712963 on Build GRCh38
rs150502156 Chr.9: 4712904-4712904 on Build GRCh38
rs761350717 Chr.9: 4713084-4713084 on Build GRCh38
rs746411663 Chr.9: 4713048-4713048 on Build GRCh38
rs142499153 Chr.9: 4712881-4712881 on Build GRCh38
rs747143567 Chr.9: 4713040-4713040 on Build GRCh38
rs768160584 Chr.9: 4713140-4713140 on Build GRCh38
rs915726706 Chr.9: 4712817-4712817 on Build GRCh38
rs773084111 Chr.9: 4713088-4713088 on Build GRCh38
rs977089245 Chr.9: 4712932-4712935 on Build GRCh38
rs62542955 Chr.9: 4712865-4712865 on Build GRCh38
rs377461213 Chr.9: 4712933-4712933 on Build GRCh38
rs998575279 Chr.9: 4713229-4713229 on Build GRCh38
rs999344155 Chr.9: 4713189-4713189 on Build GRCh38
rs779041330 Chr.9: 4712955-4712955 on Build GRCh38
rs200235726 Chr.9: 4712957-4712957 on Build GRCh38
rs758359605 Chr.9: 4712958-4712958 on Build GRCh38
rs940107693 Chr.9: 4713151-4713151 on Build GRCh38
rs559439993 Chr.9: 4713091-4713091 on Build GRCh38
rs747820244 Chr.9: 4713144-4713144 on Build GRCh38
rs747406734 Chr.9: 4712967-4712967 on Build GRCh38
rs781448075 Chr.9: 4713126-4713126 on Build GRCh38
rs775444155 Chr.9: 4712867-4712867 on Build GRCh38
rs1038405175 Chr.9: 4712992-4712992 on Build GRCh38
rs751865761 Chr.9: 4713020-4713020 on Build GRCh38
rs1031266685 Chr.9: 4713193-4713193 on Build GRCh38
rs368886471 Chr.9: 4713132-4713132 on Build GRCh38
rs184342788 Chr.9: 4713240-4713240 on Build GRCh38
rs372240880 Chr.9: 4713131-4713131 on Build GRCh38
rs757373618 Chr.9: 4713125-4713125 on Build GRCh38
rs768049334 Chr.9: 4713179-4713179 on Build GRCh38
rs777711779 Chr.9: 4713036-4713036 on Build GRCh38
rs878974781 Chr.9: 4713209-4713209 on Build GRCh38
rs373524316 Chr.9: 4712927-4712927 on Build GRCh38
rs370649949 Chr.9: 4712974-4712974 on Build GRCh38
rs148411833 Chr.9: 4712869-4712869 on Build GRCh38
rs760617107 Chr.9: 4713090-4713090 on Build GRCh38
rs757819525 Chr.9: 4713043-4713043 on Build GRCh38
rs746286687 Chr.9: 4712990-4712990 on Build GRCh38
rs763458227 Chr.9: 4713015-4713015 on Build GRCh38
rs778493453 Chr.9: 4713141-4713141 on Build GRCh38
rs751661410 Chr.9: 4713105-4713105 on Build GRCh38
rs764426366 Chr.9: 4713102-4713102 on Build GRCh38
rs1018654999 Chr.9: 4712924-4712924 on Build GRCh38
rs763073392 Chr.9: 4713099-4713099 on Build GRCh38
rs774126176 Chr.9: 4713196-4713197 on Build GRCh38
rs539106203 Chr.9: 4712854-4712854 on Build GRCh38
rs749337725 Chr.9: 4713052-4713054 on Build GRCh38
rs764442201 Chr.9: 4713018-4713018 on Build GRCh38
rs574591271 Chr.9: 4712936-4712936 on Build GRCh38
rs955540786 Chr.9: 4712903-4712903 on Build GRCh38
rs770635351 Chr.9: 4713022-4713022 on Build GRCh38
rs1049978544 Chr.9: 4713010-4713010 on Build GRCh38
rs140003401 Chr.9: 4713058-4713058 on Build GRCh38
rs777122333 Chr.9: 4712987-4712987 on Build GRCh38
rs891413181 Chr.9: 4713195-4713195 on Build GRCh38
rs148528033 Chr.9: 4713045-4713045 on Build GRCh38
rs766217778 Chr.9: 4713032-4713032 on Build GRCh38
rs780862870 Chr.9: 4713096-4713096 on Build GRCh38
rs773707269 Chr.9: 4713072-4713072 on Build GRCh38
rs968230081 Chr.9: 4713184-4713184 on Build GRCh38
rs950197300 Chr.9: 4712827-4712827 on Build GRCh38
rs371011526 Chr.9: 4712943-4712943 on Build GRCh38
rs117224155 Chr.9: 4713164-4713164 on Build GRCh38

View Full Product Details