Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 4722216-4722723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK3
Entrez Gene ID
50808
Gene Name
adenylate kinase 3
Gene Aliases
AK3L1, AK6, AKL3L, AKL3L1, FIX
UniGene
Hs.732022
Gene Chromosome Location
Chr.9: 4709556-4742043 on Build GRCh38
Transcripts
NM_001199852.1 NM_001199853.1 NM_001199855.1 NM_001199856.1 NM_016282.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
304405 c.226C>T p.L76F Chr.9: 4722551-4722551 on Build GRCh38
1462546 c.159C>T p.G53G Chr.9: 4722618-4722618 on Build GRCh38
5041907 c.271+1G>A p.? Chr.9: 4722505-4722505 on Build GRCh38
6597341 c.158G>T p.G53V Chr.9: 4722619-4722619 on Build GRCh38
69597 c.213G>C p.M71I Chr.9: 4722564-4722564 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs895518931 Chr.9: 4722429-4722429 on Build GRCh38
rs2274820 Chr.9: 4722318-4722318 on Build GRCh38
rs756993396 Chr.9: 4722473-4722473 on Build GRCh38
rs760470441 Chr.9: 4722631-4722631 on Build GRCh38
rs148233791 Chr.9: 4722554-4722554 on Build GRCh38
rs896522996 Chr.9: 4722276-4722276 on Build GRCh38
rs377403539 Chr.9: 4722560-4722560 on Build GRCh38
rs75894792 Chr.9: 4722420-4722420 on Build GRCh38
rs370765513 Chr.9: 4722519-4722519 on Build GRCh38
rs983950419 Chr.9: 4722260-4722260 on Build GRCh38
rs769554300 Chr.9: 4722530-4722530 on Build GRCh38
rs979242238 Chr.9: 4722642-4722642 on Build GRCh38
rs764256078 Chr.9: 4722644-4722644 on Build GRCh38
rs915477851 Chr.9: 4722360-4722360 on Build GRCh38
rs905627210 Chr.9: 4722392-4722392 on Build GRCh38
rs747550190 Chr.9: 4722527-4722527 on Build GRCh38
rs1045103147 Chr.9: 4722476-4722476 on Build GRCh38
rs921705309 Chr.9: 4722390-4722390 on Build GRCh38
rs905350349 Chr.9: 4722314-4722314 on Build GRCh38
rs758783814 Chr.9: 4722693-4722693 on Build GRCh38
rs201139373 Chr.9: 4722464-4722464 on Build GRCh38
rs774227130 Chr.9: 4722559-4722559 on Build GRCh38
rs964140114 Chr.9: 4722283-4722283 on Build GRCh38
rs951727036 Chr.9: 4722337-4722337 on Build GRCh38
rs146101604 Chr.9: 4722610-4722610 on Build GRCh38
rs10758645 Chr.9: 4722685-4722685 on Build GRCh38
rs189771264 Chr.9: 4722599-4722599 on Build GRCh38
rs374594568 Chr.9: 4722622-4722622 on Build GRCh38
rs757805581 Chr.9: 4722658-4722658 on Build GRCh38
rs1015001787 Chr.9: 4722352-4722352 on Build GRCh38
rs905297937 Chr.9: 4722517-4722517 on Build GRCh38
rs758811386 Chr.9: 4722515-4722515 on Build GRCh38
rs899704484 Chr.9: 4722699-4722699 on Build GRCh38
rs199938754 Chr.9: 4722682-4722682 on Build GRCh38
rs202157194 Chr.9: 4722637-4722637 on Build GRCh38
rs12555579 Chr.9: 4722242-4722242 on Build GRCh38
rs755288788 Chr.9: 4722494-4722494 on Build GRCh38
rs574801486 Chr.9: 4722677-4722677 on Build GRCh38
rs752053724 Chr.9: 4722598-4722598 on Build GRCh38
rs184922072 Chr.9: 4722403-4722403 on Build GRCh38
rs373608693 Chr.9: 4722696-4722696 on Build GRCh38
rs535104701 Chr.9: 4722638-4722638 on Build GRCh38
rs763982122 Chr.9: 4722554-4722554 on Build GRCh38
rs976753070 Chr.9: 4722306-4722306 on Build GRCh38
rs767308470 Chr.9: 4722475-4722475 on Build GRCh38
rs956592317 Chr.9: 4722684-4722684 on Build GRCh38
rs563499214 Chr.9: 4722366-4722366 on Build GRCh38
rs761261048 Chr.9: 4722562-4722562 on Build GRCh38
rs925184020 Chr.9: 4722411-4722411 on Build GRCh38
rs774828441 Chr.9: 4722545-4722545 on Build GRCh38
rs750302001 Chr.9: 4722564-4722564 on Build GRCh38
rs752236904 Chr.9: 4722618-4722618 on Build GRCh38
rs752864043 Chr.9: 4722499-4722499 on Build GRCh38
rs367908486 Chr.9: 4722402-4722402 on Build GRCh38
rs372869525 Chr.9: 4722526-4722526 on Build GRCh38
rs766118936 Chr.9: 4722632-4722632 on Build GRCh38
rs141767297 Chr.9: 4722574-4722574 on Build GRCh38
rs772862853 Chr.9: 4722625-4722625 on Build GRCh38
rs998305999 Chr.9: 4722531-4722531 on Build GRCh38
rs916976792 Chr.9: 4722664-4722664 on Build GRCh38
rs112690046 Chr.9: 4722628-4722628 on Build GRCh38
rs1052394727 Chr.9: 4722444-4722444 on Build GRCh38
rs993789454 Chr.9: 4722436-4722436 on Build GRCh38
rs768173564 Chr.9: 4722558-4722558 on Build GRCh38
rs762694618 Chr.9: 4722535-4722538 on Build GRCh38
rs369322522 Chr.9: 4722692-4722692 on Build GRCh38
rs1025538039 Chr.9: 4722442-4722442 on Build GRCh38
rs1045154926 Chr.9: 4722295-4722295 on Build GRCh38
rs922564362 Chr.9: 4722315-4722315 on Build GRCh38
rs75542723 Chr.9: 4722398-4722398 on Build GRCh38
rs753358552 Chr.9: 4722369-4722369 on Build GRCh38
rs779704775 Chr.9: 4722607-4722607 on Build GRCh38
rs138113344 Chr.9: 4722594-4722594 on Build GRCh38
rs1018790782 Chr.9: 4722277-4722277 on Build GRCh38
rs757280003 Chr.9: 4722652-4722653 on Build GRCh38
rs775722325 Chr.9: 4722338-4722338 on Build GRCh38
rs764056709 Chr.9: 4722463-4722463 on Build GRCh38
rs769305592 Chr.9: 4722605-4722605 on Build GRCh38
rs146261577 Chr.9: 4722577-4722577 on Build GRCh38
rs577270589 Chr.9: 4722379-4722379 on Build GRCh38
rs773912544 Chr.9: 4722617-4722617 on Build GRCh38
rs796972974 Chr.9: 4722355-4722362 on Build GRCh38
rs751338127 Chr.9: 4722641-4722642 on Build GRCh38
rs113217148 Chr.9: 4722364-4722364 on Build GRCh38
rs780980928 Chr.9: 4722292-4722292 on Build GRCh38
rs148328480 Chr.9: 4722648-4722648 on Build GRCh38
rs775274025 Chr.9: 4722508-4722509 on Build GRCh38
rs753953279 Chr.9: 4722479-4722479 on Build GRCh38
rs561982310 Chr.9: 4722466-4722466 on Build GRCh38
rs370754329 Chr.9: 4722575-4722575 on Build GRCh38
rs545310859 Chr.9: 4722376-4722376 on Build GRCh38
rs749468628 Chr.9: 4722367-4722367 on Build GRCh38
rs745492011 Chr.9: 4722529-4722529 on Build GRCh38
rs750913919 Chr.9: 4722662-4722662 on Build GRCh38
rs988433029 Chr.9: 4722351-4722351 on Build GRCh38
rs766309182 Chr.9: 4722372-4722372 on Build GRCh38
rs768117144 Chr.9: 4722661-4722661 on Build GRCh38
rs2274821 Chr.9: 4722462-4722462 on Build GRCh38
rs780281905 Chr.9: 4722675-4722675 on Build GRCh38
rs1056847522 Chr.9: 4722650-4722650 on Build GRCh38
rs775122125 Chr.9: 4722456-4722456 on Build GRCh38
rs756457506 Chr.9: 4722671-4722671 on Build GRCh38
rs781699339 Chr.9: 4722528-4722528 on Build GRCh38
rs748919341 Chr.9: 4722609-4722609 on Build GRCh38
rs574764553 Chr.9: 4722326-4722326 on Build GRCh38

View Full Product Details