Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 4740737-4741235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK3
Entrez Gene ID
50808
Gene Name
adenylate kinase 3
Gene Aliases
AK3L1, AK6, AKL3L, AKL3L1, FIX
UniGene
Hs.732022
Gene Chromosome Location
Chr.9: 4709556-4742043 on Build GRCh38
Transcripts
NM_001199852.1 NM_001199853.1 NM_001199855.1 NM_016282.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4884591 c.19C>A p.L7M Chr.9: 4741069-4741069 on Build GRCh38
6597342 c.77G>A p.R26H Chr.9: 4741011-4741011 on Build GRCh38
4822892 c.53C>G p.S18W Chr.9: 4741035-4741035 on Build GRCh38
353645 c.99G>C p.L33L Chr.9: 4740989-4740989 on Build GRCh38
5092612 c.7delG p.A3fs*9 Chr.9: 4741081-4741081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560090929 Chr.9: 4740818-4740818 on Build GRCh38
rs754695612 Chr.9: 4740887-4740887 on Build GRCh38
rs1052115359 Chr.9: 4741135-4741135 on Build GRCh38
rs764448561 Chr.9: 4740914-4740914 on Build GRCh38
rs892243556 Chr.9: 4741039-4741044 on Build GRCh38
rs761932600 Chr.9: 4740923-4740923 on Build GRCh38
rs374558108 Chr.9: 4740922-4740922 on Build GRCh38
rs750870747 Chr.9: 4740930-4740930 on Build GRCh38
rs999184780 Chr.9: 4741126-4741126 on Build GRCh38
rs367566616 Chr.9: 4741114-4741114 on Build GRCh38
rs568768085 Chr.9: 4740971-4740971 on Build GRCh38
rs148762993 Chr.9: 4741071-4741071 on Build GRCh38
rs757859480 Chr.9: 4740961-4740961 on Build GRCh38
rs149999263 Chr.9: 4741042-4741042 on Build GRCh38
rs774671664 Chr.9: 4740970-4740970 on Build GRCh38
rs771506827 Chr.9: 4740905-4740905 on Build GRCh38
rs562565613 Chr.9: 4740855-4740855 on Build GRCh38
rs765577063 Chr.9: 4740814-4740814 on Build GRCh38
rs11539608 Chr.9: 4741078-4741078 on Build GRCh38
rs377642074 Chr.9: 4740770-4740770 on Build GRCh38
rs538104678 Chr.9: 4741143-4741143 on Build GRCh38
rs948030788 Chr.9: 4740840-4740840 on Build GRCh38
rs997957122 Chr.9: 4740873-4740873 on Build GRCh38
rs374607747 Chr.9: 4740993-4740993 on Build GRCh38
rs967802771 Chr.9: 4740804-4740804 on Build GRCh38
rs776535386 Chr.9: 4740966-4740966 on Build GRCh38
rs531405820 Chr.9: 4741022-4741022 on Build GRCh38
rs992624883 Chr.9: 4740809-4740809 on Build GRCh38
rs1049091251 Chr.9: 4740824-4740824 on Build GRCh38
rs540349658 Chr.9: 4741173-4741173 on Build GRCh38
rs750294319 Chr.9: 4741044-4741046 on Build GRCh38
rs995444910 Chr.9: 4741183-4741183 on Build GRCh38
rs773163326 Chr.9: 4740906-4740906 on Build GRCh38
rs960852830 Chr.9: 4740915-4740915 on Build GRCh38
rs1001865293 Chr.9: 4741216-4741216 on Build GRCh38
rs1009226807 Chr.9: 4740894-4740894 on Build GRCh38
rs767572914 Chr.9: 4740977-4740977 on Build GRCh38
rs760032497 Chr.9: 4741111-4741111 on Build GRCh38
rs763067596 Chr.9: 4740788-4740788 on Build GRCh38
rs780396403 Chr.9: 4740973-4740976 on Build GRCh38
rs773321603 Chr.9: 4741035-4741035 on Build GRCh38
rs933506411 Chr.9: 4741019-4741019 on Build GRCh38
rs992438100 Chr.9: 4741101-4741101 on Build GRCh38
rs929401691 Chr.9: 4740955-4740955 on Build GRCh38
rs540263976 Chr.9: 4740810-4740810 on Build GRCh38
rs751245203 Chr.9: 4741067-4741067 on Build GRCh38
rs754243455 Chr.9: 4740942-4740942 on Build GRCh38
rs763416138 Chr.9: 4740913-4740913 on Build GRCh38
rs912692851 Chr.9: 4741095-4741095 on Build GRCh38
rs529221670 Chr.9: 4740911-4740911 on Build GRCh38
rs767519584 Chr.9: 4740919-4740919 on Build GRCh38
rs1043749432 Chr.9: 4741123-4741123 on Build GRCh38
rs945145584 Chr.9: 4741205-4741205 on Build GRCh38
rs370541411 Chr.9: 4740910-4740910 on Build GRCh38
rs773236307 Chr.9: 4740978-4740978 on Build GRCh38
rs756641369 Chr.9: 4740936-4740936 on Build GRCh38
rs746926143 Chr.9: 4740907-4740907 on Build GRCh38
rs777333105 Chr.9: 4741005-4741005 on Build GRCh38
rs888066099 Chr.9: 4740845-4740845 on Build GRCh38
rs766846196 Chr.9: 4740983-4740983 on Build GRCh38
rs754367908 Chr.9: 4740992-4740992 on Build GRCh38
rs892194976 Chr.9: 4740881-4740881 on Build GRCh38
rs1039904205 Chr.9: 4741102-4741102 on Build GRCh38
rs558314823 Chr.9: 4741156-4741156 on Build GRCh38
rs1009236632 Chr.9: 4741040-4741040 on Build GRCh38
rs749089910 Chr.9: 4741082-4741082 on Build GRCh38
rs951409898 Chr.9: 4741079-4741079 on Build GRCh38
rs767522337 Chr.9: 4740927-4740927 on Build GRCh38
rs747708086 Chr.9: 4740899-4740899 on Build GRCh38
rs533872729 Chr.9: 4741080-4741080 on Build GRCh38
rs139274633 Chr.9: 4741023-4741023 on Build GRCh38
rs745705265 Chr.9: 4740967-4740967 on Build GRCh38
rs975965033 Chr.9: 4741207-4741207 on Build GRCh38
rs747830739 Chr.9: 4740959-4740959 on Build GRCh38
rs777057896 Chr.9: 4741045-4741045 on Build GRCh38
rs185585254 Chr.9: 4740781-4740781 on Build GRCh38
rs752288305 Chr.9: 4741000-4741000 on Build GRCh38
rs915182461 Chr.9: 4740778-4740778 on Build GRCh38
rs373732889 Chr.9: 4741103-4741103 on Build GRCh38
rs368574635 Chr.9: 4741048-4741048 on Build GRCh38
rs751069757 Chr.9: 4741007-4741007 on Build GRCh38
rs916540713 Chr.9: 4740820-4740820 on Build GRCh38
rs376326632 Chr.9: 4740939-4740939 on Build GRCh38
rs765592032 Chr.9: 4740998-4740998 on Build GRCh38
rs753126260 Chr.9: 4741058-4741058 on Build GRCh38
rs747260927 Chr.9: 4741091-4741091 on Build GRCh38
rs770840297 Chr.9: 4740964-4740964 on Build GRCh38
rs202145269 Chr.9: 4741002-4741002 on Build GRCh38
rs1023486115 Chr.9: 4741160-4741160 on Build GRCh38
rs573704398 Chr.9: 4741204-4741204 on Build GRCh38
rs982806950 Chr.9: 4741202-4741202 on Build GRCh38
rs778490978 Chr.9: 4740946-4740946 on Build GRCh38
rs575849424 Chr.9: 4741140-4741140 on Build GRCh38
rs779967886 Chr.9: 4741081-4741081 on Build GRCh38
rs776750541 Chr.9: 4741110-4741110 on Build GRCh38
rs544752505 Chr.9: 4740780-4740780 on Build GRCh38
rs185573888 Chr.9: 4741097-4741097 on Build GRCh38
rs748401164 Chr.9: 4741026-4741026 on Build GRCh38
rs1026938850 Chr.9: 4741188-4741188 on Build GRCh38
rs987809103 Chr.9: 4740773-4740773 on Build GRCh38
rs560178647 Chr.9: 4741193-4741193 on Build GRCh38
rs200329877 Chr.9: 4741089-4741089 on Build GRCh38
rs370723156 Chr.9: 4740980-4740980 on Build GRCh38
rs947664432 Chr.9: 4741115-4741115 on Build GRCh38
rs763177471 Chr.9: 4741118-4741118 on Build GRCh38
rs901983614 Chr.9: 4740872-4740872 on Build GRCh38
rs781223984 Chr.9: 4741021-4741021 on Build GRCh38
rs578053372 Chr.9: 4741157-4741157 on Build GRCh38
rs763700487 Chr.9: 4741064-4741064 on Build GRCh38
rs1047223148 Chr.9: 4740900-4740900 on Build GRCh38
rs757194911 Chr.9: 4741012-4741012 on Build GRCh38
rs144288990 Chr.9: 4741049-4741049 on Build GRCh38
rs553834152 Chr.9: 4741085-4741085 on Build GRCh38
rs533238384 Chr.9: 4741124-4741124 on Build GRCh38
rs939470353 Chr.9: 4740799-4740799 on Build GRCh38
rs965506392 Chr.9: 4741205-4741205 on Build GRCh38
rs760626418 Chr.9: 4741037-4741037 on Build GRCh38
rs142691641 Chr.9: 4740962-4740962 on Build GRCh38
rs999236879 Chr.9: 4740988-4740988 on Build GRCh38
rs775812686 Chr.9: 4740861-4740861 on Build GRCh38
rs778366515 Chr.9: 4740898-4740898 on Build GRCh38
rs974130363 Chr.9: 4740856-4740856 on Build GRCh38
rs764206199 Chr.9: 4740854-4740854 on Build GRCh38
rs969671955 Chr.9: 4741163-4741163 on Build GRCh38
rs902078235 Chr.9: 4740997-4740997 on Build GRCh38
rs1013443071 Chr.9: 4741034-4741034 on Build GRCh38
rs909224178 Chr.9: 4741144-4741144 on Build GRCh38
rs775899441 Chr.9: 4741116-4741116 on Build GRCh38
rs960576330 Chr.9: 4740817-4740817 on Build GRCh38
rs967935388 Chr.9: 4740779-4740779 on Build GRCh38
rs201009389 Chr.9: 4741075-4741075 on Build GRCh38
rs759854674 Chr.9: 4740994-4740994 on Build GRCh38
rs150823530 Chr.9: 4741015-4741015 on Build GRCh38
rs189245844 Chr.9: 4740783-4740783 on Build GRCh38
rs762634528 Chr.9: 4741063-4741063 on Build GRCh38

View Full Product Details