Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 5022001-5022499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INSL6
Entrez Gene ID
11172
Gene Name
insulin like 6
Gene Aliases
RIF1
UniGene
Hs.632648
Gene Chromosome Location
Chr.9: 4992408-5185706 on Build GRCh38
Transcripts
XM_011517702.2 NM_001322194.1 NM_001322195.1 NM_001322196.1 NM_001322198.1 NM_001322199.1 NM_004972.3

Gene Symbol
JAK2
Entrez Gene ID
3717
Gene Name
Janus kinase 2
Gene Aliases
JTK10, THCYT3
UniGene
Hs.656213
Gene Chromosome Location
Chr.9: 4985086-5128183 on Build GRCh38
Transcripts
XM_011517702.2 NM_001322194.1 NM_001322195.1 NM_001322196.1 NM_001322198.1 NM_001322199.1 NM_004972.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4986864 c.62A>G p.Q21R Chr.9: 5022049-5022049 on Build GRCh38
4138959 c.109C>G p.P37A Chr.9: 5022096-5022096 on Build GRCh38
455996 c.164T>C p.L55P Chr.9: 5022151-5022151 on Build GRCh38
6949306 c.33G>T p.M11I Chr.9: 5022020-5022020 on Build GRCh38
455995 c.132C>A p.Y44* Chr.9: 5022119-5022119 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897433600 Chr.9: 5022267-5022267 on Build GRCh38
rs1022049606 Chr.9: 5022283-5022283 on Build GRCh38
rs369833874 Chr.9: 5022141-5022141 on Build GRCh38
rs745366597 Chr.9: 5022442-5022442 on Build GRCh38
rs968235029 Chr.9: 5022287-5022287 on Build GRCh38
rs774158685 Chr.9: 5022134-5022134 on Build GRCh38
rs200322825 Chr.9: 5022031-5022031 on Build GRCh38
rs754086152 Chr.9: 5022163-5022163 on Build GRCh38
rs139087749 Chr.9: 5022172-5022172 on Build GRCh38
rs899674385 Chr.9: 5022222-5022223 on Build GRCh38
rs919479063 Chr.9: 5022335-5022335 on Build GRCh38
rs138655335 Chr.9: 5022124-5022124 on Build GRCh38
rs572697100 Chr.9: 5022294-5022294 on Build GRCh38
rs577082095 Chr.9: 5022420-5022420 on Build GRCh38
rs772519132 Chr.9: 5022065-5022065 on Build GRCh38
rs926799779 Chr.9: 5022323-5022323 on Build GRCh38
rs369748023 Chr.9: 5022113-5022113 on Build GRCh38
rs772608321 Chr.9: 5022358-5022358 on Build GRCh38
rs767076792 Chr.9: 5022140-5022140 on Build GRCh38
rs757639029 Chr.9: 5022184-5022184 on Build GRCh38
rs771216385 Chr.9: 5022099-5022099 on Build GRCh38
rs767273014 Chr.9: 5022158-5022158 on Build GRCh38
rs773146013 Chr.9: 5022180-5022180 on Build GRCh38
rs759331856 Chr.9: 5022119-5022119 on Build GRCh38
rs906594522 Chr.9: 5022045-5022045 on Build GRCh38
rs773649932 Chr.9: 5022379-5022379 on Build GRCh38
rs755035784 Chr.9: 5022054-5022054 on Build GRCh38
rs775309638 Chr.9: 5022230-5022230 on Build GRCh38
rs538474116 Chr.9: 5022085-5022085 on Build GRCh38
rs554149143 Chr.9: 5022298-5022298 on Build GRCh38
rs769371550 Chr.9: 5022460-5022460 on Build GRCh38
rs776336648 Chr.9: 5022110-5022110 on Build GRCh38
rs779030695 Chr.9: 5022055-5022055 on Build GRCh38
rs1039077144 Chr.9: 5022395-5022395 on Build GRCh38
rs759031245 Chr.9: 5022025-5022025 on Build GRCh38
rs769518348 Chr.9: 5022227-5022227 on Build GRCh38
rs780253547 Chr.9: 5022052-5022052 on Build GRCh38
rs748500115 Chr.9: 5022097-5022097 on Build GRCh38
rs761588246 Chr.9: 5022091-5022091 on Build GRCh38
rs748737601 Chr.9: 5022231-5022231 on Build GRCh38
rs745497028 Chr.9: 5022218-5022218 on Build GRCh38
rs768062555 Chr.9: 5022233-5022233 on Build GRCh38
rs765467595 Chr.9: 5022170-5022170 on Build GRCh38
rs980512819 Chr.9: 5022288-5022288 on Build GRCh38
rs989175828 Chr.9: 5022401-5022401 on Build GRCh38
rs1009626169 Chr.9: 5022276-5022276 on Build GRCh38
rs778174935 Chr.9: 5022081-5022081 on Build GRCh38
rs929324978 Chr.9: 5022349-5022349 on Build GRCh38
rs575013582 Chr.9: 5022209-5022209 on Build GRCh38
rs150159583 Chr.9: 5022042-5022042 on Build GRCh38
rs763955675 Chr.9: 5022035-5022035 on Build GRCh38
rs954453682 Chr.9: 5022291-5022291 on Build GRCh38
rs182383262 Chr.9: 5022274-5022274 on Build GRCh38
rs899707345 Chr.9: 5022413-5022413 on Build GRCh38
rs762570559 Chr.9: 5022129-5022129 on Build GRCh38
rs913803119 Chr.9: 5022406-5022406 on Build GRCh38
rs150829797 Chr.9: 5022332-5022332 on Build GRCh38
rs761608925 Chr.9: 5022144-5022144 on Build GRCh38
rs957936164 Chr.9: 5022394-5022394 on Build GRCh38
rs927790168 Chr.9: 5022463-5022463 on Build GRCh38
rs143227399 Chr.9: 5022130-5022130 on Build GRCh38
rs774331379 Chr.9: 5022244-5022244 on Build GRCh38
rs763029603 Chr.9: 5022032-5022032 on Build GRCh38
rs764901971 Chr.9: 5022028-5022028 on Build GRCh38
rs544252806 Chr.9: 5022462-5022462 on Build GRCh38
rs778287602 Chr.9: 5022179-5022179 on Build GRCh38
rs781453873 Chr.9: 5022212-5022212 on Build GRCh38
rs998283965 Chr.9: 5022270-5022270 on Build GRCh38
rs551194689 Chr.9: 5022204-5022204 on Build GRCh38
rs1029983116 Chr.9: 5022234-5022234 on Build GRCh38
rs748622306 Chr.9: 5022064-5022064 on Build GRCh38
rs537424087 Chr.9: 5022217-5022217 on Build GRCh38
rs1051041681 Chr.9: 5022438-5022438 on Build GRCh38
rs879845794 Chr.9: 5022318-5022318 on Build GRCh38
rs558390002 Chr.9: 5022356-5022356 on Build GRCh38
rs758479739 Chr.9: 5022174-5022174 on Build GRCh38
rs761721827 Chr.9: 5022261-5022261 on Build GRCh38
rs149333413 Chr.9: 5022128-5022128 on Build GRCh38

View Full Product Details