Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 176864439-176864943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASTN1
Entrez Gene ID
460
Gene Name
astrotactin 1
Gene Aliases
ASTN
UniGene
Hs.495897
Gene Chromosome Location
Chr.1: 176855118-177164904 on Build GRCh38
Transcripts
NM_001286164.1 NM_004319.2 XM_017001340.1 XM_017001341.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4946223 c.3697C>A p.L1233I Chr.1: 176864496-176864496 on Build GRCh38
3478946 c.3648G>A p.R1216R Chr.1: 176864521-176864521 on Build GRCh38
7414398 c.3721A>T p.N1241Y Chr.1: 176864472-176864472 on Build GRCh38
5452683 c.3717C>A p.P1239P Chr.1: 176864452-176864452 on Build GRCh38
463472 c.3730C>T p.R1244W Chr.1: 176864439-176864439 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs79858089 Chr.1: 176864688-176864689 on Build GRCh38
rs1030436034 Chr.1: 176864919-176864919 on Build GRCh38
rs916256430 Chr.1: 176864608-176864608 on Build GRCh38
rs997519135 Chr.1: 176864826-176864826 on Build GRCh38
rs78225550 Chr.1: 176864691-176864693 on Build GRCh38
rs181806963 Chr.1: 176864482-176864482 on Build GRCh38
rs147625964 Chr.1: 176864508-176864508 on Build GRCh38
rs759681664 Chr.1: 176864546-176864550 on Build GRCh38
rs544183517 Chr.1: 176864911-176864911 on Build GRCh38
rs900543121 Chr.1: 176864815-176864815 on Build GRCh38
rs773916714 Chr.1: 176864687-176864691 on Build GRCh38
rs769906645 Chr.1: 176864536-176864536 on Build GRCh38
rs948372495 Chr.1: 176864708-176864711 on Build GRCh38
rs141553974 Chr.1: 176864688-176864692 on Build GRCh38
rs776677973 Chr.1: 176864511-176864511 on Build GRCh38
rs900530655 Chr.1: 176864890-176864890 on Build GRCh38
rs934541206 Chr.1: 176864689-176864689 on Build GRCh38
rs963596613 Chr.1: 176864593-176864593 on Build GRCh38
rs949078008 Chr.1: 176864622-176864622 on Build GRCh38
rs1030419322 Chr.1: 176864487-176864487 on Build GRCh38
rs561802069 Chr.1: 176864685-176864685 on Build GRCh38
rs1004730992 Chr.1: 176864895-176864895 on Build GRCh38
rs955202445 Chr.1: 176864648-176864648 on Build GRCh38
rs866701769 Chr.1: 176864477-176864477 on Build GRCh38
rs386637032 Chr.1: 176864688-176864693 on Build GRCh38
rs886651000 Chr.1: 176864548-176864548 on Build GRCh38
rs982301767 Chr.1: 176864646-176864646 on Build GRCh38
rs1048327503 Chr.1: 176864789-176864789 on Build GRCh38
rs199889735 Chr.1: 176864506-176864506 on Build GRCh38
rs893404892 Chr.1: 176864711-176864711 on Build GRCh38
rs577026489 Chr.1: 176864498-176864498 on Build GRCh38
rs950565411 Chr.1: 176864500-176864500 on Build GRCh38
rs990077627 Chr.1: 176864690-176864690 on Build GRCh38
rs754856836 Chr.1: 176864600-176864600 on Build GRCh38
rs868785371 Chr.1: 176864522-176864522 on Build GRCh38
rs938865973 Chr.1: 176864759-176864759 on Build GRCh38
rs778850800 Chr.1: 176864665-176864665 on Build GRCh38
rs1006922816 Chr.1: 176864808-176864808 on Build GRCh38
rs974846515 Chr.1: 176864612-176864612 on Build GRCh38
rs553179262 Chr.1: 176864617-176864617 on Build GRCh38
rs1006228148 Chr.1: 176864562-176864562 on Build GRCh38
rs775374543 Chr.1: 176864546-176864546 on Build GRCh38
rs1016193585 Chr.1: 176864590-176864590 on Build GRCh38
rs763241906 Chr.1: 176864566-176864566 on Build GRCh38
rs983250712 Chr.1: 176864572-176864572 on Build GRCh38
rs1030429418 Chr.1: 176864872-176864872 on Build GRCh38
rs189605323 Chr.1: 176864802-176864802 on Build GRCh38
rs947657181 Chr.1: 176864728-176864728 on Build GRCh38
rs746012262 Chr.1: 176864525-176864525 on Build GRCh38
rs185089201 Chr.1: 176864671-176864671 on Build GRCh38
rs906933075 Chr.1: 176864739-176864739 on Build GRCh38
rs759902061 Chr.1: 176864707-176864708 on Build GRCh38
rs1044154766 Chr.1: 176864708-176864708 on Build GRCh38
rs777653388 Chr.1: 176864496-176864496 on Build GRCh38
rs371440390 Chr.1: 176864693-176864693 on Build GRCh38
rs201237272 Chr.1: 176864687-176864688 on Build GRCh38
rs764185818 Chr.1: 176864567-176864567 on Build GRCh38
rs746751064 Chr.1: 176864501-176864501 on Build GRCh38
rs572992538 Chr.1: 176864621-176864621 on Build GRCh38
rs886485277 Chr.1: 176864878-176864878 on Build GRCh38

View Full Product Details