Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 177229758-177230280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRINP2
Entrez Gene ID
57795
Gene Name
BMP/retinoic acid inducible neural specific 2
Gene Aliases
DBCCR1L2, FAM5B
UniGene
Hs.495918
Gene Chromosome Location
Chr.1: 177170922-177282422 on Build GRCh38
Transcripts
NM_021165.3 XM_005245379.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1166121 c.200G>A p.R67H Chr.1: 177230076-177230076 on Build GRCh38
2094284 c.261G>A p.R87R Chr.1: 177230137-177230137 on Build GRCh38
676835 c.14G>A p.C5Y Chr.1: 177229890-177229890 on Build GRCh38
6060007 c.192C>T p.P64P Chr.1: 177230068-177230068 on Build GRCh38
2094278 c.145G>A p.V49I Chr.1: 177230021-177230021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs79000061 Chr.1: 177230181-177230181 on Build GRCh38
rs777303846 Chr.1: 177230008-177230008 on Build GRCh38
rs138435872 Chr.1: 177230077-177230077 on Build GRCh38
rs757041919 Chr.1: 177230021-177230021 on Build GRCh38
rs16850984 Chr.1: 177230088-177230088 on Build GRCh38
rs182854673 Chr.1: 177230238-177230238 on Build GRCh38
rs920926796 Chr.1: 177230069-177230069 on Build GRCh38
rs775532892 Chr.1: 177229938-177229938 on Build GRCh38
rs558022486 Chr.1: 177230037-177230037 on Build GRCh38
rs986474947 Chr.1: 177230102-177230102 on Build GRCh38
rs775336666 Chr.1: 177230051-177230051 on Build GRCh38
rs912146352 Chr.1: 177229987-177229987 on Build GRCh38
rs760724811 Chr.1: 177229848-177229848 on Build GRCh38
rs746624639 Chr.1: 177230020-177230020 on Build GRCh38
rs755648048 Chr.1: 177229916-177229916 on Build GRCh38
rs140915160 Chr.1: 177229917-177229917 on Build GRCh38
rs535197966 Chr.1: 177229924-177229924 on Build GRCh38
rs920899184 Chr.1: 177229951-177229951 on Build GRCh38
rs748749541 Chr.1: 177229834-177229834 on Build GRCh38
rs267598199 Chr.1: 177229903-177229903 on Build GRCh38
rs930724445 Chr.1: 177230253-177230253 on Build GRCh38
rs770771129 Chr.1: 177229857-177229857 on Build GRCh38
rs147698296 Chr.1: 177229909-177229909 on Build GRCh38
rs372391592 Chr.1: 177229868-177229868 on Build GRCh38
rs759549339 Chr.1: 177229865-177229865 on Build GRCh38
rs777691956 Chr.1: 177229918-177229918 on Build GRCh38
rs1002503609 Chr.1: 177229873-177229873 on Build GRCh38
rs753767194 Chr.1: 177230192-177230192 on Build GRCh38
rs745666848 Chr.1: 177230095-177230095 on Build GRCh38
rs966266136 Chr.1: 177230199-177230199 on Build GRCh38
rs367853087 Chr.1: 177230162-177230162 on Build GRCh38
rs752251952 Chr.1: 177230006-177230006 on Build GRCh38
rs748381466 Chr.1: 177230112-177230112 on Build GRCh38
rs149441185 Chr.1: 177230005-177230005 on Build GRCh38
rs750937304 Chr.1: 177229999-177229999 on Build GRCh38
rs768728535 Chr.1: 177230060-177230060 on Build GRCh38
rs764152225 Chr.1: 177230087-177230088 on Build GRCh38
rs764499091 Chr.1: 177229877-177229877 on Build GRCh38
rs761135465 Chr.1: 177230117-177230117 on Build GRCh38
rs975086864 Chr.1: 177229858-177229858 on Build GRCh38
rs986749782 Chr.1: 177229969-177229969 on Build GRCh38
rs762102096 Chr.1: 177229986-177229986 on Build GRCh38
rs200815682 Chr.1: 177230143-177230143 on Build GRCh38
rs199631180 Chr.1: 177229930-177229930 on Build GRCh38
rs150664972 Chr.1: 177230107-177230107 on Build GRCh38
rs751744392 Chr.1: 177229977-177229977 on Build GRCh38
rs759437536 Chr.1: 177229935-177229935 on Build GRCh38
rs962477177 Chr.1: 177230059-177230059 on Build GRCh38
rs774464143 Chr.1: 177230062-177230062 on Build GRCh38
rs763621067 Chr.1: 177230085-177230085 on Build GRCh38
rs1049073734 Chr.1: 177230050-177230050 on Build GRCh38
rs569593343 Chr.1: 177229907-177229907 on Build GRCh38
rs528623536 Chr.1: 177229824-177229824 on Build GRCh38
rs751983601 Chr.1: 177230078-177230078 on Build GRCh38
rs944890706 Chr.1: 177229990-177229990 on Build GRCh38
rs779789828 Chr.1: 177230191-177230191 on Build GRCh38
rs772863226 Chr.1: 177229847-177229847 on Build GRCh38
rs781673030 Chr.1: 177230165-177230165 on Build GRCh38
rs865789221 Chr.1: 177230145-177230145 on Build GRCh38
rs376274006 Chr.1: 177229867-177229867 on Build GRCh38
rs781018149 Chr.1: 177230036-177230036 on Build GRCh38
rs190682585 Chr.1: 177230216-177230216 on Build GRCh38
rs1012892954 Chr.1: 177229804-177229804 on Build GRCh38
rs757573324 Chr.1: 177229893-177229893 on Build GRCh38
rs972246834 Chr.1: 177230230-177230230 on Build GRCh38
rs3176441 Chr.1: 177229862-177229862 on Build GRCh38
rs756669472 Chr.1: 177230003-177230003 on Build GRCh38
rs147833227 Chr.1: 177229885-177229885 on Build GRCh38
rs768655368 Chr.1: 177230111-177230111 on Build GRCh38
rs377272745 Chr.1: 177230044-177230044 on Build GRCh38
rs779678300 Chr.1: 177229832-177229832 on Build GRCh38
rs546164217 Chr.1: 177230226-177230226 on Build GRCh38
rs763568462 Chr.1: 177230140-177230140 on Build GRCh38
rs139209134 Chr.1: 177230092-177230092 on Build GRCh38
rs553424360 Chr.1: 177230114-177230114 on Build GRCh38
rs765260341 Chr.1: 177229866-177229866 on Build GRCh38
rs112761040 Chr.1: 177230072-177230072 on Build GRCh38
rs767880163 Chr.1: 177229989-177229989 on Build GRCh38
rs752035032 Chr.1: 177229831-177229831 on Build GRCh38
rs756920458 Chr.1: 177230087-177230087 on Build GRCh38
rs924857789 Chr.1: 177229898-177229898 on Build GRCh38
rs569033840 Chr.1: 177229942-177229942 on Build GRCh38
rs151068470 Chr.1: 177229829-177229829 on Build GRCh38
rs367628989 Chr.1: 177229841-177229841 on Build GRCh38
rs890584786 Chr.1: 177229822-177229822 on Build GRCh38
rs1054612397 Chr.1: 177230170-177230170 on Build GRCh38
rs3176442 Chr.1: 177230187-177230187 on Build GRCh38
rs934220861 Chr.1: 177229864-177229864 on Build GRCh38
rs373037602 Chr.1: 177230192-177230192 on Build GRCh38
rs146202383 Chr.1: 177230086-177230086 on Build GRCh38
rs376635257 Chr.1: 177230056-177230056 on Build GRCh38
rs370573955 Chr.1: 177229842-177229842 on Build GRCh38
rs201739935 Chr.1: 177230136-177230136 on Build GRCh38
rs773700754 Chr.1: 177230075-177230075 on Build GRCh38
rs200656202 Chr.1: 177230115-177230115 on Build GRCh38
rs763602017 Chr.1: 177230064-177230064 on Build GRCh38
rs769910100 Chr.1: 177229937-177229937 on Build GRCh38
rs200249029 Chr.1: 177229913-177229913 on Build GRCh38
rs953661445 Chr.1: 177229963-177229963 on Build GRCh38
rs564425687 Chr.1: 177230214-177230214 on Build GRCh38
rs1050628511 Chr.1: 177229819-177229819 on Build GRCh38
rs758005507 Chr.1: 177230082-177230082 on Build GRCh38
rs772035276 Chr.1: 177229843-177229843 on Build GRCh38
rs562095758 Chr.1: 177230173-177230173 on Build GRCh38
rs932200722 Chr.1: 177229805-177229805 on Build GRCh38
rs969260394 Chr.1: 177229818-177229818 on Build GRCh38
rs755843993 Chr.1: 177230105-177230105 on Build GRCh38
rs893220586 Chr.1: 177229798-177229798 on Build GRCh38
rs568417156 Chr.1: 177229921-177229921 on Build GRCh38
rs548466314 Chr.1: 177229825-177229825 on Build GRCh38
rs747854235 Chr.1: 177229914-177229914 on Build GRCh38
rs763137751 Chr.1: 177229875-177229875 on Build GRCh38
rs766803056 Chr.1: 177230119-177230119 on Build GRCh38
rs764224385 Chr.1: 177229958-177229958 on Build GRCh38
rs767825106 Chr.1: 177230065-177230065 on Build GRCh38
rs777310352 Chr.1: 177230128-177230128 on Build GRCh38

View Full Product Details