Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 87974599-87975037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK20
Entrez Gene ID
23552
Gene Name
cyclin dependent kinase 20
Gene Aliases
CCRK, CDCH, P42, PNQALRE
UniGene
Hs.522274
Gene Chromosome Location
Chr.9: 87966441-87974780 on Build GRCh38
Transcripts
NM_001039803.2 NM_001170639.1 NM_001170640.1 NM_012119.4 NM_178432.3 XM_017014561.1 XM_017014562.1 XM_017014563.1 XM_017014564.1

SNP Detail

SNP ID Chromosome Location
rs945030048 Chr.9: 87974897-87974897 on Build GRCh38
rs773052079 Chr.9: 87974669-87974669 on Build GRCh38
rs540223094 Chr.9: 87974829-87974829 on Build GRCh38
rs139380442 Chr.9: 87974944-87974944 on Build GRCh38
rs915059805 Chr.9: 87974763-87974763 on Build GRCh38
rs1029177614 Chr.9: 87974974-87974974 on Build GRCh38
rs1009133184 Chr.9: 87974991-87974991 on Build GRCh38
rs889222605 Chr.9: 87974777-87974777 on Build GRCh38
rs987880420 Chr.9: 87974868-87974868 on Build GRCh38
rs920485884 Chr.9: 87974698-87974698 on Build GRCh38
rs905110872 Chr.9: 87974774-87974774 on Build GRCh38
rs532445573 Chr.9: 87974857-87974857 on Build GRCh38
rs946492302 Chr.9: 87974877-87974877 on Build GRCh38
rs946565991 Chr.9: 87974765-87974765 on Build GRCh38
rs761904712 Chr.9: 87974958-87974958 on Build GRCh38
rs568003663 Chr.9: 87974716-87974716 on Build GRCh38
rs926406167 Chr.9: 87974731-87974731 on Build GRCh38
rs1018883533 Chr.9: 87974657-87974657 on Build GRCh38
rs1045351624 Chr.9: 87974770-87974770 on Build GRCh38
rs994976718 Chr.9: 87974825-87974825 on Build GRCh38
rs1057143941 Chr.9: 87974795-87974795 on Build GRCh38
rs905850832 Chr.9: 87974632-87974632 on Build GRCh38
rs982662221 Chr.9: 87974665-87974665 on Build GRCh38
rs555376182 Chr.9: 87974819-87974819 on Build GRCh38
rs953129495 Chr.9: 87974722-87974722 on Build GRCh38
rs576173609 Chr.9: 87974783-87974783 on Build GRCh38
rs539569736 Chr.9: 87974766-87974766 on Build GRCh38
rs960502929 Chr.9: 87974882-87974882 on Build GRCh38
rs182447501 Chr.9: 87974776-87974776 on Build GRCh38
rs1036143603 Chr.9: 87974664-87974664 on Build GRCh38
rs1008891889 Chr.9: 87974634-87974634 on Build GRCh38
rs987515213 Chr.9: 87974846-87974846 on Build GRCh38
rs767703183 Chr.9: 87974990-87974990 on Build GRCh38
rs28364934 Chr.9: 87974673-87974673 on Build GRCh38
rs939098903 Chr.9: 87974732-87974732 on Build GRCh38
rs1051060249 Chr.9: 87974944-87974945 on Build GRCh38
rs553846365 Chr.9: 87974757-87974757 on Build GRCh38
rs980647707 Chr.9: 87974893-87974893 on Build GRCh38
rs765626161 Chr.9: 87974715-87974715 on Build GRCh38
rs545926199 Chr.9: 87974875-87974875 on Build GRCh38
rs1008922843 Chr.9: 87974778-87974778 on Build GRCh38
rs926501392 Chr.9: 87974898-87974898 on Build GRCh38
rs1045042877 Chr.9: 87974620-87974620 on Build GRCh38
rs940731906 Chr.9: 87974928-87974928 on Build GRCh38
rs1056806885 Chr.9: 87974908-87974908 on Build GRCh38
rs771806437 Chr.9: 87974626-87974626 on Build GRCh38
rs915000717 Chr.9: 87974872-87974872 on Build GRCh38
rs1052213352 Chr.9: 87974762-87974762 on Build GRCh38
rs895815979 Chr.9: 87974923-87974923 on Build GRCh38
rs1028018343 Chr.9: 87974854-87974854 on Build GRCh38
rs567474768 Chr.9: 87974621-87974622 on Build GRCh38
rs373817050 Chr.9: 87974929-87974929 on Build GRCh38
rs993426336 Chr.9: 87974836-87974836 on Build GRCh38
rs1019653521 Chr.9: 87975012-87975012 on Build GRCh38
rs897905705 Chr.9: 87975003-87975003 on Build GRCh38
rs560158135 Chr.9: 87974855-87974855 on Build GRCh38
rs944852208 Chr.9: 87974954-87974954 on Build GRCh38
rs186539701 Chr.9: 87974799-87974799 on Build GRCh38
rs961963098 Chr.9: 87974668-87974668 on Build GRCh38
rs1056174149 Chr.9: 87974756-87974756 on Build GRCh38
rs34581063 Chr.9: 87974760-87974760 on Build GRCh38
rs1004903218 Chr.9: 87974804-87974804 on Build GRCh38
rs1029496928 Chr.9: 87974623-87974623 on Build GRCh38
rs41286033 Chr.9: 87974720-87974720 on Build GRCh38
rs548614722 Chr.9: 87974714-87974714 on Build GRCh38
rs963535170 Chr.9: 87974684-87974684 on Build GRCh38
rs189873388 Chr.9: 87974705-87974705 on Build GRCh38
rs991928920 Chr.9: 87974889-87974889 on Build GRCh38
rs763312791 Chr.9: 87974894-87974894 on Build GRCh38
rs1014999388 Chr.9: 87974805-87974805 on Build GRCh38
rs1001550648 Chr.9: 87974635-87974635 on Build GRCh38
rs911608688 Chr.9: 87974758-87974758 on Build GRCh38
rs531817180 Chr.9: 87974907-87974907 on Build GRCh38
rs574735036 Chr.9: 87974826-87974826 on Build GRCh38
rs562706836 Chr.9: 87974878-87974878 on Build GRCh38
rs956383557 Chr.9: 87974861-87974861 on Build GRCh38

View Full Product Details