Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92500713-92501223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPP
Entrez Gene ID
401541
Gene Name
centromere protein P
Gene Aliases
CENP-P
UniGene
Hs.713775
Gene Chromosome Location
Chr.9: 92325457-92615164 on Build GRCh38
Transcripts
NM_001012267.2 NM_001286969.1 NM_001197295.1 NM_001197296.1 NM_001393.3

Gene Symbol
ECM2
Entrez Gene ID
1842
Gene Name
extracellular matrix protein 2
Gene Aliases
-
UniGene
Hs.117060
Gene Chromosome Location
Chr.9: 92493547-92536092 on Build GRCh38
Transcripts
NM_001012267.2 NM_001286969.1 NM_001197295.1 NM_001197296.1 NM_001393.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3927223 c.1644G>A p.R548R Chr.9: 92500948-92500948 on Build GRCh38
1463594 c.1865C>G p.S622C Chr.9: 92500793-92500793 on Build GRCh38
5546314 c.1628G>A p.G543E Chr.9: 92500964-92500964 on Build GRCh38
456237 c.1652C>T p.P551L Chr.9: 92501006-92501006 on Build GRCh38
291334 c.1831C>A p.L611I Chr.9: 92500761-92500761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759070099 Chr.9: 92500917-92500918 on Build GRCh38
rs748535545 Chr.9: 92500983-92500983 on Build GRCh38
rs778464812 Chr.9: 92500826-92500826 on Build GRCh38
rs776930712 Chr.9: 92500757-92500757 on Build GRCh38
rs942352232 Chr.9: 92500843-92500843 on Build GRCh38
rs199713916 Chr.9: 92501005-92501005 on Build GRCh38
rs755447923 Chr.9: 92501081-92501081 on Build GRCh38
rs1026622587 Chr.9: 92501079-92501079 on Build GRCh38
rs984003366 Chr.9: 92501159-92501159 on Build GRCh38
rs777770493 Chr.9: 92501130-92501130 on Build GRCh38
rs770032853 Chr.9: 92500762-92500762 on Build GRCh38
rs570789332 Chr.9: 92500859-92500859 on Build GRCh38
rs7861257 Chr.9: 92501125-92501125 on Build GRCh38
rs978764730 Chr.9: 92500916-92500917 on Build GRCh38
rs138868045 Chr.9: 92500807-92500807 on Build GRCh38
rs771864666 Chr.9: 92500913-92500913 on Build GRCh38
rs774424324 Chr.9: 92500766-92500766 on Build GRCh38
rs763992107 Chr.9: 92500939-92500939 on Build GRCh38
rs191100785 Chr.9: 92501102-92501102 on Build GRCh38
rs756731840 Chr.9: 92500825-92500825 on Build GRCh38
rs887344082 Chr.9: 92501107-92501107 on Build GRCh38
rs757923206 Chr.9: 92500954-92500954 on Build GRCh38
rs575737141 Chr.9: 92501034-92501034 on Build GRCh38
rs34868349 Chr.9: 92501030-92501031 on Build GRCh38
rs150249574 Chr.9: 92500788-92500788 on Build GRCh38
rs752368753 Chr.9: 92500787-92500787 on Build GRCh38
rs778172021 Chr.9: 92501002-92501002 on Build GRCh38
rs141859598 Chr.9: 92500873-92500873 on Build GRCh38
rs758832338 Chr.9: 92500855-92500855 on Build GRCh38
rs1010689 Chr.9: 92501091-92501091 on Build GRCh38
rs775521249 Chr.9: 92500925-92500925 on Build GRCh38
rs34518968 Chr.9: 92500934-92500934 on Build GRCh38
rs772897322 Chr.9: 92501031-92501031 on Build GRCh38
rs925160568 Chr.9: 92500928-92500937 on Build GRCh38
rs186648835 Chr.9: 92500831-92500831 on Build GRCh38
rs767576492 Chr.9: 92500780-92500780 on Build GRCh38
rs781440673 Chr.9: 92501010-92501010 on Build GRCh38
rs761622714 Chr.9: 92501018-92501018 on Build GRCh38
rs143452414 Chr.9: 92500963-92500963 on Build GRCh38
rs764817722 Chr.9: 92501029-92501029 on Build GRCh38
rs557095017 Chr.9: 92501011-92501011 on Build GRCh38
rs764663897 Chr.9: 92500922-92500923 on Build GRCh38
rs775427154 Chr.9: 92500928-92500929 on Build GRCh38
rs534691103 Chr.9: 92500885-92500885 on Build GRCh38
rs755291775 Chr.9: 92500967-92500967 on Build GRCh38
rs776356270 Chr.9: 92501015-92501015 on Build GRCh38
rs199600306 Chr.9: 92500950-92500950 on Build GRCh38
rs763129979 Chr.9: 92500902-92500902 on Build GRCh38
rs147126273 Chr.9: 92500897-92500897 on Build GRCh38
rs1039336331 Chr.9: 92500846-92500846 on Build GRCh38
rs763121296 Chr.9: 92500897-92500910 on Build GRCh38
rs975493466 Chr.9: 92501164-92501164 on Build GRCh38
rs780400206 Chr.9: 92500955-92500955 on Build GRCh38
rs775261362 Chr.9: 92501006-92501006 on Build GRCh38
rs780638094 Chr.9: 92500857-92500857 on Build GRCh38
rs765973788 Chr.9: 92501056-92501056 on Build GRCh38
rs781662954 Chr.9: 92500979-92500979 on Build GRCh38
rs761774737 Chr.9: 92500758-92500758 on Build GRCh38
rs769980008 Chr.9: 92500894-92500894 on Build GRCh38
rs754777850 Chr.9: 92500992-92500992 on Build GRCh38
rs371112363 Chr.9: 92501076-92501076 on Build GRCh38
rs376628552 Chr.9: 92501039-92501039 on Build GRCh38
rs543477797 Chr.9: 92501086-92501086 on Build GRCh38
rs184512785 Chr.9: 92501133-92501133 on Build GRCh38
rs750921084 Chr.9: 92500839-92500839 on Build GRCh38
rs763015482 Chr.9: 92500932-92500939 on Build GRCh38
rs781238711 Chr.9: 92500879-92500879 on Build GRCh38
rs369582601 Chr.9: 92501087-92501087 on Build GRCh38
rs764243970 Chr.9: 92500746-92500746 on Build GRCh38
rs201745324 Chr.9: 92500847-92500847 on Build GRCh38
rs776211051 Chr.9: 92500860-92500860 on Build GRCh38
rs752007103 Chr.9: 92501067-92501067 on Build GRCh38
rs982943174 Chr.9: 92500819-92500822 on Build GRCh38
rs970729664 Chr.9: 92501101-92501101 on Build GRCh38
rs965965918 Chr.9: 92501141-92501141 on Build GRCh38
rs770074932 Chr.9: 92500999-92500999 on Build GRCh38
rs763606251 Chr.9: 92500803-92500803 on Build GRCh38
rs770551588 Chr.9: 92500848-92500850 on Build GRCh38
rs764887308 Chr.9: 92500947-92500947 on Build GRCh38
rs753431713 Chr.9: 92500943-92500943 on Build GRCh38
rs373764507 Chr.9: 92500945-92500945 on Build GRCh38
rs752980805 Chr.9: 92501106-92501106 on Build GRCh38
rs547551173 Chr.9: 92500785-92500785 on Build GRCh38
rs140976209 Chr.9: 92500776-92500776 on Build GRCh38
rs190254490 Chr.9: 92501200-92501200 on Build GRCh38
rs778108579 Chr.9: 92501100-92501100 on Build GRCh38

View Full Product Details