Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92511923-92512425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPP
Entrez Gene ID
401541
Gene Name
centromere protein P
Gene Aliases
CENP-P
UniGene
Hs.713775
Gene Chromosome Location
Chr.9: 92325457-92615164 on Build GRCh38
Transcripts
NM_001012267.2 NM_001286969.1 NM_001197295.1 NM_001197296.1 NM_001393.3 XM_017014376.1 XM_017014377.1

Gene Symbol
ECM2
Entrez Gene ID
1842
Gene Name
extracellular matrix protein 2
Gene Aliases
-
UniGene
Hs.117060
Gene Chromosome Location
Chr.9: 92493547-92536092 on Build GRCh38
Transcripts
NM_001012267.2 NM_001286969.1 NM_001197295.1 NM_001197296.1 NM_001393.3 XM_017014376.1 XM_017014377.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
753878 c.1084A>G p.I362V Chr.9: 92512031-92512031 on Build GRCh38
1111141 c.1142C>A p.S381Y Chr.9: 92512039-92512039 on Build GRCh38
422318 c.1120C>T p.L374L Chr.9: 92512061-92512061 on Build GRCh38
7291636 c.1085C>T p.A362V Chr.9: 92512096-92512096 on Build GRCh38
6359002 c.1094A>C p.K365T Chr.9: 92512021-92512021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049929177 Chr.9: 92512202-92512202 on Build GRCh38
rs780113287 Chr.9: 92512140-92512140 on Build GRCh38
rs940261808 Chr.9: 92512179-92512179 on Build GRCh38
rs537279656 Chr.9: 92512178-92512179 on Build GRCh38
rs544614974 Chr.9: 92512082-92512082 on Build GRCh38
rs767046393 Chr.9: 92512051-92512051 on Build GRCh38
rs762833336 Chr.9: 92512070-92512070 on Build GRCh38
rs762010252 Chr.9: 92512154-92512154 on Build GRCh38
rs189594615 Chr.9: 92511986-92511986 on Build GRCh38
rs894682254 Chr.9: 92512131-92512131 on Build GRCh38
rs776770919 Chr.9: 92511978-92511978 on Build GRCh38
rs536872293 Chr.9: 92512317-92512317 on Build GRCh38
rs762919894 Chr.9: 92512001-92512001 on Build GRCh38
rs1018603804 Chr.9: 92512277-92512277 on Build GRCh38
rs368447634 Chr.9: 92512141-92512141 on Build GRCh38
rs993514151 Chr.9: 92512228-92512228 on Build GRCh38
rs146339911 Chr.9: 92512024-92512024 on Build GRCh38
rs150792948 Chr.9: 92512182-92512182 on Build GRCh38
rs907464010 Chr.9: 92512081-92512081 on Build GRCh38
rs1010143442 Chr.9: 92512312-92512312 on Build GRCh38
rs1021585210 Chr.9: 92512332-92512332 on Build GRCh38
rs927073042 Chr.9: 92512102-92512102 on Build GRCh38
rs757106445 Chr.9: 92512295-92512295 on Build GRCh38
rs760636627 Chr.9: 92511970-92511970 on Build GRCh38
rs776796835 Chr.9: 92512148-92512148 on Build GRCh38
rs2296667 Chr.9: 92512056-92512056 on Build GRCh38
rs1016387145 Chr.9: 92512177-92512177 on Build GRCh38
rs995592485 Chr.9: 92511997-92511997 on Build GRCh38
rs931513900 Chr.9: 92512199-92512199 on Build GRCh38
rs987050558 Chr.9: 92511977-92511977 on Build GRCh38
rs753894390 Chr.9: 92512271-92512271 on Build GRCh38
rs759408059 Chr.9: 92512038-92512038 on Build GRCh38
rs986693742 Chr.9: 92512256-92512256 on Build GRCh38
rs142186054 Chr.9: 92512115-92512115 on Build GRCh38
rs751352419 Chr.9: 92512036-92512036 on Build GRCh38
rs369961555 Chr.9: 92512351-92512351 on Build GRCh38
rs113355942 Chr.9: 92512386-92512386 on Build GRCh38
rs2296666 Chr.9: 92511979-92511979 on Build GRCh38
rs139225550 Chr.9: 92512255-92512255 on Build GRCh38
rs773575499 Chr.9: 92511950-92511950 on Build GRCh38
rs746176588 Chr.9: 92512116-92512116 on Build GRCh38
rs568094846 Chr.9: 92512380-92512380 on Build GRCh38
rs200699120 Chr.9: 92512142-92512142 on Build GRCh38
rs1056370302 Chr.9: 92512125-92512125 on Build GRCh38
rs73651314 Chr.9: 92512059-92512059 on Build GRCh38
rs1008325317 Chr.9: 92512265-92512265 on Build GRCh38
rs41280065 Chr.9: 92512244-92512244 on Build GRCh38
rs898661164 Chr.9: 92512198-92512198 on Build GRCh38
rs778793161 Chr.9: 92512341-92512341 on Build GRCh38
rs1019602478 Chr.9: 92512282-92512282 on Build GRCh38
rs374663122 Chr.9: 92512137-92512137 on Build GRCh38
rs141995151 Chr.9: 92512000-92512000 on Build GRCh38
rs529992696 Chr.9: 92511985-92511985 on Build GRCh38
rs199972367 Chr.9: 92512030-92512030 on Build GRCh38
rs1030302688 Chr.9: 92512241-92512242 on Build GRCh38
rs980301595 Chr.9: 92512398-92512398 on Build GRCh38
rs757702618 Chr.9: 92512103-92512103 on Build GRCh38
rs988745902 Chr.9: 92512048-92512048 on Build GRCh38
rs772134102 Chr.9: 92511962-92511962 on Build GRCh38
rs775809629 Chr.9: 92511965-92511965 on Build GRCh38
rs954070567 Chr.9: 92512011-92512011 on Build GRCh38
rs546631933 Chr.9: 92512302-92512302 on Build GRCh38
rs1034192305 Chr.9: 92512399-92512399 on Build GRCh38
rs745655359 Chr.9: 92512347-92512347 on Build GRCh38
rs914343898 Chr.9: 92512380-92512381 on Build GRCh38
rs141793116 Chr.9: 92512121-92512121 on Build GRCh38

View Full Product Details