Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 93002966-93003507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGD3
Entrez Gene ID
89846
Gene Name
FYVE, RhoGEF and PH domain containing 3
Gene Aliases
ZFYVE5
UniGene
Hs.411081
Gene Chromosome Location
Chr.9: 92947319-93036236 on Build GRCh38
Transcripts
NM_001083536.1 NM_001286993.1 NM_033086.2 XM_017015274.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6380537 c.530A>G p.H177R Chr.9: 93003001-93003001 on Build GRCh38
1463636 c.523C>T p.R175W Chr.9: 93002994-93002994 on Build GRCh38
456254 c.531C>A p.H177Q Chr.9: 93003002-93003002 on Build GRCh38
5531234 c.508G>A p.E170K Chr.9: 93002979-93002979 on Build GRCh38
6380538 c.530A>G p.H177R Chr.9: 93003001-93003001 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900431511 Chr.9: 93003109-93003109 on Build GRCh38
rs541355148 Chr.9: 93003425-93003425 on Build GRCh38
rs577140695 Chr.9: 93003420-93003420 on Build GRCh38
rs909640761 Chr.9: 93003437-93003437 on Build GRCh38
rs1021210293 Chr.9: 93003035-93003035 on Build GRCh38
rs373012437 Chr.9: 93003414-93003414 on Build GRCh38
rs944505314 Chr.9: 93003408-93003408 on Build GRCh38
rs752591220 Chr.9: 93003049-93003049 on Build GRCh38
rs554416626 Chr.9: 93003239-93003239 on Build GRCh38
rs1005400749 Chr.9: 93003191-93003191 on Build GRCh38
rs759581803 Chr.9: 93003039-93003039 on Build GRCh38
rs900399025 Chr.9: 93003424-93003424 on Build GRCh38
rs981815063 Chr.9: 93003068-93003068 on Build GRCh38
rs186090338 Chr.9: 93003291-93003291 on Build GRCh38
rs189120671 Chr.9: 93003167-93003167 on Build GRCh38
rs933876024 Chr.9: 93003272-93003272 on Build GRCh38
rs139284637 Chr.9: 93003286-93003286 on Build GRCh38
rs887809501 Chr.9: 93003439-93003439 on Build GRCh38
rs746650199 Chr.9: 93003024-93003025 on Build GRCh38
rs113069932 Chr.9: 93003436-93003436 on Build GRCh38
rs376038788 Chr.9: 93003133-93003133 on Build GRCh38
rs1042566813 Chr.9: 93003388-93003388 on Build GRCh38
rs923796590 Chr.9: 93003404-93003404 on Build GRCh38
rs774156647 Chr.9: 93003033-93003033 on Build GRCh38
rs1051002177 Chr.9: 93003282-93003282 on Build GRCh38
rs147120978 Chr.9: 93003473-93003473 on Build GRCh38
rs958956393 Chr.9: 93003288-93003288 on Build GRCh38
rs767340631 Chr.9: 93003046-93003046 on Build GRCh38
rs1040309965 Chr.9: 93003098-93003098 on Build GRCh38
rs201243454 Chr.9: 93003409-93003409 on Build GRCh38
rs946474677 Chr.9: 93003021-93003021 on Build GRCh38
rs755946776 Chr.9: 93003051-93003051 on Build GRCh38
rs747350454 Chr.9: 93002995-93002995 on Build GRCh38
rs775900978 Chr.9: 93002994-93002994 on Build GRCh38
rs751686326 Chr.9: 93003123-93003132 on Build GRCh38
rs755312907 Chr.9: 93003354-93003354 on Build GRCh38
rs763874088 Chr.9: 93003055-93003055 on Build GRCh38
rs765299985 Chr.9: 93003024-93003024 on Build GRCh38
rs571345593 Chr.9: 93003381-93003381 on Build GRCh38
rs959070131 Chr.9: 93003426-93003426 on Build GRCh38
rs981608735 Chr.9: 93003254-93003254 on Build GRCh38
rs770438386 Chr.9: 93003031-93003031 on Build GRCh38
rs761819923 Chr.9: 93003017-93003017 on Build GRCh38
rs890490927 Chr.9: 93003282-93003282 on Build GRCh38
rs914913253 Chr.9: 93003339-93003339 on Build GRCh38
rs536338740 Chr.9: 93003190-93003190 on Build GRCh38
rs962744419 Chr.9: 93003462-93003462 on Build GRCh38
rs781564624 Chr.9: 93003380-93003380 on Build GRCh38
rs990054508 Chr.9: 93003034-93003034 on Build GRCh38
rs145868231 Chr.9: 93003090-93003090 on Build GRCh38
rs79177813 Chr.9: 93003466-93003468 on Build GRCh38
rs918520970 Chr.9: 93003147-93003147 on Build GRCh38
rs1015084967 Chr.9: 93003215-93003215 on Build GRCh38
rs997122811 Chr.9: 93003466-93003466 on Build GRCh38
rs927610159 Chr.9: 93003080-93003080 on Build GRCh38
rs780434476 Chr.9: 93003073-93003073 on Build GRCh38
rs768903942 Chr.9: 93003000-93003000 on Build GRCh38
rs546347417 Chr.9: 93003072-93003072 on Build GRCh38
rs779942091 Chr.9: 93003189-93003189 on Build GRCh38
rs776793657 Chr.9: 93003012-93003012 on Build GRCh38
rs547392364 Chr.9: 93003133-93003133 on Build GRCh38
rs893859220 Chr.9: 93003341-93003341 on Build GRCh38
rs753493925 Chr.9: 93003059-93003059 on Build GRCh38
rs1028083767 Chr.9: 93003169-93003169 on Build GRCh38
rs946472165 Chr.9: 93003378-93003378 on Build GRCh38
rs959016873 Chr.9: 93003096-93003096 on Build GRCh38
rs181454093 Chr.9: 93003253-93003253 on Build GRCh38

View Full Product Details