Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 95100617-95101123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf3
Entrez Gene ID
84909
Gene Name
chromosome 9 open reading frame 3
Gene Aliases
AOPEP, AP-O, APO, C90RF3, ONPEP
UniGene
Hs.434253
Gene Chromosome Location
Chr.9: 94726604-95148264 on Build GRCh38
Transcripts
XM_011519121.2 NM_000136.2 NM_001243743.1 XM_005251802.3 XM_006717001.2 XM_011518365.2 XM_011518367.2 XM_017014452.1 XM_017014453.1 XM_017014454.1

Gene Symbol
FANCC
Entrez Gene ID
2176
Gene Name
Fanconi anemia complementation group C
Gene Aliases
FA3, FAC, FACC
UniGene
Hs.494529
Gene Chromosome Location
Chr.9: 95099054-95317730 on Build GRCh38
Transcripts
XM_011519121.2 NM_000136.2 NM_001243743.1 XM_005251802.3 XM_006717001.2 XM_011518365.2 XM_011518367.2 XM_017014452.1 XM_017014453.1 XM_017014454.1

SNP Detail

SNP ID Chromosome Location
rs931513834 Chr.9: 95100687-95100687 on Build GRCh38
rs919705909 Chr.9: 95100918-95100918 on Build GRCh38
rs945289900 Chr.9: 95100792-95100792 on Build GRCh38
rs1052130713 Chr.9: 95100793-95100793 on Build GRCh38
rs996840650 Chr.9: 95101015-95101015 on Build GRCh38
rs1025115331 Chr.9: 95100711-95100711 on Build GRCh38
rs899912241 Chr.9: 95100841-95100841 on Build GRCh38
rs996871661 Chr.9: 95100761-95100761 on Build GRCh38
rs537983135 Chr.9: 95100662-95100662 on Build GRCh38
rs34302466 Chr.9: 95100648-95100648 on Build GRCh38
rs1026945471 Chr.9: 95100927-95100927 on Build GRCh38
rs4647553 Chr.9: 95100999-95100999 on Build GRCh38
rs956856849 Chr.9: 95100833-95100833 on Build GRCh38
rs971315647 Chr.9: 95100712-95100712 on Build GRCh38
rs554177200 Chr.9: 95100931-95100931 on Build GRCh38
rs983439044 Chr.9: 95100707-95100707 on Build GRCh38
rs964149259 Chr.9: 95100661-95100661 on Build GRCh38
rs936463547 Chr.9: 95100744-95100744 on Build GRCh38
rs1047677594 Chr.9: 95100936-95100936 on Build GRCh38
rs1041307863 Chr.9: 95100949-95100949 on Build GRCh38
rs1038927110 Chr.9: 95100823-95100823 on Build GRCh38
rs1027024042 Chr.9: 95100693-95100693 on Build GRCh38
rs114612660 Chr.9: 95101047-95101047 on Build GRCh38
rs1043137022 Chr.9: 95101035-95101035 on Build GRCh38
rs865887946 Chr.9: 95100720-95100720 on Build GRCh38
rs887127195 Chr.9: 95100971-95100971 on Build GRCh38
rs1022149785 Chr.9: 95100768-95100768 on Build GRCh38
rs956557360 Chr.9: 95101087-95101087 on Build GRCh38
rs756756014 Chr.9: 95101039-95101039 on Build GRCh38
rs554500052 Chr.9: 95100699-95100699 on Build GRCh38
rs56059656 Chr.9: 95100724-95100724 on Build GRCh38
rs890408612 Chr.9: 95100816-95100816 on Build GRCh38
rs572664166 Chr.9: 95100998-95100998 on Build GRCh38
rs908769148 Chr.9: 95100942-95100942 on Build GRCh38
rs991797942 Chr.9: 95100870-95100870 on Build GRCh38
rs753765376 Chr.9: 95101023-95101023 on Build GRCh38
rs56250966 Chr.9: 95100981-95100984 on Build GRCh38
rs3780559 Chr.9: 95100723-95100723 on Build GRCh38
rs989098196 Chr.9: 95100756-95100756 on Build GRCh38
rs760485290 Chr.9: 95100787-95100787 on Build GRCh38
rs1057515699 Chr.9: 95100823-95100823 on Build GRCh38
rs957750212 Chr.9: 95101069-95101069 on Build GRCh38
rs150462386 Chr.9: 95101040-95101040 on Build GRCh38
rs944306522 Chr.9: 95100948-95100948 on Build GRCh38
rs763956310 Chr.9: 95100888-95100888 on Build GRCh38
rs931462868 Chr.9: 95100925-95100925 on Build GRCh38
rs556229977 Chr.9: 95100679-95100679 on Build GRCh38
rs917576719 Chr.9: 95100878-95100878 on Build GRCh38
rs1057515700 Chr.9: 95100882-95100882 on Build GRCh38
rs980515995 Chr.9: 95100903-95100903 on Build GRCh38
rs976747246 Chr.9: 95101034-95101034 on Build GRCh38
rs560893486 Chr.9: 95101046-95101046 on Build GRCh38
rs979684000 Chr.9: 95100819-95100819 on Build GRCh38
rs887364685 Chr.9: 95100700-95100700 on Build GRCh38
rs943180279 Chr.9: 95100822-95100822 on Build GRCh38
rs1030880241 Chr.9: 95100828-95100828 on Build GRCh38
rs1044866059 Chr.9: 95100648-95100648 on Build GRCh38

View Full Product Details