Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97309326-97309823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC180
Entrez Gene ID
100499483
Gene Name
coiled-coil domain containing 180
Gene Aliases
BDAG1, C9orf174, KIAA1529
UniGene
Hs.435629
Gene Chromosome Location
Chr.9: 97307628-97377295 on Build GRCh38
Transcripts
NM_020893.2 NR_036527.1 NR_036528.1 NR_036529.1

Gene Symbol
LOC100499484-C9ORF174
Entrez Gene ID
57653
Gene Name
LOC100499484-C9orf174 readthrough
Gene Aliases
-
UniGene
Hs.435629
Gene Chromosome Location
Chr.9: 97238426-97377295 on Build GRCh38
Transcripts
NM_020893.2 NR_036527.1 NR_036528.1 NR_036529.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7286638 c.655C>T p.R219W Chr.9: 97309450-97309450 on Build GRCh38
4542074 c.311G>A p.G104E Chr.9: 97309523-97309523 on Build GRCh38
6550280 c.311G>T p.G104V Chr.9: 97309523-97309523 on Build GRCh38
3432672 c.323C>A p.S108Y Chr.9: 97309535-97309535 on Build GRCh38
1464277 c.753G>T p.Q251H Chr.9: 97309548-97309548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750621394 Chr.9: 97309519-97309519 on Build GRCh38
rs780105852 Chr.9: 97309532-97309537 on Build GRCh38
rs750550333 Chr.9: 97309386-97309386 on Build GRCh38
rs775715262 Chr.9: 97309614-97309614 on Build GRCh38
rs533554585 Chr.9: 97309408-97309408 on Build GRCh38
rs768434399 Chr.9: 97309573-97309573 on Build GRCh38
rs3747506 Chr.9: 97309707-97309707 on Build GRCh38
rs779743714 Chr.9: 97309571-97309571 on Build GRCh38
rs1039073188 Chr.9: 97309714-97309714 on Build GRCh38
rs778356346 Chr.9: 97309566-97309566 on Build GRCh38
rs561709807 Chr.9: 97309550-97309550 on Build GRCh38
rs758452152 Chr.9: 97309389-97309389 on Build GRCh38
rs551390148 Chr.9: 97309655-97309655 on Build GRCh38
rs890484318 Chr.9: 97309494-97309494 on Build GRCh38
rs149588398 Chr.9: 97309517-97309517 on Build GRCh38
rs751767974 Chr.9: 97309462-97309462 on Build GRCh38
rs995494397 Chr.9: 97309557-97309557 on Build GRCh38
rs755390553 Chr.9: 97309414-97309414 on Build GRCh38
rs753768660 Chr.9: 97309451-97309451 on Build GRCh38
rs776566171 Chr.9: 97309718-97309718 on Build GRCh38
rs974352955 Chr.9: 97309701-97309701 on Build GRCh38
rs759190211 Chr.9: 97309437-97309437 on Build GRCh38
rs767557058 Chr.9: 97309602-97309602 on Build GRCh38
rs945175125 Chr.9: 97309663-97309663 on Build GRCh38
rs915107315 Chr.9: 97309662-97309662 on Build GRCh38
rs755389900 Chr.9: 97309568-97309569 on Build GRCh38
rs771794426 Chr.9: 97309440-97309440 on Build GRCh38
rs1049282859 Chr.9: 97309770-97309770 on Build GRCh38
rs778326464 Chr.9: 97309489-97309489 on Build GRCh38
rs754230941 Chr.9: 97309631-97309631 on Build GRCh38
rs535694548 Chr.9: 97309372-97309372 on Build GRCh38
rs766939975 Chr.9: 97309726-97309726 on Build GRCh38
rs1015914803 Chr.9: 97309680-97309680 on Build GRCh38
rs906286935 Chr.9: 97309801-97309801 on Build GRCh38
rs771745384 Chr.9: 97309516-97309516 on Build GRCh38
rs769820686 Chr.9: 97309587-97309587 on Build GRCh38
rs1015313360 Chr.9: 97309568-97309568 on Build GRCh38
rs1039015969 Chr.9: 97309495-97309495 on Build GRCh38
rs562561806 Chr.9: 97309565-97309565 on Build GRCh38
rs930936575 Chr.9: 97309762-97309762 on Build GRCh38
rs778182900 Chr.9: 97309431-97309431 on Build GRCh38
rs143592372 Chr.9: 97309561-97309561 on Build GRCh38
rs765712567 Chr.9: 97309637-97309637 on Build GRCh38
rs374421284 Chr.9: 97309509-97309509 on Build GRCh38
rs540169968 Chr.9: 97309529-97309529 on Build GRCh38
rs780405795 Chr.9: 97309392-97309392 on Build GRCh38
rs144353260 Chr.9: 97309555-97309555 on Build GRCh38
rs557395065 Chr.9: 97309444-97309444 on Build GRCh38
rs941642922 Chr.9: 97309493-97309493 on Build GRCh38
rs776631023 Chr.9: 97309530-97309530 on Build GRCh38
rs3747507 Chr.9: 97309446-97309446 on Build GRCh38
rs547247520 Chr.9: 97309784-97309784 on Build GRCh38
rs763115795 Chr.9: 97309459-97309459 on Build GRCh38
rs757922685 Chr.9: 97309647-97309647 on Build GRCh38
rs1005017898 Chr.9: 97309793-97309793 on Build GRCh38
rs374121300 Chr.9: 97309463-97309463 on Build GRCh38
rs10817403 Chr.9: 97309794-97309794 on Build GRCh38
rs1039807831 Chr.9: 97309792-97309793 on Build GRCh38
rs930171330 Chr.9: 97309754-97309754 on Build GRCh38
rs751232499 Chr.9: 97309541-97309541 on Build GRCh38
rs781694688 Chr.9: 97309416-97309416 on Build GRCh38
rs748547217 Chr.9: 97309420-97309420 on Build GRCh38
rs748809506 Chr.9: 97309434-97309434 on Build GRCh38
rs770362825 Chr.9: 97309423-97309423 on Build GRCh38
rs756387081 Chr.9: 97309529-97309530 on Build GRCh38
rs779505806 Chr.9: 97309569-97309570 on Build GRCh38
rs367967039 Chr.9: 97309474-97309474 on Build GRCh38
rs770486284 Chr.9: 97309435-97309435 on Build GRCh38
rs765185130 Chr.9: 97309380-97309380 on Build GRCh38
rs760185118 Chr.9: 97309645-97309645 on Build GRCh38
rs756492243 Chr.9: 97309560-97309560 on Build GRCh38
rs770361437 Chr.9: 97309498-97309498 on Build GRCh38
rs757661591 Chr.9: 97309632-97309632 on Build GRCh38
rs761850815 Chr.9: 97309531-97309531 on Build GRCh38
rs760819889 Chr.9: 97309616-97309616 on Build GRCh38
rs764469370 Chr.9: 97309625-97309625 on Build GRCh38
rs773328626 Chr.9: 97309593-97309593 on Build GRCh38
rs774886818 Chr.9: 97309436-97309436 on Build GRCh38
rs748643123 Chr.9: 97309634-97309635 on Build GRCh38
rs761762906 Chr.9: 97309755-97309755 on Build GRCh38
rs1042484153 Chr.9: 97309475-97309475 on Build GRCh38
rs144942620 Chr.9: 97309569-97309569 on Build GRCh38
rs977896155 Chr.9: 97309667-97309667 on Build GRCh38
rs891186482 Chr.9: 97309518-97309518 on Build GRCh38
rs759671250 Chr.9: 97309599-97309599 on Build GRCh38
rs557777905 Chr.9: 97309492-97309492 on Build GRCh38
rs760496954 Chr.9: 97309528-97309528 on Build GRCh38
rs147529206 Chr.9: 97309572-97309572 on Build GRCh38
rs766637317 Chr.9: 97309460-97309460 on Build GRCh38
rs528775824 Chr.9: 97309739-97309739 on Build GRCh38
rs911993626 Chr.9: 97309683-97309683 on Build GRCh38
rs987350893 Chr.9: 97309758-97309758 on Build GRCh38
rs751888188 Chr.9: 97309396-97309396 on Build GRCh38
rs545746004 Chr.9: 97309450-97309450 on Build GRCh38
rs370298824 Chr.9: 97309507-97309507 on Build GRCh38
rs367601533 Chr.9: 97309513-97309513 on Build GRCh38
rs745352802 Chr.9: 97309567-97309567 on Build GRCh38
rs775083413 Chr.9: 97309579-97309579 on Build GRCh38
rs766512576 Chr.9: 97309540-97309540 on Build GRCh38
rs749397882 Chr.9: 97309538-97309539 on Build GRCh38

View Full Product Details