Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98120843-98121345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO2A
Entrez Gene ID
7464
Gene Name
coronin 2A
Gene Aliases
CLIPINB, IR10, WDR2
UniGene
Hs.113094
Gene Chromosome Location
Chr.9: 98120975-98192674 on Build GRCh38
Transcripts
NM_003389.3 NM_052820.3 XM_011518986.2

SNP Detail

SNP ID Chromosome Location
rs989797446 Chr.9: 98121144-98121144 on Build GRCh38
rs930942914 Chr.9: 98121307-98121307 on Build GRCh38
rs974402996 Chr.9: 98121296-98121296 on Build GRCh38
rs745806635 Chr.9: 98121230-98121230 on Build GRCh38
rs945087437 Chr.9: 98121213-98121222 on Build GRCh38
rs546511963 Chr.9: 98121318-98121318 on Build GRCh38
rs182264823 Chr.9: 98121103-98121103 on Build GRCh38
rs530511081 Chr.9: 98120876-98120876 on Build GRCh38
rs771977617 Chr.9: 98121311-98121311 on Build GRCh38
rs906939893 Chr.9: 98121209-98121209 on Build GRCh38
rs1026265040 Chr.9: 98120872-98120872 on Build GRCh38
rs1044197719 Chr.9: 98121207-98121207 on Build GRCh38
rs954760206 Chr.9: 98121065-98121067 on Build GRCh38
rs921587888 Chr.9: 98121305-98121305 on Build GRCh38
rs959307303 Chr.9: 98121136-98121136 on Build GRCh38
rs550270099 Chr.9: 98120930-98120930 on Build GRCh38
rs1016069628 Chr.9: 98121292-98121292 on Build GRCh38
rs575094892 Chr.9: 98120886-98120887 on Build GRCh38
rs977875810 Chr.9: 98121224-98121224 on Build GRCh38
rs1049256753 Chr.9: 98120904-98120904 on Build GRCh38
rs570285552 Chr.9: 98120993-98120993 on Build GRCh38
rs1030352116 Chr.9: 98121029-98121029 on Build GRCh38
rs989662005 Chr.9: 98121111-98121111 on Build GRCh38
rs537467009 Chr.9: 98121269-98121269 on Build GRCh38
rs978488049 Chr.9: 98121069-98121069 on Build GRCh38
rs886365467 Chr.9: 98121252-98121252 on Build GRCh38
rs1034333437 Chr.9: 98121125-98121125 on Build GRCh38
rs1063628 Chr.9: 98121238-98121238 on Build GRCh38
rs886353528 Chr.9: 98120881-98120884 on Build GRCh38
rs1005005337 Chr.9: 98120941-98120941 on Build GRCh38
rs1001514425 Chr.9: 98121101-98121101 on Build GRCh38
rs1016015880 Chr.9: 98120926-98120926 on Build GRCh38
rs907217432 Chr.9: 98121094-98121094 on Build GRCh38
rs911612870 Chr.9: 98121231-98121231 on Build GRCh38
rs924253940 Chr.9: 98121072-98121072 on Build GRCh38
rs778811547 Chr.9: 98121205-98121205 on Build GRCh38
rs966907537 Chr.9: 98121216-98121216 on Build GRCh38

View Full Product Details