Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98122501-98122998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO2A
Entrez Gene ID
7464
Gene Name
coronin 2A
Gene Aliases
CLIPINB, IR10, WDR2
UniGene
Hs.113094
Gene Chromosome Location
Chr.9: 98120975-98192674 on Build GRCh38
Transcripts
NM_003389.3 NM_052820.3 XM_011518986.2

SNP Detail

SNP ID Chromosome Location
rs963109347 Chr.9: 98122568-98122568 on Build GRCh38
rs115062197 Chr.9: 98122569-98122569 on Build GRCh38
rs887365063 Chr.9: 98122543-98122543 on Build GRCh38
rs774648585 Chr.9: 98122893-98122893 on Build GRCh38
rs145467027 Chr.9: 98122746-98122746 on Build GRCh38
rs192924976 Chr.9: 98122558-98122558 on Build GRCh38
rs924688677 Chr.9: 98122877-98122877 on Build GRCh38
rs996962865 Chr.9: 98122896-98122896 on Build GRCh38
rs1891323 Chr.9: 98122656-98122656 on Build GRCh38
rs770687005 Chr.9: 98122565-98122565 on Build GRCh38
rs560249230 Chr.9: 98122740-98122740 on Build GRCh38
rs905267690 Chr.9: 98122823-98122823 on Build GRCh38
rs760635615 Chr.9: 98122554-98122554 on Build GRCh38
rs563788677 Chr.9: 98122589-98122589 on Build GRCh38
rs942164 Chr.9: 98122752-98122752 on Build GRCh38
rs184740384 Chr.9: 98122816-98122816 on Build GRCh38
rs947491826 Chr.9: 98122566-98122566 on Build GRCh38
rs976752112 Chr.9: 98122890-98122890 on Build GRCh38
rs1050325397 Chr.9: 98122950-98122950 on Build GRCh38
rs931889056 Chr.9: 98122651-98122651 on Build GRCh38
rs765128828 Chr.9: 98122655-98122655 on Build GRCh38
rs1028738969 Chr.9: 98122961-98122961 on Build GRCh38
rs953120449 Chr.9: 98122975-98122975 on Build GRCh38
rs1017123482 Chr.9: 98122889-98122889 on Build GRCh38
rs991680115 Chr.9: 98122538-98122538 on Build GRCh38
rs958880063 Chr.9: 98122536-98122536 on Build GRCh38
rs147680752 Chr.9: 98122769-98122769 on Build GRCh38
rs931941455 Chr.9: 98122915-98122915 on Build GRCh38
rs866650275 Chr.9: 98122679-98122679 on Build GRCh38
rs988395086 Chr.9: 98122744-98122744 on Build GRCh38
rs701378 Chr.9: 98122860-98122860 on Build GRCh38
rs771079098 Chr.9: 98122888-98122888 on Build GRCh38
rs756921021 Chr.9: 98122654-98122654 on Build GRCh38
rs777776973 Chr.9: 98122793-98122793 on Build GRCh38
rs759335832 Chr.9: 98122688-98122688 on Build GRCh38
rs879250803 Chr.9: 98122588-98122588 on Build GRCh38
rs780017364 Chr.9: 98122723-98122723 on Build GRCh38
rs897509095 Chr.9: 98122711-98122711 on Build GRCh38
rs190407678 Chr.9: 98122549-98122549 on Build GRCh38
rs142428490 Chr.9: 98122807-98122807 on Build GRCh38
rs968872933 Chr.9: 98122848-98122848 on Build GRCh38
rs939387836 Chr.9: 98122841-98122841 on Build GRCh38
rs1056429342 Chr.9: 98122851-98122851 on Build GRCh38
rs12238311 Chr.9: 98122614-98122614 on Build GRCh38
rs571709369 Chr.9: 98122820-98122820 on Build GRCh38
rs574302185 Chr.9: 98122591-98122591 on Build GRCh38
rs903774194 Chr.9: 98122883-98122883 on Build GRCh38
rs547576993 Chr.9: 98122952-98122952 on Build GRCh38

View Full Product Details