Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 99217440-99217950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG2
Entrez Gene ID
85365
Gene Name
ALG2, alpha-1,3/1,6-mannosyltransferase
Gene Aliases
CDGIi, CMS14, CMSTA3, NET38, hALPG2
UniGene
Hs.40919
Gene Chromosome Location
Chr.9: 99216425-99221964 on Build GRCh38
Transcripts
NM_033087.3 NR_024532.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1103199 c.1240C>T p.L414L Chr.9: 99217945-99217945 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755097173 Chr.9: 99217864-99217864 on Build GRCh38
rs987407355 Chr.9: 99217546-99217546 on Build GRCh38
rs113080479 Chr.9: 99217681-99217681 on Build GRCh38
rs985665216 Chr.9: 99217575-99217575 on Build GRCh38
rs777096197 Chr.9: 99217787-99217788 on Build GRCh38
rs566212682 Chr.9: 99217788-99217788 on Build GRCh38
rs551384792 Chr.9: 99217783-99217783 on Build GRCh38
rs753611171 Chr.9: 99217834-99217834 on Build GRCh38
rs766834738 Chr.9: 99217591-99217592 on Build GRCh38
rs775344567 Chr.9: 99217813-99217813 on Build GRCh38
rs144947827 Chr.9: 99217641-99217641 on Build GRCh38
rs778009043 Chr.9: 99217916-99217916 on Build GRCh38
rs929989190 Chr.9: 99217667-99217667 on Build GRCh38
rs545165833 Chr.9: 99217738-99217738 on Build GRCh38
rs547977189 Chr.9: 99217657-99217657 on Build GRCh38
rs1043505507 Chr.9: 99217646-99217646 on Build GRCh38
rs533399840 Chr.9: 99217516-99217516 on Build GRCh38
rs868582111 Chr.9: 99217750-99217750 on Build GRCh38
rs988800917 Chr.9: 99217563-99217563 on Build GRCh38
rs941587788 Chr.9: 99217633-99217633 on Build GRCh38
rs1030115315 Chr.9: 99217559-99217559 on Build GRCh38
rs764898893 Chr.9: 99217920-99217928 on Build GRCh38
rs781431140 Chr.9: 99217687-99217687 on Build GRCh38
rs780011889 Chr.9: 99217548-99217548 on Build GRCh38
rs1005121928 Chr.9: 99217722-99217722 on Build GRCh38
rs769497390 Chr.9: 99217724-99217724 on Build GRCh38
rs955516968 Chr.9: 99217570-99217570 on Build GRCh38
rs781235728 Chr.9: 99217870-99217870 on Build GRCh38
rs1056458604 Chr.9: 99217728-99217728 on Build GRCh38
rs567866273 Chr.9: 99217702-99217702 on Build GRCh38
rs1031361341 Chr.9: 99217917-99217917 on Build GRCh38
rs913217206 Chr.9: 99217565-99217565 on Build GRCh38
rs994598506 Chr.9: 99217763-99217763 on Build GRCh38
rs370330270 Chr.9: 99217781-99217781 on Build GRCh38
rs953151416 Chr.9: 99217902-99217902 on Build GRCh38
rs1004585539 Chr.9: 99217495-99217495 on Build GRCh38
rs773752365 Chr.9: 99217770-99217770 on Build GRCh38
rs375055192 Chr.9: 99217869-99217869 on Build GRCh38
rs760403292 Chr.9: 99217819-99217819 on Build GRCh38
rs1001200979 Chr.9: 99217470-99217470 on Build GRCh38
rs13283030 Chr.9: 99217692-99217692 on Build GRCh38
rs773136928 Chr.9: 99217746-99217746 on Build GRCh38
rs753789244 Chr.9: 99217509-99217509 on Build GRCh38
rs748006857 Chr.9: 99217882-99217882 on Build GRCh38
rs953298422 Chr.9: 99217535-99217535 on Build GRCh38
rs373058210 Chr.9: 99217863-99217865 on Build GRCh38
rs186134722 Chr.9: 99217858-99217858 on Build GRCh38
rs552176290 Chr.9: 99217636-99217636 on Build GRCh38
rs763802412 Chr.9: 99217825-99217825 on Build GRCh38
rs757166552 Chr.9: 99217845-99217845 on Build GRCh38
rs770647305 Chr.9: 99217675-99217675 on Build GRCh38
rs534640878 Chr.9: 99217656-99217656 on Build GRCh38
rs577290860 Chr.9: 99217863-99217863 on Build GRCh38
rs773617864 Chr.9: 99217739-99217739 on Build GRCh38
rs142781699 Chr.9: 99217512-99217512 on Build GRCh38
rs767116632 Chr.9: 99217800-99217800 on Build GRCh38
rs763370114 Chr.9: 99217754-99217754 on Build GRCh38
rs748435948 Chr.9: 99217892-99217892 on Build GRCh38
rs960318802 Chr.9: 99217491-99217491 on Build GRCh38
rs759160900 Chr.9: 99217784-99217784 on Build GRCh38
rs771409082 Chr.9: 99217761-99217761 on Build GRCh38
rs756258238 Chr.9: 99217900-99217900 on Build GRCh38
rs759852086 Chr.9: 99217860-99217860 on Build GRCh38
rs1057293165 Chr.9: 99217707-99217707 on Build GRCh38
rs1036314771 Chr.9: 99217477-99217477 on Build GRCh38
rs963149772 Chr.9: 99217508-99217508 on Build GRCh38
rs748346515 Chr.9: 99217717-99217717 on Build GRCh38
rs921803279 Chr.9: 99217525-99217525 on Build GRCh38
rs777702474 Chr.9: 99217505-99217505 on Build GRCh38
rs911395619 Chr.9: 99217625-99217625 on Build GRCh38

View Full Product Details