Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 100099099-100099592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INVS
Entrez Gene ID
27130
Gene Name
inversin
Gene Aliases
INV, NPH2, NPHP2
UniGene
Hs.558477
Gene Chromosome Location
Chr.9: 100099185-100302175 on Build GRCh38
Transcripts
NM_001318381.1 NM_001318382.1 NM_014425.4 NR_134606.1

SNP Detail

SNP ID Chromosome Location
rs1033714301 Chr.9: 100099478-100099478 on Build GRCh38
rs1022527144 Chr.9: 100099223-100099223 on Build GRCh38
rs912320934 Chr.9: 100099352-100099352 on Build GRCh38
rs533889420 Chr.9: 100099312-100099312 on Build GRCh38
rs961216008 Chr.9: 100099289-100099289 on Build GRCh38
rs867234496 Chr.9: 100099529-100099529 on Build GRCh38
rs536258005 Chr.9: 100099411-100099411 on Build GRCh38
rs1012400302 Chr.9: 100099558-100099558 on Build GRCh38
rs537621596 Chr.9: 100099249-100099249 on Build GRCh38
rs770885080 Chr.9: 100099429-100099429 on Build GRCh38
rs939184392 Chr.9: 100099132-100099132 on Build GRCh38
rs6479002 Chr.9: 100099166-100099166 on Build GRCh38
rs1045839589 Chr.9: 100099213-100099213 on Build GRCh38
rs370987236 Chr.9: 100099453-100099453 on Build GRCh38
rs1054107425 Chr.9: 100099387-100099387 on Build GRCh38
rs973907990 Chr.9: 100099533-100099533 on Build GRCh38
rs980253275 Chr.9: 100099325-100099325 on Build GRCh38
rs902924409 Chr.9: 100099220-100099220 on Build GRCh38
rs997100255 Chr.9: 100099271-100099271 on Build GRCh38
rs932305987 Chr.9: 100099167-100099167 on Build GRCh38
rs974234893 Chr.9: 100099124-100099124 on Build GRCh38
rs1038286142 Chr.9: 100099163-100099163 on Build GRCh38
rs968070744 Chr.9: 100099227-100099227 on Build GRCh38
rs992361239 Chr.9: 100099144-100099144 on Build GRCh38
rs571502726 Chr.9: 100099288-100099288 on Build GRCh38
rs1008595968 Chr.9: 100099480-100099480 on Build GRCh38
rs991048057 Chr.9: 100099160-100099160 on Build GRCh38
rs540077792 Chr.9: 100099146-100099146 on Build GRCh38
rs959750175 Chr.9: 100099553-100099553 on Build GRCh38
rs556269258 Chr.9: 100099412-100099412 on Build GRCh38
rs756648020 Chr.9: 100099539-100099539 on Build GRCh38
rs142849038 Chr.9: 100099228-100099228 on Build GRCh38
rs879293808 Chr.9: 100099177-100099177 on Build GRCh38
rs1008210608 Chr.9: 100099305-100099305 on Build GRCh38
rs1050048469 Chr.9: 100099424-100099424 on Build GRCh38
rs960746121 Chr.9: 100099251-100099251 on Build GRCh38
rs755935488 Chr.9: 100099274-100099274 on Build GRCh38
rs1025604788 Chr.9: 100099564-100099564 on Build GRCh38
rs62577237 Chr.9: 100099286-100099286 on Build GRCh38
rs971412130 Chr.9: 100099354-100099354 on Build GRCh38
rs920125231 Chr.9: 100099129-100099129 on Build GRCh38
rs897120500 Chr.9: 100099498-100099498 on Build GRCh38
rs868400727 Chr.9: 100099390-100099390 on Build GRCh38
rs1044015510 Chr.9: 100099379-100099379 on Build GRCh38
rs992247628 Chr.9: 100099283-100099283 on Build GRCh38
rs555449965 Chr.9: 100099517-100099517 on Build GRCh38
rs904065758 Chr.9: 100099410-100099410 on Build GRCh38
rs1009071035 Chr.9: 100099200-100099200 on Build GRCh38
rs545569313 Chr.9: 100099546-100099546 on Build GRCh38
rs1015663835 Chr.9: 100099518-100099518 on Build GRCh38
rs1010273757 Chr.9: 100099222-100099222 on Build GRCh38
rs752368953 Chr.9: 100099212-100099212 on Build GRCh38
rs934327967 Chr.9: 100099381-100099381 on Build GRCh38
rs1009725507 Chr.9: 100099365-100099365 on Build GRCh38
rs770460255 Chr.9: 100099406-100099406 on Build GRCh38
rs1015640482 Chr.9: 100099308-100099308 on Build GRCh38
rs961514748 Chr.9: 100099526-100099526 on Build GRCh38
rs112431315 Chr.9: 100099221-100099221 on Build GRCh38
rs567598200 Chr.9: 100099154-100099154 on Build GRCh38
rs908464167 Chr.9: 100099159-100099159 on Build GRCh38
rs892651935 Chr.9: 100099358-100099358 on Build GRCh38
rs890853749 Chr.9: 100099193-100099193 on Build GRCh38
rs932486850 Chr.9: 100099408-100099408 on Build GRCh38
rs1054211647 Chr.9: 100099239-100099239 on Build GRCh38
rs924316671 Chr.9: 100099373-100099373 on Build GRCh38
rs918164220 Chr.9: 100099353-100099353 on Build GRCh38
rs891064227 Chr.9: 100099300-100099300 on Build GRCh38
rs1036957513 Chr.9: 100099489-100099489 on Build GRCh38
rs7024375 Chr.9: 100099331-100099331 on Build GRCh38
rs1002211378 Chr.9: 100099445-100099445 on Build GRCh38
rs886063263 Chr.9: 100099263-100099263 on Build GRCh38
rs961446430 Chr.9: 100099309-100099309 on Build GRCh38
rs1049480127 Chr.9: 100099183-100099183 on Build GRCh38
rs981073893 Chr.9: 100099360-100099360 on Build GRCh38
rs959544769 Chr.9: 100099346-100099346 on Build GRCh38
rs573639725 Chr.9: 100099347-100099347 on Build GRCh38

View Full Product Details