Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 100225866-100226330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INVS
Entrez Gene ID
27130
Gene Name
inversin
Gene Aliases
INV, NPH2, NPHP2
UniGene
Hs.558477
Gene Chromosome Location
Chr.9: 100099185-100302175 on Build GRCh38
Transcripts
NM_001318381.1 NM_001318382.1 NM_014425.4 NR_134606.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3902558 c.283C>T p.R95C Chr.9: 100226071-100226071 on Build GRCh38
1458916 c.284G>A p.R95H Chr.9: 100226072-100226072 on Build GRCh38
3902559 c.329A>G p.K110R Chr.9: 100226117-100226117 on Build GRCh38
3902560 c.41A>G p.K14R Chr.9: 100226117-100226117 on Build GRCh38
3902561 c.372C>T p.H124H Chr.9: 100226160-100226160 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056975326 Chr.9: 100226187-100226187 on Build GRCh38
rs755572098 Chr.9: 100226028-100226029 on Build GRCh38
rs201366153 Chr.9: 100226048-100226048 on Build GRCh38
rs560851395 Chr.9: 100226152-100226152 on Build GRCh38
rs755654469 Chr.9: 100226248-100226248 on Build GRCh38
rs375188577 Chr.9: 100226074-100226074 on Build GRCh38
rs887804052 Chr.9: 100225991-100225991 on Build GRCh38
rs761539455 Chr.9: 100226025-100226025 on Build GRCh38
rs975952124 Chr.9: 100226294-100226294 on Build GRCh38
rs776737973 Chr.9: 100226108-100226108 on Build GRCh38
rs866469135 Chr.9: 100225962-100225962 on Build GRCh38
rs770506909 Chr.9: 100226271-100226271 on Build GRCh38
rs750863293 Chr.9: 100226095-100226095 on Build GRCh38
rs768984655 Chr.9: 100226139-100226139 on Build GRCh38
rs781529497 Chr.9: 100226239-100226239 on Build GRCh38
rs1012910834 Chr.9: 100226011-100226011 on Build GRCh38
rs757623556 Chr.9: 100226089-100226089 on Build GRCh38
rs375209595 Chr.9: 100226198-100226198 on Build GRCh38
rs528036309 Chr.9: 100226307-100226307 on Build GRCh38
rs996740227 Chr.9: 100226247-100226247 on Build GRCh38
rs747572974 Chr.9: 100226125-100226125 on Build GRCh38
rs745703727 Chr.9: 100226221-100226221 on Build GRCh38
rs759346863 Chr.9: 100226056-100226056 on Build GRCh38
rs907280207 Chr.9: 100226021-100226021 on Build GRCh38
rs548854031 Chr.9: 100226185-100226185 on Build GRCh38
rs61147858 Chr.9: 100226047-100226048 on Build GRCh38
rs778745264 Chr.9: 100226127-100226128 on Build GRCh38
rs767356831 Chr.9: 100226174-100226174 on Build GRCh38
rs751547848 Chr.9: 100226052-100226052 on Build GRCh38
rs747606826 Chr.9: 100226154-100226154 on Build GRCh38
rs111681340 Chr.9: 100226297-100226297 on Build GRCh38
rs766345404 Chr.9: 100226050-100226050 on Build GRCh38
rs779261194 Chr.9: 100226092-100226092 on Build GRCh38
rs373485994 Chr.9: 100226147-100226147 on Build GRCh38
rs757534991 Chr.9: 100226156-100226156 on Build GRCh38
rs369271547 Chr.9: 100226205-100226205 on Build GRCh38
rs747735892 Chr.9: 100226245-100226245 on Build GRCh38
rs372088206 Chr.9: 100226072-100226072 on Build GRCh38
rs748955353 Chr.9: 100226270-100226270 on Build GRCh38
rs765593554 Chr.9: 100226196-100226196 on Build GRCh38
rs372662270 Chr.9: 100226263-100226263 on Build GRCh38
rs955020033 Chr.9: 100225913-100225913 on Build GRCh38
rs935519829 Chr.9: 100226003-100226003 on Build GRCh38
rs768677542 Chr.9: 100226085-100226088 on Build GRCh38
rs145532725 Chr.9: 100225908-100225909 on Build GRCh38
rs919187465 Chr.9: 100225930-100225930 on Build GRCh38
rs755575787 Chr.9: 100226232-100226232 on Build GRCh38
rs60607451 Chr.9: 100225909-100225910 on Build GRCh38
rs149315279 Chr.9: 100226155-100226155 on Build GRCh38
rs779668166 Chr.9: 100226040-100226044 on Build GRCh38
rs759526374 Chr.9: 100226168-100226168 on Build GRCh38
rs770737798 Chr.9: 100226163-100226163 on Build GRCh38
rs1012219252 Chr.9: 100226135-100226135 on Build GRCh38
rs758802357 Chr.9: 100226096-100226096 on Build GRCh38
rs776825894 Chr.9: 100226181-100226181 on Build GRCh38
rs769369619 Chr.9: 100226029-100226029 on Build GRCh38
rs1049107496 Chr.9: 100225977-100225977 on Build GRCh38
rs138668044 Chr.9: 100226145-100226145 on Build GRCh38
rs56840267 Chr.9: 100225952-100225952 on Build GRCh38
rs775953086 Chr.9: 100226012-100226012 on Build GRCh38
rs1032223225 Chr.9: 100226121-100226121 on Build GRCh38
rs751473961 Chr.9: 100226046-100226047 on Build GRCh38
rs867086899 Chr.9: 100226117-100226117 on Build GRCh38
rs1021818693 Chr.9: 100226137-100226137 on Build GRCh38
rs766778206 Chr.9: 100226225-100226225 on Build GRCh38
rs566435222 Chr.9: 100226040-100226040 on Build GRCh38
rs369509413 Chr.9: 100226100-100226100 on Build GRCh38
rs778677145 Chr.9: 100226215-100226215 on Build GRCh38

View Full Product Details