Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 110138440-110138937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP2
Entrez Gene ID
11217
Gene Name
A-kinase anchoring protein 2
Gene Aliases
AKAP-2, AKAPKL, MISP2, PRKA2
UniGene
Hs.591908
Gene Chromosome Location
Chr.9: 110048598-110172512 on Build GRCh38
Transcripts
NM_001004065.4 NM_001136562.2 NM_001198656.1 NM_007203.4 NM_147150.2

Gene Symbol
PALM2-AKAP2
Entrez Gene ID
445815
Gene Name
PALM2-AKAP2 readthrough
Gene Aliases
AKAP2
UniGene
Hs.591908
Gene Chromosome Location
Chr.9: 109780297-110172512 on Build GRCh38
Transcripts
NM_001004065.4 NM_001136562.2 NM_001198656.1 NM_007203.4 NM_147150.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1701515 c.2539C>T p.P847S Chr.9: 110138509-110138509 on Build GRCh38
7422948 c.2555A>T p.Y852F Chr.9: 110138525-110138525 on Build GRCh38
6369803 c.2503A>G p.T835A Chr.9: 110138473-110138473 on Build GRCh38
3982678 c.2927G>A p.S976N Chr.9: 110138471-110138471 on Build GRCh38
3982679 c.2501G>A p.S834N Chr.9: 110138471-110138471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191043666 Chr.9: 110138863-110138863 on Build GRCh38
rs180763405 Chr.9: 110138888-110138888 on Build GRCh38
rs770005256 Chr.9: 110138539-110138539 on Build GRCh38
rs897800488 Chr.9: 110138738-110138738 on Build GRCh38
rs960967479 Chr.9: 110138772-110138772 on Build GRCh38
rs764217285 Chr.9: 110138463-110138463 on Build GRCh38
rs539552025 Chr.9: 110138831-110138831 on Build GRCh38
rs750832165 Chr.9: 110138570-110138570 on Build GRCh38
rs907989188 Chr.9: 110138898-110138898 on Build GRCh38
rs746629234 Chr.9: 110138502-110138502 on Build GRCh38
rs531486809 Chr.9: 110138573-110138573 on Build GRCh38
rs547046302 Chr.9: 110138797-110138797 on Build GRCh38
rs377019694 Chr.9: 110138497-110138497 on Build GRCh38
rs1010651082 Chr.9: 110138631-110138631 on Build GRCh38
rs768964636 Chr.9: 110138546-110138546 on Build GRCh38
rs3739456 Chr.9: 110138505-110138505 on Build GRCh38
rs769839334 Chr.9: 110138513-110138513 on Build GRCh38
rs1013036521 Chr.9: 110138893-110138893 on Build GRCh38
rs541180913 Chr.9: 110138552-110138552 on Build GRCh38
rs557443818 Chr.9: 110138842-110138842 on Build GRCh38
rs376606574 Chr.9: 110138475-110138475 on Build GRCh38
rs761783014 Chr.9: 110138555-110138555 on Build GRCh38
rs755252885 Chr.9: 110138583-110138585 on Build GRCh38
rs775877745 Chr.9: 110138534-110138534 on Build GRCh38
rs1027949247 Chr.9: 110138579-110138579 on Build GRCh38
rs202061551 Chr.9: 110138480-110138480 on Build GRCh38
rs140643568 Chr.9: 110138520-110138520 on Build GRCh38
rs910377543 Chr.9: 110138657-110138657 on Build GRCh38
rs187017185 Chr.9: 110138801-110138801 on Build GRCh38
rs754236658 Chr.9: 110138521-110138523 on Build GRCh38
rs777511698 Chr.9: 110138519-110138519 on Build GRCh38
rs749670412 Chr.9: 110138614-110138614 on Build GRCh38
rs373087862 Chr.9: 110138474-110138474 on Build GRCh38
rs551354793 Chr.9: 110138862-110138862 on Build GRCh38
rs747582238 Chr.9: 110138508-110138508 on Build GRCh38
rs966848096 Chr.9: 110138700-110138700 on Build GRCh38
rs371019109 Chr.9: 110138884-110138884 on Build GRCh38
rs765789312 Chr.9: 110138567-110138567 on Build GRCh38
rs556904046 Chr.9: 110138913-110138913 on Build GRCh38
rs761260209 Chr.9: 110138465-110138466 on Build GRCh38
rs1022091473 Chr.9: 110138688-110138688 on Build GRCh38
rs774760624 Chr.9: 110138821-110138821 on Build GRCh38
rs984838625 Chr.9: 110138880-110138880 on Build GRCh38
rs978282203 Chr.9: 110138721-110138721 on Build GRCh38
rs750860393 Chr.9: 110138496-110138496 on Build GRCh38
rs567882419 Chr.9: 110138705-110138705 on Build GRCh38
rs3739455 Chr.9: 110138727-110138727 on Build GRCh38
rs765655099 Chr.9: 110138494-110138494 on Build GRCh38
rs761023069 Chr.9: 110138544-110138544 on Build GRCh38
rs780805317 Chr.9: 110138507-110138507 on Build GRCh38
rs776835403 Chr.9: 110138551-110138551 on Build GRCh38
rs993481966 Chr.9: 110138764-110138764 on Build GRCh38
rs749098354 Chr.9: 110138522-110138522 on Build GRCh38
rs74612901 Chr.9: 110138601-110138601 on Build GRCh38
rs770808786 Chr.9: 110138524-110138524 on Build GRCh38
rs767033135 Chr.9: 110138496-110138496 on Build GRCh38

View Full Product Details