Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 69076787-69077290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXN
Entrez Gene ID
2395
Gene Name
frataxin
Gene Aliases
CyaY, FA, FARR, FRDA, X25
UniGene
Hs.20685
Gene Chromosome Location
Chr.9: 69035563-69100178 on Build GRCh38
Transcripts
NM_000144.4 NM_001161706.1 NM_181425.2

SNP Detail

SNP ID Chromosome Location
rs529810877 Chr.9: 69077270-69077270 on Build GRCh38
rs181918026 Chr.9: 69077072-69077072 on Build GRCh38
rs530843434 Chr.9: 69076949-69076949 on Build GRCh38
rs1021885635 Chr.9: 69077253-69077253 on Build GRCh38
rs977183852 Chr.9: 69076926-69076926 on Build GRCh38
rs188899351 Chr.9: 69076886-69076886 on Build GRCh38
rs973802192 Chr.9: 69076915-69076915 on Build GRCh38
rs763967317 Chr.9: 69076889-69076889 on Build GRCh38
rs910502651 Chr.9: 69076973-69076973 on Build GRCh38
rs981123559 Chr.9: 69076893-69076893 on Build GRCh38
rs941939544 Chr.9: 69076974-69076974 on Build GRCh38
rs900495424 Chr.9: 69077116-69077116 on Build GRCh38
rs933001522 Chr.9: 69076931-69076931 on Build GRCh38
rs796604602 Chr.9: 69077076-69077076 on Build GRCh38
rs527521799 Chr.9: 69076832-69076832 on Build GRCh38
rs892993279 Chr.9: 69077178-69077178 on Build GRCh38
rs922990305 Chr.9: 69076928-69076928 on Build GRCh38
rs185073365 Chr.9: 69076839-69076839 on Build GRCh38
rs576320282 Chr.9: 69077093-69077093 on Build GRCh38
rs1011563208 Chr.9: 69077240-69077240 on Build GRCh38
rs536322979 Chr.9: 69077047-69077047 on Build GRCh38
rs764714873 Chr.9: 69076977-69076977 on Build GRCh38
rs1030092126 Chr.9: 69077049-69077049 on Build GRCh38
rs552453290 Chr.9: 69076978-69076978 on Build GRCh38
rs929149640 Chr.9: 69077163-69077163 on Build GRCh38
rs1032835863 Chr.9: 69076820-69076820 on Build GRCh38
rs1011823668 Chr.9: 69077026-69077026 on Build GRCh38
rs1025628229 Chr.9: 69076847-69076847 on Build GRCh38
rs541223608 Chr.9: 69076883-69076883 on Build GRCh38
rs938749018 Chr.9: 69077218-69077218 on Build GRCh38
rs1050478668 Chr.9: 69076963-69076963 on Build GRCh38
rs11145061 Chr.9: 69077001-69077001 on Build GRCh38
rs141798960 Chr.9: 69077000-69077000 on Build GRCh38
rs957208909 Chr.9: 69076845-69076845 on Build GRCh38
rs571014160 Chr.9: 69076946-69076946 on Build GRCh38
rs529911504 Chr.9: 69077074-69077074 on Build GRCh38
rs746694402 Chr.9: 69077073-69077073 on Build GRCh38
rs919150852 Chr.9: 69077102-69077102 on Build GRCh38
rs993262489 Chr.9: 69076996-69076996 on Build GRCh38
rs760340650 Chr.9: 69076887-69076887 on Build GRCh38
rs541263315 Chr.9: 69077198-69077198 on Build GRCh38
rs532384045 Chr.9: 69077260-69077260 on Build GRCh38
rs967558808 Chr.9: 69077039-69077039 on Build GRCh38
rs11145063 Chr.9: 69077221-69077221 on Build GRCh38
rs757256914 Chr.9: 69076954-69076954 on Build GRCh38
rs34235465 Chr.9: 69076914-69076915 on Build GRCh38
rs990375399 Chr.9: 69076905-69076905 on Build GRCh38
rs1055829203 Chr.9: 69077219-69077219 on Build GRCh38
rs894752553 Chr.9: 69077022-69077022 on Build GRCh38
rs1020514577 Chr.9: 69077210-69077210 on Build GRCh38
rs973306828 Chr.9: 69077101-69077101 on Build GRCh38
rs941986270 Chr.9: 69077100-69077100 on Build GRCh38
rs543710417 Chr.9: 69077103-69077103 on Build GRCh38
rs1002910654 Chr.9: 69077004-69077004 on Build GRCh38
rs12554971 Chr.9: 69077019-69077019 on Build GRCh38
rs569515511 Chr.9: 69076922-69076922 on Build GRCh38
rs566191092 Chr.9: 69077038-69077038 on Build GRCh38
rs577112663 Chr.9: 69077192-69077192 on Build GRCh38
rs34356733 Chr.9: 69076982-69076983 on Build GRCh38
rs967150624 Chr.9: 69076910-69076910 on Build GRCh38
rs910191022 Chr.9: 69077075-69077075 on Build GRCh38
rs7874105 Chr.9: 69077164-69077164 on Build GRCh38
rs548189845 Chr.9: 69076909-69076909 on Build GRCh38
rs999057015 Chr.9: 69077268-69077268 on Build GRCh38
rs1010607705 Chr.9: 69076875-69076875 on Build GRCh38

View Full Product Details