Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 85546446-85546988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGTPBP1
Entrez Gene ID
23287
Gene Name
ATP/GTP binding protein 1
Gene Aliases
CCP1, NNA1
UniGene
Hs.719980
Gene Chromosome Location
Chr.9: 85543799-85742029 on Build GRCh38
Transcripts
NM_001286715.1 NM_001286717.1 NM_015239.2 XM_005251847.3 XM_005251848.2 XM_005251849.3 XM_011518420.2 XM_017014544.1 XM_017014546.1 XM_017014548.1

SNP Detail

SNP ID Chromosome Location
rs1007449159 Chr.9: 85546531-85546531 on Build GRCh38
rs944591387 Chr.9: 85546522-85546522 on Build GRCh38
rs902183690 Chr.9: 85546918-85546918 on Build GRCh38
rs778653044 Chr.9: 85546648-85546648 on Build GRCh38
rs970274693 Chr.9: 85546662-85546662 on Build GRCh38
rs918189669 Chr.9: 85546748-85546750 on Build GRCh38
rs1014389643 Chr.9: 85546637-85546637 on Build GRCh38
rs761207368 Chr.9: 85546700-85546700 on Build GRCh38
rs949547507 Chr.9: 85546778-85546778 on Build GRCh38
rs1012427328 Chr.9: 85546947-85546947 on Build GRCh38
rs896197791 Chr.9: 85546934-85546934 on Build GRCh38
rs1039289035 Chr.9: 85546552-85546552 on Build GRCh38
rs984877465 Chr.9: 85546684-85546684 on Build GRCh38
rs532240899 Chr.9: 85546741-85546741 on Build GRCh38
rs902214757 Chr.9: 85546619-85546619 on Build GRCh38
rs999662625 Chr.9: 85546624-85546624 on Build GRCh38
rs946620705 Chr.9: 85546901-85546901 on Build GRCh38
rs944792438 Chr.9: 85546855-85546855 on Build GRCh38
rs934376422 Chr.9: 85546859-85546859 on Build GRCh38
rs1034813885 Chr.9: 85546631-85546631 on Build GRCh38
rs566639533 Chr.9: 85546736-85546736 on Build GRCh38
rs546964504 Chr.9: 85546693-85546693 on Build GRCh38
rs189422340 Chr.9: 85546805-85546805 on Build GRCh38
rs192876044 Chr.9: 85546924-85546924 on Build GRCh38
rs747102708 Chr.9: 85546797-85546797 on Build GRCh38
rs530500405 Chr.9: 85546494-85546494 on Build GRCh38
rs900390705 Chr.9: 85546571-85546571 on Build GRCh38
rs1027638916 Chr.9: 85546678-85546678 on Build GRCh38
rs141421425 Chr.9: 85546919-85546919 on Build GRCh38
rs371155837 Chr.9: 85546495-85546495 on Build GRCh38
rs550189746 Chr.9: 85546873-85546873 on Build GRCh38
rs950787746 Chr.9: 85546708-85546708 on Build GRCh38
rs1017957826 Chr.9: 85546562-85546562 on Build GRCh38
rs924113867 Chr.9: 85546855-85546855 on Build GRCh38
rs561128018 Chr.9: 85546497-85546497 on Build GRCh38
rs946656728 Chr.9: 85546605-85546605 on Build GRCh38
rs921778605 Chr.9: 85546896-85546896 on Build GRCh38
rs938026849 Chr.9: 85546706-85546706 on Build GRCh38
rs895963874 Chr.9: 85546634-85546634 on Build GRCh38
rs973679482 Chr.9: 85546668-85546668 on Build GRCh38
rs1043646855 Chr.9: 85546908-85546908 on Build GRCh38
rs1054628442 Chr.9: 85546872-85546872 on Build GRCh38
rs1038937441 Chr.9: 85546933-85546933 on Build GRCh38
rs3185651 Chr.9: 85546612-85546612 on Build GRCh38
rs796786669 Chr.9: 85546854-85546855 on Build GRCh38
rs909184289 Chr.9: 85546701-85546701 on Build GRCh38
rs991067374 Chr.9: 85546733-85546733 on Build GRCh38

View Full Product Details