Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 86944263-86944793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS1
Entrez Gene ID
2619
Gene Name
growth arrest specific 1
Gene Aliases
-
UniGene
Hs.65029
Gene Chromosome Location
Chr.9: 86944362-86947189 on Build GRCh38
Transcripts
NM_002048.2

SNP Detail

SNP ID Chromosome Location
rs1038872420 Chr.9: 86944403-86944403 on Build GRCh38
rs920580189 Chr.9: 86944460-86944460 on Build GRCh38
rs961226660 Chr.9: 86944768-86944768 on Build GRCh38
rs901633551 Chr.9: 86944423-86944423 on Build GRCh38
rs150795265 Chr.9: 86944318-86944318 on Build GRCh38
rs928986846 Chr.9: 86944336-86944336 on Build GRCh38
rs14441 Chr.9: 86944568-86944568 on Build GRCh38
rs981166232 Chr.9: 86944326-86944326 on Build GRCh38
rs991038035 Chr.9: 86944303-86944303 on Build GRCh38
rs778679503 Chr.9: 86944358-86944358 on Build GRCh38
rs902592704 Chr.9: 86944468-86944468 on Build GRCh38
rs891048190 Chr.9: 86944712-86944712 on Build GRCh38
rs923141683 Chr.9: 86944737-86944737 on Build GRCh38
rs918940471 Chr.9: 86944333-86944333 on Build GRCh38
rs1028625146 Chr.9: 86944707-86944707 on Build GRCh38
rs933183510 Chr.9: 86944746-86944746 on Build GRCh38
rs918045265 Chr.9: 86944304-86944304 on Build GRCh38
rs962140857 Chr.9: 86944440-86944440 on Build GRCh38
rs955376596 Chr.9: 86944414-86944415 on Build GRCh38
rs996702848 Chr.9: 86944375-86944375 on Build GRCh38
rs972885576 Chr.9: 86944586-86944586 on Build GRCh38
rs930654142 Chr.9: 86944460-86944461 on Build GRCh38
rs1020965629 Chr.9: 86944453-86944453 on Build GRCh38
rs1009146671 Chr.9: 86944324-86944324 on Build GRCh38
rs111308204 Chr.9: 86944544-86944544 on Build GRCh38
rs972029299 Chr.9: 86944456-86944456 on Build GRCh38
rs1056606349 Chr.9: 86944307-86944307 on Build GRCh38
rs746159929 Chr.9: 86944631-86944631 on Build GRCh38
rs574443340 Chr.9: 86944335-86944336 on Build GRCh38
rs766902654 Chr.9: 86944415-86944415 on Build GRCh38
rs949832798 Chr.9: 86944315-86944315 on Build GRCh38
rs182783172 Chr.9: 86944753-86944753 on Build GRCh38
rs548631933 Chr.9: 86944646-86944646 on Build GRCh38
rs996503424 Chr.9: 86944682-86944682 on Build GRCh38
rs943300944 Chr.9: 86944685-86944685 on Build GRCh38
rs997573662 Chr.9: 86944425-86944425 on Build GRCh38
rs991441239 Chr.9: 86944557-86944557 on Build GRCh38
rs903529198 Chr.9: 86944677-86944677 on Build GRCh38
rs748088409 Chr.9: 86944386-86944386 on Build GRCh38
rs1032584457 Chr.9: 86944291-86944291 on Build GRCh38
rs1025161090 Chr.9: 86944580-86944580 on Build GRCh38
rs777698497 Chr.9: 86944492-86944492 on Build GRCh38
rs149438850 Chr.9: 86944717-86944717 on Build GRCh38
rs758318639 Chr.9: 86944491-86944491 on Build GRCh38
rs34813745 Chr.9: 86944677-86944678 on Build GRCh38
rs1043481977 Chr.9: 86944671-86944671 on Build GRCh38
rs1042184050 Chr.9: 86944751-86944751 on Build GRCh38
rs546372210 Chr.9: 86944302-86944302 on Build GRCh38
rs971276472 Chr.9: 86944584-86944584 on Build GRCh38
rs1039339072 Chr.9: 86944721-86944721 on Build GRCh38
rs943270312 Chr.9: 86944400-86944400 on Build GRCh38
rs780359012 Chr.9: 86944487-86944487 on Build GRCh38
rs1056636083 Chr.9: 86944608-86944608 on Build GRCh38
rs563221386 Chr.9: 86944600-86944600 on Build GRCh38
rs7869285 Chr.9: 86944619-86944619 on Build GRCh38
rs11795098 Chr.9: 86944642-86944642 on Build GRCh38

View Full Product Details