Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 86946109-86946603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS1
Entrez Gene ID
2619
Gene Name
growth arrest specific 1
Gene Aliases
-
UniGene
Hs.65029
Gene Chromosome Location
Chr.9: 86944362-86947189 on Build GRCh38
Transcripts
NM_002048.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4887438 c.367C>A p.P123T Chr.9: 86946413-86946413 on Build GRCh38
5485095 c.208G>A p.E70K Chr.9: 86946572-86946572 on Build GRCh38
4938663 c.610C>T p.R204C Chr.9: 86946170-86946170 on Build GRCh38
5550948 c.519C>A p.C173* Chr.9: 86946261-86946261 on Build GRCh38
1207923 c.608G>A p.C203Y Chr.9: 86946172-86946172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs387907165 Chr.9: 86946181-86946181 on Build GRCh38
rs994346671 Chr.9: 86946178-86946178 on Build GRCh38
rs752317122 Chr.9: 86946537-86946537 on Build GRCh38
rs977195580 Chr.9: 86946543-86946543 on Build GRCh38
rs994313500 Chr.9: 86946495-86946495 on Build GRCh38
rs758112692 Chr.9: 86946223-86946223 on Build GRCh38
rs772529497 Chr.9: 86946386-86946386 on Build GRCh38
rs759704477 Chr.9: 86946195-86946195 on Build GRCh38
rs907485406 Chr.9: 86946294-86946295 on Build GRCh38
rs762845548 Chr.9: 86946471-86946471 on Build GRCh38
rs781205733 Chr.9: 86946531-86946531 on Build GRCh38
rs1034143238 Chr.9: 86946513-86946513 on Build GRCh38
rs911193660 Chr.9: 86946390-86946390 on Build GRCh38
rs766061802 Chr.9: 86946496-86946496 on Build GRCh38
rs752725446 Chr.9: 86946216-86946216 on Build GRCh38
rs755684527 Chr.9: 86946541-86946541 on Build GRCh38
rs754001203 Chr.9: 86946432-86946432 on Build GRCh38
rs867342975 Chr.9: 86946207-86946207 on Build GRCh38
rs756167741 Chr.9: 86946506-86946507 on Build GRCh38
rs1025359669 Chr.9: 86946214-86946214 on Build GRCh38
rs1041660363 Chr.9: 86946468-86946468 on Build GRCh38
rs531407961 Chr.9: 86946417-86946417 on Build GRCh38
rs896991763 Chr.9: 86946292-86946292 on Build GRCh38
rs769586642 Chr.9: 86946298-86946298 on Build GRCh38
rs767563820 Chr.9: 86946213-86946213 on Build GRCh38
rs764235371 Chr.9: 86946218-86946218 on Build GRCh38
rs917196227 Chr.9: 86946309-86946309 on Build GRCh38
rs768276315 Chr.9: 86946145-86946145 on Build GRCh38
rs773739552 Chr.9: 86946169-86946169 on Build GRCh38
rs750244637 Chr.9: 86946221-86946221 on Build GRCh38
rs533125798 Chr.9: 86946135-86946135 on Build GRCh38
rs373433865 Chr.9: 86946252-86946252 on Build GRCh38
rs564115058 Chr.9: 86946129-86946129 on Build GRCh38
rs201726102 Chr.9: 86946128-86946128 on Build GRCh38
rs1005191877 Chr.9: 86946272-86946272 on Build GRCh38
rs942754517 Chr.9: 86946412-86946412 on Build GRCh38
rs747776390 Chr.9: 86946165-86946165 on Build GRCh38
rs139565812 Chr.9: 86946144-86946144 on Build GRCh38
rs778703916 Chr.9: 86946132-86946132 on Build GRCh38
rs1001770983 Chr.9: 86946308-86946309 on Build GRCh38
rs751417484 Chr.9: 86946231-86946231 on Build GRCh38
rs568303782 Chr.9: 86946559-86946559 on Build GRCh38
rs563401753 Chr.9: 86946291-86946291 on Build GRCh38
rs1032793536 Chr.9: 86946554-86946554 on Build GRCh38
rs775553361 Chr.9: 86946387-86946387 on Build GRCh38
rs777280963 Chr.9: 86946348-86946348 on Build GRCh38
rs774972884 Chr.9: 86946380-86946380 on Build GRCh38
rs760688479 Chr.9: 86946418-86946418 on Build GRCh38
rs977225070 Chr.9: 86946341-86946341 on Build GRCh38
rs529361078 Chr.9: 86946316-86946317 on Build GRCh38
rs776459870 Chr.9: 86946153-86946153 on Build GRCh38
rs989967848 Chr.9: 86946567-86946567 on Build GRCh38
rs777526325 Chr.9: 86946565-86946565 on Build GRCh38
rs780228522 Chr.9: 86946507-86946508 on Build GRCh38
rs757093506 Chr.9: 86946131-86946131 on Build GRCh38
rs771528019 Chr.9: 86946176-86946176 on Build GRCh38
rs184053229 Chr.9: 86946516-86946516 on Build GRCh38
rs771173456 Chr.9: 86946378-86946378 on Build GRCh38
rs911318462 Chr.9: 86946576-86946576 on Build GRCh38
rs888648250 Chr.9: 86946499-86946499 on Build GRCh38
rs754995675 Chr.9: 86946524-86946524 on Build GRCh38
rs371633977 Chr.9: 86946246-86946246 on Build GRCh38
rs549876121 Chr.9: 86946217-86946217 on Build GRCh38
rs1013928267 Chr.9: 86946521-86946521 on Build GRCh38
rs749732161 Chr.9: 86946364-86946364 on Build GRCh38
rs764355861 Chr.9: 86946419-86946419 on Build GRCh38
rs924002473 Chr.9: 86946359-86946359 on Build GRCh38
rs759982942 Chr.9: 86946384-86946384 on Build GRCh38
rs901428369 Chr.9: 86946482-86946482 on Build GRCh38
rs780104648 Chr.9: 86946228-86946228 on Build GRCh38
rs993153385 Chr.9: 86946308-86946308 on Build GRCh38
rs763173224 Chr.9: 86946173-86946173 on Build GRCh38
rs201340761 Chr.9: 86946470-86946470 on Build GRCh38
rs769518745 Chr.9: 86946168-86946168 on Build GRCh38

View Full Product Details