Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 87159675-87160219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf170
Entrez Gene ID
401535
Gene Name
chromosome 9 open reading frame 170
Gene Aliases
-
UniGene
Hs.657740
Gene Chromosome Location
Chr.9: 87148644-87159726 on Build GRCh38
Transcripts
NM_001001709.2

SNP Detail

SNP ID Chromosome Location
rs573424475 Chr.9: 87160110-87160110 on Build GRCh38
rs550833047 Chr.9: 87159846-87159846 on Build GRCh38
rs971617459 Chr.9: 87159828-87159828 on Build GRCh38
rs748932828 Chr.9: 87159815-87159815 on Build GRCh38
rs778826845 Chr.9: 87160026-87160026 on Build GRCh38
rs981609265 Chr.9: 87159866-87159866 on Build GRCh38
rs775183212 Chr.9: 87159765-87159765 on Build GRCh38
rs951169222 Chr.9: 87159766-87159766 on Build GRCh38
rs969362888 Chr.9: 87159728-87159728 on Build GRCh38
rs934298811 Chr.9: 87159926-87159926 on Build GRCh38
rs990282011 Chr.9: 87159909-87159909 on Build GRCh38
rs951096301 Chr.9: 87160155-87160155 on Build GRCh38
rs760399516 Chr.9: 87160131-87160131 on Build GRCh38
rs910025092 Chr.9: 87159794-87159794 on Build GRCh38
rs767385725 Chr.9: 87160097-87160097 on Build GRCh38
rs914256207 Chr.9: 87159923-87159923 on Build GRCh38
rs923224281 Chr.9: 87160033-87160033 on Build GRCh38
rs538742087 Chr.9: 87160152-87160152 on Build GRCh38
rs571138574 Chr.9: 87159713-87159713 on Build GRCh38
rs893614254 Chr.9: 87160034-87160034 on Build GRCh38
rs866693349 Chr.9: 87159734-87159734 on Build GRCh38
rs559039443 Chr.9: 87160179-87160179 on Build GRCh38
rs921496650 Chr.9: 87159897-87159897 on Build GRCh38
rs934707611 Chr.9: 87160008-87160008 on Build GRCh38
rs962475345 Chr.9: 87160187-87160187 on Build GRCh38
rs982529359 Chr.9: 87159784-87159784 on Build GRCh38
rs955817585 Chr.9: 87159901-87159901 on Build GRCh38
rs975607663 Chr.9: 87159895-87159895 on Build GRCh38
rs1035056508 Chr.9: 87160146-87160146 on Build GRCh38
rs1010683839 Chr.9: 87160055-87160055 on Build GRCh38
rs1034724896 Chr.9: 87159755-87159755 on Build GRCh38
rs529766458 Chr.9: 87159969-87159969 on Build GRCh38
rs975576523 Chr.9: 87160188-87160188 on Build GRCh38
rs1038835475 Chr.9: 87160141-87160141 on Build GRCh38
rs553412904 Chr.9: 87159891-87159892 on Build GRCh38
rs1012904443 Chr.9: 87159791-87159791 on Build GRCh38
rs536407367 Chr.9: 87160088-87160088 on Build GRCh38
rs185467056 Chr.9: 87160063-87160063 on Build GRCh38
rs140202217 Chr.9: 87159748-87159748 on Build GRCh38
rs940140809 Chr.9: 87160125-87160125 on Build GRCh38
rs1051528179 Chr.9: 87159932-87159932 on Build GRCh38
rs1010757544 Chr.9: 87159707-87159707 on Build GRCh38
rs575681971 Chr.9: 87160182-87160182 on Build GRCh38
rs1012908424 Chr.9: 87160195-87160195 on Build GRCh38
rs277752 Chr.9: 87160100-87160100 on Build GRCh38
rs1023523018 Chr.9: 87159724-87159724 on Build GRCh38

View Full Product Details