Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136367386-136367919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD9
Entrez Gene ID
64170
Gene Name
caspase recruitment domain family member 9
Gene Aliases
CANDF2, hCARD9
UniGene
Hs.694071
Gene Chromosome Location
Chr.9: 136363956-136373681 on Build GRCh38
Transcripts
NM_052813.4 NM_052814.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
487223 c.1147A>C p.K383Q Chr.9: 136367759-136367759 on Build GRCh38
4878003 c.1240A>G p.R414G Chr.9: 136367666-136367666 on Build GRCh38
4997032 c.1183C>G p.L395V Chr.9: 136367723-136367723 on Build GRCh38
6873433 c.1158G>A p.R386R Chr.9: 136367748-136367748 on Build GRCh38
1624722 c.1091G>A p.R364Q Chr.9: 136367815-136367815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781345390 Chr.9: 136367444-136367444 on Build GRCh38
rs761143967 Chr.9: 136367656-136367656 on Build GRCh38
rs935302620 Chr.9: 136367440-136367440 on Build GRCh38
rs139454076 Chr.9: 136367691-136367691 on Build GRCh38
rs956705516 Chr.9: 136367673-136367673 on Build GRCh38
rs745787091 Chr.9: 136367620-136367621 on Build GRCh38
rs745746683 Chr.9: 136367682-136367682 on Build GRCh38
rs931812368 Chr.9: 136367899-136367899 on Build GRCh38
rs980067180 Chr.9: 136367703-136367703 on Build GRCh38
rs752111756 Chr.9: 136367569-136367569 on Build GRCh38
rs371771167 Chr.9: 136367620-136367620 on Build GRCh38
rs569181286 Chr.9: 136367529-136367529 on Build GRCh38
rs770769219 Chr.9: 136367534-136367534 on Build GRCh38
rs988070503 Chr.9: 136367675-136367675 on Build GRCh38
rs754119986 Chr.9: 136367663-136367663 on Build GRCh38
rs752685448 Chr.9: 136367669-136367669 on Build GRCh38
rs559114246 Chr.9: 136367643-136367643 on Build GRCh38
rs145698391 Chr.9: 136367600-136367600 on Build GRCh38
rs749349863 Chr.9: 136367510-136367510 on Build GRCh38
rs940518611 Chr.9: 136367765-136367765 on Build GRCh38
rs376357452 Chr.9: 136367793-136367793 on Build GRCh38
rs570573080 Chr.9: 136367837-136367837 on Build GRCh38
rs747038239 Chr.9: 136367603-136367603 on Build GRCh38
rs117344324 Chr.9: 136367849-136367849 on Build GRCh38
rs767880614 Chr.9: 136367647-136367647 on Build GRCh38
rs550873526 Chr.9: 136367778-136367778 on Build GRCh38
rs3812555 Chr.9: 136367481-136367481 on Build GRCh38
rs1004762750 Chr.9: 136367468-136367468 on Build GRCh38
rs747645575 Chr.9: 136367723-136367723 on Build GRCh38
rs576925149 Chr.9: 136367679-136367679 on Build GRCh38
rs771660974 Chr.9: 136367724-136367724 on Build GRCh38
rs3812556 Chr.9: 136367539-136367539 on Build GRCh38
rs532713129 Chr.9: 136367767-136367767 on Build GRCh38
rs146829177 Chr.9: 136367810-136367810 on Build GRCh38
rs989407851 Chr.9: 136367690-136367690 on Build GRCh38
rs994614202 Chr.9: 136367436-136367436 on Build GRCh38
rs145574224 Chr.9: 136367792-136367792 on Build GRCh38
rs200523846 Chr.9: 136367826-136367826 on Build GRCh38
rs747580987 Chr.9: 136367787-136367787 on Build GRCh38
rs147340708 Chr.9: 136367662-136367662 on Build GRCh38
rs750740690 Chr.9: 136367448-136367448 on Build GRCh38
rs199848610 Chr.9: 136367685-136367685 on Build GRCh38
rs764216689 Chr.9: 136367798-136367798 on Build GRCh38
rs778454812 Chr.9: 136367769-136367769 on Build GRCh38
rs769777642 Chr.9: 136367775-136367777 on Build GRCh38
rs745691628 Chr.9: 136367612-136367612 on Build GRCh38
rs558475431 Chr.9: 136367625-136367625 on Build GRCh38
rs763452178 Chr.9: 136367560-136367560 on Build GRCh38
rs746866702 Chr.9: 136367530-136367530 on Build GRCh38
rs774516640 Chr.9: 136367848-136367848 on Build GRCh38
rs759838683 Chr.9: 136367739-136367739 on Build GRCh38
rs564811361 Chr.9: 136367520-136367520 on Build GRCh38
rs776755973 Chr.9: 136367556-136367556 on Build GRCh38
rs202124994 Chr.9: 136367857-136367857 on Build GRCh38
rs142533596 Chr.9: 136367730-136367730 on Build GRCh38
rs149712114 Chr.9: 136367788-136367788 on Build GRCh38
rs201988948 Chr.9: 136367850-136367850 on Build GRCh38
rs773952828 Chr.9: 136367702-136367702 on Build GRCh38
rs769142671 Chr.9: 136367834-136367834 on Build GRCh38
rs998085876 Chr.9: 136367493-136367493 on Build GRCh38
rs567395579 Chr.9: 136367731-136367731 on Build GRCh38
rs751616314 Chr.9: 136367762-136367762 on Build GRCh38
rs144193548 Chr.9: 136367684-136367684 on Build GRCh38
rs376063829 Chr.9: 136367579-136367579 on Build GRCh38
rs757745178 Chr.9: 136367818-136367818 on Build GRCh38
rs759837396 Chr.9: 136367789-136367789 on Build GRCh38
rs780456845 Chr.9: 136367598-136367598 on Build GRCh38
rs939899334 Chr.9: 136367889-136367889 on Build GRCh38
rs1052488634 Chr.9: 136367441-136367441 on Build GRCh38
rs374295826 Chr.9: 136367642-136367642 on Build GRCh38
rs762471346 Chr.9: 136367559-136367559 on Build GRCh38
rs948364270 Chr.9: 136367893-136367893 on Build GRCh38
rs754828452 Chr.9: 136367549-136367549 on Build GRCh38
rs781001992 Chr.9: 136367607-136367607 on Build GRCh38
rs754067141 Chr.9: 136367599-136367599 on Build GRCh38
rs560188869 Chr.9: 136367738-136367738 on Build GRCh38
rs760842007 Chr.9: 136367415-136367415 on Build GRCh38
rs566481759 Chr.9: 136367805-136367805 on Build GRCh38
rs549380094 Chr.9: 136367888-136367888 on Build GRCh38
rs572036783 Chr.9: 136367897-136367897 on Build GRCh38
rs533832731 Chr.9: 136367604-136367604 on Build GRCh38
rs372539632 Chr.9: 136367766-136367766 on Build GRCh38
rs780330253 Chr.9: 136367504-136367504 on Build GRCh38
rs3124993 Chr.9: 136367753-136367753 on Build GRCh38
rs538768455 Chr.9: 136367433-136367433 on Build GRCh38
rs962483785 Chr.9: 136367573-136367573 on Build GRCh38
rs1038471982 Chr.9: 136367892-136367892 on Build GRCh38
rs533844645 Chr.9: 136367812-136367812 on Build GRCh38
rs368200571 Chr.9: 136367681-136367681 on Build GRCh38
rs151211588 Chr.9: 136367816-136367816 on Build GRCh38
rs1028697702 Chr.9: 136367628-136367628 on Build GRCh38
rs778706578 Chr.9: 136367815-136367815 on Build GRCh38
rs368479737 Chr.9: 136367615-136367615 on Build GRCh38
rs747779269 Chr.9: 136367777-136367777 on Build GRCh38
rs746544609 Chr.9: 136367821-136367821 on Build GRCh38
rs777420944 Chr.9: 136367785-136367785 on Build GRCh38
rs1049117042 Chr.9: 136367750-136367750 on Build GRCh38
rs574560841 Chr.9: 136367694-136367694 on Build GRCh38
rs541901067 Chr.9: 136367887-136367887 on Build GRCh38
rs756446760 Chr.9: 136367594-136367594 on Build GRCh38
rs141270365 Chr.9: 136367801-136367801 on Build GRCh38
rs746317918 Chr.9: 136367833-136367833 on Build GRCh38
rs926183337 Chr.9: 136367554-136367554 on Build GRCh38
rs1027476446 Chr.9: 136367437-136367437 on Build GRCh38
rs775080826 Chr.9: 136367557-136367557 on Build GRCh38
rs752913001 Chr.9: 136367749-136367749 on Build GRCh38
rs768834477 Chr.9: 136367836-136367836 on Build GRCh38
rs765405752 Chr.9: 136367747-136367747 on Build GRCh38
rs773817103 Chr.9: 136367654-136367654 on Build GRCh38
rs112000318 Chr.9: 136367896-136367896 on Build GRCh38
rs765420080 Chr.9: 136367667-136367667 on Build GRCh38
rs921094576 Chr.9: 136367693-136367693 on Build GRCh38
rs775135852 Chr.9: 136367616-136367616 on Build GRCh38
rs572840818 Chr.9: 136367601-136367601 on Build GRCh38
rs145052557 Chr.9: 136367802-136367802 on Build GRCh38
rs371122898 Chr.9: 136367485-136367485 on Build GRCh38
rs1044401985 Chr.9: 136367593-136367593 on Build GRCh38
rs769402703 Chr.9: 136367551-136367551 on Build GRCh38
rs139646302 Chr.9: 136367768-136367768 on Build GRCh38
rs540891739 Chr.9: 136367761-136367761 on Build GRCh38
rs750795840 Chr.9: 136367588-136367588 on Build GRCh38
rs965714063 Chr.9: 136367482-136367482 on Build GRCh38
rs142757984 Chr.9: 136367646-136367646 on Build GRCh38
rs764093590 Chr.9: 136367758-136367758 on Build GRCh38
rs767984480 Chr.9: 136367584-136367584 on Build GRCh38
rs759231279 Chr.9: 136367550-136367550 on Build GRCh38

View Full Product Details