Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136429684-136430179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5E
Entrez Gene ID
56623
Gene Name
inositol polyphosphate-5-phosphatase E
Gene Aliases
CORS1, CPD4, JBTS1, MORMS, PPI5PIV
UniGene
Hs.120998
Gene Chromosome Location
Chr.9: 136428615-136439861 on Build GRCh38
Transcripts
NM_001318502.1 NM_019892.5 XM_017014926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4692604 c.1842C>T p.Y614Y Chr.9: 136429768-136429768 on Build GRCh38
5512633 c.1809G>A p.P603P Chr.9: 136429801-136429801 on Build GRCh38
3655736 c.1911C>T p.S637S Chr.9: 136429699-136429699 on Build GRCh38
6453898 c.1869G>A p.S623S Chr.9: 136429741-136429741 on Build GRCh38
1106786 c.1834G>T p.E612* Chr.9: 136429776-136429776 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746372090 Chr.9: 136429802-136429802 on Build GRCh38
rs982275578 Chr.9: 136430082-136430082 on Build GRCh38
rs190316580 Chr.9: 136429827-136429827 on Build GRCh38
rs979206393 Chr.9: 136429863-136429881 on Build GRCh38
rs770339939 Chr.9: 136429792-136429792 on Build GRCh38
rs931297186 Chr.9: 136429940-136429940 on Build GRCh38
rs550781430 Chr.9: 136429972-136429972 on Build GRCh38
rs144720715 Chr.9: 136429729-136429729 on Build GRCh38
rs888006187 Chr.9: 136429929-136429929 on Build GRCh38
rs999858067 Chr.9: 136429861-136429861 on Build GRCh38
rs68142670 Chr.9: 136430121-136430121 on Build GRCh38
rs1056991920 Chr.9: 136429772-136429772 on Build GRCh38
rs532667515 Chr.9: 136429971-136429971 on Build GRCh38
rs780279027 Chr.9: 136430013-136430013 on Build GRCh38
rs375147334 Chr.9: 136429873-136429873 on Build GRCh38
rs540871489 Chr.9: 136429950-136429950 on Build GRCh38
rs879708308 Chr.9: 136430033-136430033 on Build GRCh38
rs776136400 Chr.9: 136429794-136429794 on Build GRCh38
rs565476813 Chr.9: 136429969-136429969 on Build GRCh38
rs757272800 Chr.9: 136429733-136429733 on Build GRCh38
rs573682018 Chr.9: 136429996-136429996 on Build GRCh38
rs561573256 Chr.9: 136429919-136429919 on Build GRCh38
rs771894087 Chr.9: 136429782-136429782 on Build GRCh38
rs1057330185 Chr.9: 136429961-136429961 on Build GRCh38
rs141403616 Chr.9: 136430062-136430064 on Build GRCh38
rs760420960 Chr.9: 136429736-136429736 on Build GRCh38
rs73670263 Chr.9: 136430145-136430145 on Build GRCh38
rs770286818 Chr.9: 136429818-136429818 on Build GRCh38
rs558289264 Chr.9: 136429859-136429859 on Build GRCh38
rs1007372343 Chr.9: 136430015-136430015 on Build GRCh38
rs541784197 Chr.9: 136429903-136429903 on Build GRCh38
rs35539554 Chr.9: 136430119-136430120 on Build GRCh38
rs77350282 Chr.9: 136429875-136429875 on Build GRCh38
rs566437568 Chr.9: 136430097-136430097 on Build GRCh38
rs1032160292 Chr.9: 136430028-136430028 on Build GRCh38
rs771705093 Chr.9: 136430138-136430138 on Build GRCh38
rs781634065 Chr.9: 136429881-136429881 on Build GRCh38
rs750470490 Chr.9: 136429844-136429844 on Build GRCh38
rs761979487 Chr.9: 136430037-136430037 on Build GRCh38
rs779919473 Chr.9: 136429810-136429810 on Build GRCh38
rs863225199 Chr.9: 136429712-136429713 on Build GRCh38
rs773063131 Chr.9: 136430144-136430144 on Build GRCh38
rs576569302 Chr.9: 136429860-136429860 on Build GRCh38
rs199875003 Chr.9: 136429780-136429780 on Build GRCh38
rs68142669 Chr.9: 136430121-136430121 on Build GRCh38
rs972820366 Chr.9: 136429911-136429911 on Build GRCh38
rs1046143445 Chr.9: 136429874-136429874 on Build GRCh38
rs769679427 Chr.9: 136430114-136430116 on Build GRCh38
rs533739662 Chr.9: 136429858-136429858 on Build GRCh38
rs926363483 Chr.9: 136430088-136430088 on Build GRCh38
rs777549235 Chr.9: 136429784-136429784 on Build GRCh38
rs957713025 Chr.9: 136429830-136429830 on Build GRCh38
rs763271458 Chr.9: 136429824-136429824 on Build GRCh38
rs148539728 Chr.9: 136429735-136429735 on Build GRCh38
rs142759730 Chr.9: 136429749-136429749 on Build GRCh38
rs1042220932 Chr.9: 136430136-136430136 on Build GRCh38
rs953200980 Chr.9: 136429744-136429744 on Build GRCh38
rs34184721 Chr.9: 136430065-136430067 on Build GRCh38
rs756651906 Chr.9: 136429997-136429997 on Build GRCh38
rs200294819 Chr.9: 136430066-136430068 on Build GRCh38
rs927187221 Chr.9: 136429949-136429949 on Build GRCh38
rs765239506 Chr.9: 136429856-136429856 on Build GRCh38
rs749437887 Chr.9: 136430018-136430018 on Build GRCh38
rs999805870 Chr.9: 136430000-136430000 on Build GRCh38
rs78191714 Chr.9: 136429840-136429840 on Build GRCh38
rs569300457 Chr.9: 136430007-136430007 on Build GRCh38
rs148865848 Chr.9: 136430032-136430032 on Build GRCh38
rs768806260 Chr.9: 136429963-136429980 on Build GRCh38
rs555221247 Chr.9: 136429848-136429863 on Build GRCh38
rs895660048 Chr.9: 136429965-136429965 on Build GRCh38
rs754906374 Chr.9: 136429846-136429846 on Build GRCh38
rs754887212 Chr.9: 136429742-136429742 on Build GRCh38
rs904305556 Chr.9: 136429978-136429978 on Build GRCh38
rs574332963 Chr.9: 136429898-136429898 on Build GRCh38
rs997604012 Chr.9: 136429723-136429723 on Build GRCh38
rs796644246 Chr.9: 136430121-136430122 on Build GRCh38
rs528757201 Chr.9: 136429942-136429942 on Build GRCh38
rs775863954 Chr.9: 136429819-136429819 on Build GRCh38
rs548517449 Chr.9: 136430067-136430067 on Build GRCh38
rs774753290 Chr.9: 136429948-136429948 on Build GRCh38
rs749045132 Chr.9: 136429812-136429814 on Build GRCh38
rs763184652 Chr.9: 136429713-136429713 on Build GRCh38
rs375144690 Chr.9: 136429790-136429790 on Build GRCh38
rs72775768 Chr.9: 136430122-136430122 on Build GRCh38
rs931184217 Chr.9: 136429714-136429714 on Build GRCh38
rs764347779 Chr.9: 136429826-136429826 on Build GRCh38
rs776349871 Chr.9: 136430106-136430106 on Build GRCh38
rs555224966 Chr.9: 136429886-136429887 on Build GRCh38
rs918708296 Chr.9: 136429937-136429937 on Build GRCh38
rs757518929 Chr.9: 136429880-136429880 on Build GRCh38
rs1020669897 Chr.9: 136430050-136430050 on Build GRCh38
rs764259572 Chr.9: 136429716-136429716 on Build GRCh38
rs121918127 Chr.9: 136429731-136429731 on Build GRCh38

View Full Product Details