Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137251897-137252392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf173-AS1
Entrez Gene ID
100129722
Gene Name
C9orf173 antisense RNA 1
Gene Aliases
-
UniGene
Hs.656347
Gene Chromosome Location
Chr.9: 137250219-137253497 on Build GRCh38
Transcripts
NR_038389.1 NM_001004353.3 NM_001256699.1 NM_001256700.1 NM_001256701.1 NR_046339.1 NR_046340.1 XM_006717113.2 XM_006717115.2

Gene Symbol
STPG3
Entrez Gene ID
441476
Gene Name
sperm-tail PG-rich repeat containing 3
Gene Aliases
C9orf173
UniGene
Hs.372640
Gene Chromosome Location
Chr.9: 137250840-137253483 on Build GRCh38
Transcripts
NR_038389.1 NM_001004353.3 NM_001256699.1 NM_001256700.1 NM_001256701.1 NR_046339.1 NR_046340.1 XM_006717113.2 XM_006717115.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4453593 c.319A>G p.T107A Chr.9: 137252051-137252051 on Build GRCh38
1624789 c.351delA p.E118fs*83 Chr.9: 137252083-137252083 on Build GRCh38
6053733 c.350G>A p.R117Q Chr.9: 137252082-137252082 on Build GRCh38
3655996 c.344C>T p.S115F Chr.9: 137252076-137252076 on Build GRCh38
6616891 c.403+1_403+2delGT p.? Chr.9: 137252136-137252137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747878784 Chr.9: 137252113-137252113 on Build GRCh38
rs529802699 Chr.9: 137251953-137251953 on Build GRCh38
rs776937400 Chr.9: 137251959-137251959 on Build GRCh38
rs895931862 Chr.9: 137252214-137252214 on Build GRCh38
rs987686234 Chr.9: 137252285-137252285 on Build GRCh38
rs536211493 Chr.9: 137252223-137252223 on Build GRCh38
rs200274542 Chr.9: 137252012-137252012 on Build GRCh38
rs780427182 Chr.9: 137252093-137252093 on Build GRCh38
rs1029237763 Chr.9: 137252272-137252272 on Build GRCh38
rs762577860 Chr.9: 137251969-137251969 on Build GRCh38
rs1011510684 Chr.9: 137251990-137251990 on Build GRCh38
rs771623567 Chr.9: 137252181-137252181 on Build GRCh38
rs1014904842 Chr.9: 137252251-137252251 on Build GRCh38
rs760153294 Chr.9: 137252019-137252019 on Build GRCh38
rs562254530 Chr.9: 137252271-137252271 on Build GRCh38
rs921730775 Chr.9: 137252355-137252355 on Build GRCh38
rs988968210 Chr.9: 137252308-137252308 on Build GRCh38
rs77141550 Chr.9: 137251952-137251952 on Build GRCh38
rs779176024 Chr.9: 137251939-137251939 on Build GRCh38
rs762080166 Chr.9: 137252145-137252145 on Build GRCh38
rs781195802 Chr.9: 137252090-137252090 on Build GRCh38
rs771601584 Chr.9: 137252114-137252114 on Build GRCh38
rs775732219 Chr.9: 137252056-137252056 on Build GRCh38
rs756596541 Chr.9: 137252155-137252155 on Build GRCh38
rs755909044 Chr.9: 137252014-137252014 on Build GRCh38
rs995041007 Chr.9: 137252268-137252268 on Build GRCh38
rs375821259 Chr.9: 137252132-137252132 on Build GRCh38
rs767567232 Chr.9: 137252146-137252146 on Build GRCh38
rs767586749 Chr.9: 137252003-137252003 on Build GRCh38
rs766253964 Chr.9: 137252119-137252119 on Build GRCh38
rs757710984 Chr.9: 137252038-137252038 on Build GRCh38
rs1035360887 Chr.9: 137252279-137252279 on Build GRCh38
rs748905558 Chr.9: 137252066-137252066 on Build GRCh38
rs28657439 Chr.9: 137252108-137252108 on Build GRCh38
rs747454041 Chr.9: 137251954-137251954 on Build GRCh38
rs751119451 Chr.9: 137251984-137251984 on Build GRCh38
rs772138076 Chr.9: 137251941-137251941 on Build GRCh38
rs542399200 Chr.9: 137252257-137252257 on Build GRCh38
rs546655671 Chr.9: 137252362-137252362 on Build GRCh38
rs770102695 Chr.9: 137251965-137251965 on Build GRCh38
rs200326940 Chr.9: 137252164-137252164 on Build GRCh38
rs59404109 Chr.9: 137252248-137252248 on Build GRCh38
rs771430538 Chr.9: 137251958-137251958 on Build GRCh38
rs574552044 Chr.9: 137252107-137252107 on Build GRCh38
rs369986377 Chr.9: 137252167-137252167 on Build GRCh38
rs1056939619 Chr.9: 137252361-137252361 on Build GRCh38
rs777758620 Chr.9: 137252027-137252027 on Build GRCh38
rs757293942 Chr.9: 137252088-137252088 on Build GRCh38
rs183312524 Chr.9: 137251970-137251970 on Build GRCh38
rs933438046 Chr.9: 137252356-137252356 on Build GRCh38
rs372285801 Chr.9: 137252153-137252153 on Build GRCh38
rs765646622 Chr.9: 137252075-137252089 on Build GRCh38
rs937161195 Chr.9: 137252343-137252343 on Build GRCh38
rs756584595 Chr.9: 137252092-137252092 on Build GRCh38
rs751827557 Chr.9: 137252087-137252087 on Build GRCh38
rs764319164 Chr.9: 137252082-137252082 on Build GRCh38
rs780404409 Chr.9: 137252165-137252165 on Build GRCh38
rs368075675 Chr.9: 137252081-137252081 on Build GRCh38
rs965132514 Chr.9: 137252297-137252297 on Build GRCh38
rs1019543772 Chr.9: 137252295-137252295 on Build GRCh38
rs140898894 Chr.9: 137252068-137252068 on Build GRCh38
rs759965569 Chr.9: 137251964-137251964 on Build GRCh38
rs927218966 Chr.9: 137252333-137252333 on Build GRCh38
rs763736353 Chr.9: 137251921-137251921 on Build GRCh38
rs1035814707 Chr.9: 137252030-137252030 on Build GRCh38
rs974590908 Chr.9: 137252351-137252351 on Build GRCh38
rs28567631 Chr.9: 137252199-137252199 on Build GRCh38
rs780984226 Chr.9: 137252067-137252067 on Build GRCh38
rs1015579276 Chr.9: 137252047-137252047 on Build GRCh38
rs760591785 Chr.9: 137252078-137252078 on Build GRCh38
rs914418782 Chr.9: 137252131-137252131 on Build GRCh38
rs74822962 Chr.9: 137252074-137252074 on Build GRCh38
rs373873739 Chr.9: 137252133-137252133 on Build GRCh38
rs750265081 Chr.9: 137251932-137251932 on Build GRCh38
rs746402737 Chr.9: 137252052-137252052 on Build GRCh38
rs764901330 Chr.9: 137252260-137252260 on Build GRCh38
rs771051728 Chr.9: 137252008-137252008 on Build GRCh38
rs571553502 Chr.9: 137251981-137251981 on Build GRCh38
rs777292581 Chr.9: 137252185-137252185 on Build GRCh38
rs749694392 Chr.9: 137252102-137252102 on Build GRCh38
rs1004908627 Chr.9: 137252035-137252035 on Build GRCh38
rs747119528 Chr.9: 137252032-137252032 on Build GRCh38
rs755109847 Chr.9: 137251938-137251938 on Build GRCh38
rs955659276 Chr.9: 137252309-137252309 on Build GRCh38
rs748363166 Chr.9: 137251940-137251940 on Build GRCh38
rs912868505 Chr.9: 137252316-137252316 on Build GRCh38
rs1023158600 Chr.9: 137252203-137252203 on Build GRCh38
rs766135937 Chr.9: 137252080-137252080 on Build GRCh38
rs143046453 Chr.9: 137252073-137252073 on Build GRCh38
rs776503611 Chr.9: 137252014-137252014 on Build GRCh38
rs781707372 Chr.9: 137252041-137252041 on Build GRCh38
rs937229736 Chr.9: 137252194-137252194 on Build GRCh38
rs757274084 Chr.9: 137251924-137251924 on Build GRCh38
rs899178700 Chr.9: 137252252-137252252 on Build GRCh38
rs554001293 Chr.9: 137252115-137252115 on Build GRCh38
rs549862258 Chr.9: 137252156-137252156 on Build GRCh38
rs201378131 Chr.9: 137251998-137251998 on Build GRCh38
rs978673282 Chr.9: 137252326-137252326 on Build GRCh38
rs892954145 Chr.9: 137251977-137251977 on Build GRCh38
rs558801799 Chr.9: 137252013-137252013 on Build GRCh38
rs947070792 Chr.9: 137252318-137252318 on Build GRCh38
rs201819623 Chr.9: 137252138-137252138 on Build GRCh38
rs758629910 Chr.9: 137252017-137252017 on Build GRCh38
rs1005392411 Chr.9: 137252246-137252246 on Build GRCh38
rs570432346 Chr.9: 137252327-137252327 on Build GRCh38
rs199636581 Chr.9: 137252166-137252166 on Build GRCh38
rs950954245 Chr.9: 137252044-137252044 on Build GRCh38
rs779784778 Chr.9: 137252015-137252015 on Build GRCh38
rs527929748 Chr.9: 137252349-137252349 on Build GRCh38
rs752954038 Chr.9: 137252016-137252016 on Build GRCh38
rs371624796 Chr.9: 137252055-137252055 on Build GRCh38
rs747748101 Chr.9: 137252175-137252175 on Build GRCh38
rs769193616 Chr.9: 137252104-137252104 on Build GRCh38
rs368386014 Chr.9: 137252140-137252140 on Build GRCh38
rs760590801 Chr.9: 137252118-137252118 on Build GRCh38
rs775177257 Chr.9: 137251967-137251967 on Build GRCh38
rs1051758219 Chr.9: 137252365-137252365 on Build GRCh38
rs374788223 Chr.9: 137252077-137252077 on Build GRCh38
rs781070039 Chr.9: 137251929-137251929 on Build GRCh38
rs953526008 Chr.9: 137252276-137252276 on Build GRCh38

View Full Product Details