Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 124877136-124877640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARPC5L
Entrez Gene ID
81873
Gene Name
actin related protein 2/3 complex subunit 5 like
Gene Aliases
ARC16-2
UniGene
Hs.132499
Gene Chromosome Location
Chr.9: 124862118-124877733 on Build GRCh38
Transcripts
NM_030978.2 XM_011519067.1

SNP Detail

SNP ID Chromosome Location
rs961959036 Chr.9: 124877534-124877534 on Build GRCh38
rs951973259 Chr.9: 124877319-124877319 on Build GRCh38
rs1033159476 Chr.9: 124877195-124877195 on Build GRCh38
rs992767918 Chr.9: 124877515-124877515 on Build GRCh38
rs952127568 Chr.9: 124877502-124877502 on Build GRCh38
rs939541496 Chr.9: 124877394-124877394 on Build GRCh38
rs748337040 Chr.9: 124877551-124877551 on Build GRCh38
rs887706634 Chr.9: 124877260-124877260 on Build GRCh38
rs139065613 Chr.9: 124877191-124877191 on Build GRCh38
rs527448400 Chr.9: 124877308-124877308 on Build GRCh38
rs202100752 Chr.9: 124877439-124877440 on Build GRCh38
rs186557918 Chr.9: 124877453-124877453 on Build GRCh38
rs540254586 Chr.9: 124877556-124877556 on Build GRCh38
rs996682499 Chr.9: 124877486-124877486 on Build GRCh38
rs374663500 Chr.9: 124877484-124877486 on Build GRCh38
rs939468562 Chr.9: 124877216-124877217 on Build GRCh38
rs955990607 Chr.9: 124877483-124877484 on Build GRCh38
rs547735802 Chr.9: 124877330-124877330 on Build GRCh38
rs752298427 Chr.9: 124877236-124877236 on Build GRCh38
rs562336505 Chr.9: 124877229-124877229 on Build GRCh38
rs561118017 Chr.9: 124877331-124877331 on Build GRCh38
rs142994484 Chr.9: 124877422-124877422 on Build GRCh38
rs961866868 Chr.9: 124877371-124877371 on Build GRCh38
rs936375001 Chr.9: 124877421-124877421 on Build GRCh38
rs117721363 Chr.9: 124877287-124877287 on Build GRCh38
rs1014848374 Chr.9: 124877529-124877529 on Build GRCh38
rs929313433 Chr.9: 124877417-124877417 on Build GRCh38
rs11552913 Chr.9: 124877340-124877340 on Build GRCh38
rs758821666 Chr.9: 124877389-124877389 on Build GRCh38
rs765490596 Chr.9: 124877376-124877376 on Build GRCh38
rs991958351 Chr.9: 124877547-124877547 on Build GRCh38
rs1006257387 Chr.9: 124877525-124877525 on Build GRCh38
rs755055086 Chr.9: 124877494-124877494 on Build GRCh38
rs907820116 Chr.9: 124877349-124877349 on Build GRCh38
rs980913351 Chr.9: 124877589-124877589 on Build GRCh38
rs574837426 Chr.9: 124877531-124877531 on Build GRCh38
rs537341560 Chr.9: 124877576-124877576 on Build GRCh38
rs147685245 Chr.9: 124877301-124877302 on Build GRCh38
rs969143835 Chr.9: 124877570-124877570 on Build GRCh38
rs1049023837 Chr.9: 124877562-124877562 on Build GRCh38
rs993926683 Chr.9: 124877256-124877256 on Build GRCh38
rs554854641 Chr.9: 124877503-124877503 on Build GRCh38
rs1051321 Chr.9: 124877211-124877211 on Build GRCh38
rs556153087 Chr.9: 124877594-124877594 on Build GRCh38
rs751616833 Chr.9: 124877258-124877259 on Build GRCh38
rs1010392933 Chr.9: 124877250-124877250 on Build GRCh38
rs974734919 Chr.9: 124877283-124877283 on Build GRCh38
rs926884815 Chr.9: 124877511-124877513 on Build GRCh38
rs531882169 Chr.9: 124877274-124877274 on Build GRCh38
rs767018203 Chr.9: 124877196-124877196 on Build GRCh38
rs56180964 Chr.9: 124877479-124877479 on Build GRCh38
rs913450098 Chr.9: 124877610-124877610 on Build GRCh38
rs1051358 Chr.9: 124877416-124877416 on Build GRCh38
rs1040525065 Chr.9: 124877449-124877449 on Build GRCh38
rs190588960 Chr.9: 124877277-124877277 on Build GRCh38
rs939530609 Chr.9: 124877513-124877513 on Build GRCh38
rs1031862577 Chr.9: 124877312-124877313 on Build GRCh38
rs181124041 Chr.9: 124877377-124877377 on Build GRCh38
rs552371717 Chr.9: 124877425-124877425 on Build GRCh38
rs540132206 Chr.9: 124877286-124877287 on Build GRCh38
rs892083947 Chr.9: 124877432-124877432 on Build GRCh38
rs1055107795 Chr.9: 124877431-124877431 on Build GRCh38
rs1052051159 Chr.9: 124877605-124877605 on Build GRCh38
rs576135544 Chr.9: 124877257-124877257 on Build GRCh38
rs1021318064 Chr.9: 124877487-124877487 on Build GRCh38
rs767507117 Chr.9: 124877355-124877356 on Build GRCh38
rs924960691 Chr.9: 124877419-124877419 on Build GRCh38
rs1031230301 Chr.9: 124877448-124877462 on Build GRCh38
rs1010319273 Chr.9: 124877434-124877434 on Build GRCh38
rs915462208 Chr.9: 124877370-124877371 on Build GRCh38
rs936329963 Chr.9: 124877600-124877600 on Build GRCh38
rs750850222 Chr.9: 124877381-124877381 on Build GRCh38
rs534661033 Chr.9: 124877413-124877413 on Build GRCh38

View Full Product Details