Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127311554-127312062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GARNL3
Entrez Gene ID
84253
Gene Name
GTPase activating Rap/RanGAP domain like 3
Gene Aliases
bA356B19.1
UniGene
Hs.29304
Gene Chromosome Location
Chr.9: 127224471-127393655 on Build GRCh38
Transcripts
NM_001286779.1 NM_032293.4 NR_104591.1 XM_005252267.3 XM_005252268.2 XM_011519086.2 XM_011519087.2 XM_011519089.2 XM_011519090.2 XM_017015202.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4609423 c.229G>T p.A77S Chr.9: 127311645-127311645 on Build GRCh38
6114645 c.230C>G p.A77G Chr.9: 127311646-127311646 on Build GRCh38
6472922 c.298T>C p.F100L Chr.9: 127311714-127311714 on Build GRCh38
6472923 c.299T>C p.F100S Chr.9: 127311715-127311715 on Build GRCh38
6241470 c.246G>T p.W82C Chr.9: 127311662-127311662 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545452691 Chr.9: 127311758-127311758 on Build GRCh38
rs758618292 Chr.9: 127311655-127311655 on Build GRCh38
rs900437894 Chr.9: 127311951-127311951 on Build GRCh38
rs759576789 Chr.9: 127311645-127311645 on Build GRCh38
rs529225227 Chr.9: 127311750-127311750 on Build GRCh38
rs369870685 Chr.9: 127311772-127311772 on Build GRCh38
rs547920013 Chr.9: 127311885-127311885 on Build GRCh38
rs549164439 Chr.9: 127311791-127311791 on Build GRCh38
rs1009797388 Chr.9: 127311996-127311996 on Build GRCh38
rs1042952391 Chr.9: 127311911-127311911 on Build GRCh38
rs566988999 Chr.9: 127312025-127312025 on Build GRCh38
rs16929767 Chr.9: 127311584-127311584 on Build GRCh38
rs771696173 Chr.9: 127311622-127311622 on Build GRCh38
rs757517107 Chr.9: 127311683-127311683 on Build GRCh38
rs760317876 Chr.9: 127311640-127311640 on Build GRCh38
rs906948991 Chr.9: 127311746-127311746 on Build GRCh38
rs865832194 Chr.9: 127311674-127311674 on Build GRCh38
rs772679098 Chr.9: 127311786-127311786 on Build GRCh38
rs866551776 Chr.9: 127311800-127311800 on Build GRCh38
rs1033860714 Chr.9: 127311586-127311586 on Build GRCh38
rs375505893 Chr.9: 127311743-127311743 on Build GRCh38
rs564707604 Chr.9: 127311901-127311901 on Build GRCh38
rs762061564 Chr.9: 127311782-127311783 on Build GRCh38
rs201202835 Chr.9: 127311597-127311597 on Build GRCh38
rs746694411 Chr.9: 127311787-127311787 on Build GRCh38
rs776960605 Chr.9: 127312027-127312027 on Build GRCh38
rs908568076 Chr.9: 127311580-127311580 on Build GRCh38
rs891365071 Chr.9: 127311946-127311946 on Build GRCh38
rs150439873 Chr.9: 127311876-127311876 on Build GRCh38
rs1030152626 Chr.9: 127312008-127312008 on Build GRCh38
rs777791714 Chr.9: 127311661-127311661 on Build GRCh38
rs772981436 Chr.9: 127311624-127311624 on Build GRCh38
rs372166937 Chr.9: 127311704-127311704 on Build GRCh38
rs774708478 Chr.9: 127311775-127311776 on Build GRCh38
rs777235839 Chr.9: 127311844-127311844 on Build GRCh38
rs765304842 Chr.9: 127311652-127311652 on Build GRCh38
rs572477023 Chr.9: 127311795-127311795 on Build GRCh38
rs574679844 Chr.9: 127311576-127311576 on Build GRCh38
rs1038419931 Chr.9: 127311869-127311869 on Build GRCh38
rs200369774 Chr.9: 127311785-127311785 on Build GRCh38
rs932664143 Chr.9: 127311906-127311906 on Build GRCh38
rs746273864 Chr.9: 127311718-127311718 on Build GRCh38
rs75828652 Chr.9: 127311813-127311813 on Build GRCh38
rs547091148 Chr.9: 127312017-127312017 on Build GRCh38
rs770644118 Chr.9: 127311788-127311788 on Build GRCh38
rs767867538 Chr.9: 127311787-127311788 on Build GRCh38
rs1051565393 Chr.9: 127311921-127311921 on Build GRCh38
rs562389326 Chr.9: 127311863-127311863 on Build GRCh38
rs986753672 Chr.9: 127311579-127311579 on Build GRCh38
rs983272965 Chr.9: 127311790-127311790 on Build GRCh38
rs891485029 Chr.9: 127311866-127311866 on Build GRCh38
rs920183809 Chr.9: 127311648-127311648 on Build GRCh38
rs959036818 Chr.9: 127311607-127311607 on Build GRCh38
rs766207673 Chr.9: 127311642-127311642 on Build GRCh38
rs146318751 Chr.9: 127311654-127311654 on Build GRCh38
rs576521049 Chr.9: 127311677-127311677 on Build GRCh38
rs533572952 Chr.9: 127312016-127312016 on Build GRCh38
rs995928746 Chr.9: 127311976-127311976 on Build GRCh38

View Full Product Details