Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127785862-127786368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK9
Entrez Gene ID
1025
Gene Name
cyclin dependent kinase 9
Gene Aliases
C-2k, CDC2L4, CTK1, PITALRE, TAK
UniGene
Hs.150423
Gene Chromosome Location
Chr.9: 127786026-127790782 on Build GRCh38
Transcripts
NM_001261.3 XM_017014184.1 NR_036055.1 NR_039767.1

Gene Symbol
MIR2861
Entrez Gene ID
100422910
Gene Name
microRNA 2861
Gene Aliases
BMND15, MIRN2861
UniGene
Gene Chromosome Location
Chr.9: 127785918-127786007 on Build GRCh38
Transcripts
NM_001261.3 XM_017014184.1 NR_036055.1 NR_039767.1

Gene Symbol
MIR3960
Entrez Gene ID
100616250
Gene Name
microRNA 3960
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.9: 127785833-127785923 on Build GRCh38
Transcripts
NM_001261.3 XM_017014184.1 NR_036055.1 NR_039767.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3326059 c.55T>C p.Y19H Chr.9: 127786203-127786203 on Build GRCh38
5795377 c.408C>G p.Y136* Chr.9: 127786205-127786205 on Build GRCh38
7360673 c.406T>C p.Y136H Chr.9: 127786203-127786203 on Build GRCh38
4965711 c.285G>T p.A95A Chr.9: 127786082-127786082 on Build GRCh38
7360671 c.18C>G p.D6E Chr.9: 127786166-127786166 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544840251 Chr.9: 127786110-127786110 on Build GRCh38
rs749997551 Chr.9: 127786092-127786092 on Build GRCh38
rs779209522 Chr.9: 127786285-127786285 on Build GRCh38
rs771229305 Chr.9: 127786082-127786082 on Build GRCh38
rs756969440 Chr.9: 127786074-127786076 on Build GRCh38
rs935629648 Chr.9: 127785995-127785995 on Build GRCh38
rs748038622 Chr.9: 127786079-127786079 on Build GRCh38
rs764963508 Chr.9: 127786145-127786145 on Build GRCh38
rs1008052903 Chr.9: 127786307-127786308 on Build GRCh38
rs772188304 Chr.9: 127786106-127786106 on Build GRCh38
rs754831384 Chr.9: 127786098-127786098 on Build GRCh38
rs779551319 Chr.9: 127786056-127786056 on Build GRCh38
rs1044227509 Chr.9: 127785879-127785905 on Build GRCh38
rs753542894 Chr.9: 127786144-127786144 on Build GRCh38
rs915617210 Chr.9: 127786006-127786006 on Build GRCh38
rs757364410 Chr.9: 127786169-127786169 on Build GRCh38
rs17849564 Chr.9: 127786154-127786154 on Build GRCh38
rs948517620 Chr.9: 127786023-127786023 on Build GRCh38
rs1054172248 Chr.9: 127785896-127785897 on Build GRCh38
rs774932151 Chr.9: 127786241-127786241 on Build GRCh38
rs765744959 Chr.9: 127786136-127786136 on Build GRCh38
rs536203650 Chr.9: 127785935-127785935 on Build GRCh38
rs778807180 Chr.9: 127786121-127786122 on Build GRCh38
rs747857637 Chr.9: 127786275-127786277 on Build GRCh38
rs868808605 Chr.9: 127785925-127785925 on Build GRCh38
rs781165539 Chr.9: 127786274-127786274 on Build GRCh38
rs755556423 Chr.9: 127786100-127786101 on Build GRCh38
rs749303038 Chr.9: 127786106-127786107 on Build GRCh38
rs775888687 Chr.9: 127786118-127786118 on Build GRCh38
rs765016720 Chr.9: 127786257-127786257 on Build GRCh38
rs772498938 Chr.9: 127786289-127786289 on Build GRCh38
rs1045249467 Chr.9: 127785900-127785900 on Build GRCh38
rs755852257 Chr.9: 127786008-127786008 on Build GRCh38
rs986342846 Chr.9: 127785940-127785940 on Build GRCh38
rs966341548 Chr.9: 127785973-127785973 on Build GRCh38
rs769065822 Chr.9: 127786284-127786284 on Build GRCh38
rs889179952 Chr.9: 127786084-127786084 on Build GRCh38
rs777792275 Chr.9: 127786163-127786163 on Build GRCh38
rs1053054981 Chr.9: 127786295-127786295 on Build GRCh38
rs755547628 Chr.9: 127786094-127786094 on Build GRCh38
rs779537539 Chr.9: 127786101-127786122 on Build GRCh38
rs930904126 Chr.9: 127786301-127786301 on Build GRCh38
rs989783533 Chr.9: 127786003-127786003 on Build GRCh38
rs748529804 Chr.9: 127786288-127786288 on Build GRCh38
rs146765767 Chr.9: 127786219-127786219 on Build GRCh38
rs747946620 Chr.9: 127786103-127786103 on Build GRCh38
rs765530478 Chr.9: 127786089-127786089 on Build GRCh38
rs535039188 Chr.9: 127786208-127786208 on Build GRCh38
rs779011982 Chr.9: 127786073-127786073 on Build GRCh38
rs757201394 Chr.9: 127786271-127786271 on Build GRCh38
rs1012795846 Chr.9: 127785900-127785901 on Build GRCh38
rs529276597 Chr.9: 127786336-127786336 on Build GRCh38
rs747148423 Chr.9: 127786108-127786108 on Build GRCh38
rs1049517320 Chr.9: 127786302-127786302 on Build GRCh38
rs781197980 Chr.9: 127786198-127786198 on Build GRCh38
rs1042735468 Chr.9: 127786316-127786316 on Build GRCh38
rs908537095 Chr.9: 127786141-127786141 on Build GRCh38
rs1014923565 Chr.9: 127785923-127785923 on Build GRCh38
rs964495120 Chr.9: 127786104-127786104 on Build GRCh38
rs770049111 Chr.9: 127786212-127786212 on Build GRCh38
rs566144307 Chr.9: 127786174-127786174 on Build GRCh38
rs780276638 Chr.9: 127786282-127786282 on Build GRCh38
rs572778243 Chr.9: 127785920-127785920 on Build GRCh38
rs530750745 Chr.9: 127786310-127786311 on Build GRCh38
rs753378672 Chr.9: 127786095-127786095 on Build GRCh38
rs751365977 Chr.9: 127786268-127786268 on Build GRCh38
rs998617610 Chr.9: 127786317-127786318 on Build GRCh38
rs560331821 Chr.9: 127786317-127786317 on Build GRCh38
rs962073919 Chr.9: 127785932-127785932 on Build GRCh38
rs11403971 Chr.9: 127786311-127786312 on Build GRCh38
rs777544420 Chr.9: 127786107-127786107 on Build GRCh38
rs1023047310 Chr.9: 127786342-127786342 on Build GRCh38
rs987036880 Chr.9: 127786124-127786124 on Build GRCh38
rs898084298 Chr.9: 127786075-127786075 on Build GRCh38
rs763730809 Chr.9: 127786267-127786267 on Build GRCh38
rs948840067 Chr.9: 127786246-127786246 on Build GRCh38
rs745986749 Chr.9: 127786276-127786276 on Build GRCh38
rs762157126 Chr.9: 127786247-127786247 on Build GRCh38
rs1028459182 Chr.9: 127786117-127786117 on Build GRCh38
rs977342159 Chr.9: 127785975-127785975 on Build GRCh38
rs953500441 Chr.9: 127785937-127785937 on Build GRCh38
rs368784318 Chr.9: 127786114-127786114 on Build GRCh38
rs754847349 Chr.9: 127786122-127786141 on Build GRCh38
rs891461471 Chr.9: 127786307-127786307 on Build GRCh38
rs758230646 Chr.9: 127786153-127786153 on Build GRCh38
rs906755142 Chr.9: 127785909-127785909 on Build GRCh38
rs768527415 Chr.9: 127786069-127786069 on Build GRCh38
rs763517666 Chr.9: 127786091-127786091 on Build GRCh38
rs1045623972 Chr.9: 127786039-127786039 on Build GRCh38
rs1018801032 Chr.9: 127785960-127785960 on Build GRCh38
rs889156287 Chr.9: 127786304-127786304 on Build GRCh38
rs906733245 Chr.9: 127786055-127786055 on Build GRCh38
rs759089495 Chr.9: 127786252-127786252 on Build GRCh38
rs776332336 Chr.9: 127786251-127786251 on Build GRCh38
rs190633494 Chr.9: 127786258-127786258 on Build GRCh38
rs749683780 Chr.9: 127786223-127786223 on Build GRCh38
rs994921744 Chr.9: 127785933-127785933 on Build GRCh38
rs749950582 Chr.9: 127786077-127786100 on Build GRCh38
rs556088007 Chr.9: 127786101-127786101 on Build GRCh38
rs774680949 Chr.9: 127786127-127786127 on Build GRCh38
rs756145683 Chr.9: 127786281-127786281 on Build GRCh38
rs940049375 Chr.9: 127786172-127786172 on Build GRCh38
rs772280389 Chr.9: 127786080-127786080 on Build GRCh38
rs752582972 Chr.9: 127786147-127786147 on Build GRCh38
rs71380090 Chr.9: 127786318-127786319 on Build GRCh38
rs794728003 Chr.9: 127785950-127785950 on Build GRCh38
rs894229025 Chr.9: 127785898-127785898 on Build GRCh38
rs370750864 Chr.9: 127786220-127786220 on Build GRCh38
rs759915993 Chr.9: 127786085-127786085 on Build GRCh38
rs773403021 Chr.9: 127786081-127786081 on Build GRCh38
rs924277493 Chr.9: 127785984-127785984 on Build GRCh38
rs575961189 Chr.9: 127786109-127786109 on Build GRCh38
rs746864836 Chr.9: 127786168-127786168 on Build GRCh38
rs1028429653 Chr.9: 127786345-127786345 on Build GRCh38
rs768849649 Chr.9: 127786232-127786232 on Build GRCh38
rs749147935 Chr.9: 127786065-127786065 on Build GRCh38
rs974181253 Chr.9: 127786217-127786217 on Build GRCh38
rs767108124 Chr.9: 127786073-127786074 on Build GRCh38
rs760251322 Chr.9: 127786133-127786133 on Build GRCh38
rs901943098 Chr.9: 127786314-127786315 on Build GRCh38
rs545030944 Chr.9: 127786249-127786249 on Build GRCh38
rs1011618476 Chr.9: 127786310-127786310 on Build GRCh38

View Full Product Details