Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127789095-127789607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK9
Entrez Gene ID
1025
Gene Name
cyclin dependent kinase 9
Gene Aliases
C-2k, CDC2L4, CTK1, PITALRE, TAK
UniGene
Hs.150423
Gene Chromosome Location
Chr.9: 127786026-127790782 on Build GRCh38
Transcripts
NM_001261.3 XM_017014184.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7360674 c.1390A>T p.S464C Chr.9: 127789463-127789463 on Build GRCh38
4980428 c.1227C>T p.I409I Chr.9: 127789300-127789300 on Build GRCh38
3952353 c.966C>T p.S322S Chr.9: 127789390-127789390 on Build GRCh38
5888037 c.754-4C>T p.? Chr.9: 127789174-127789174 on Build GRCh38
7360672 c.1369G>A p.A457T Chr.9: 127789442-127789442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749397034 Chr.9: 127789388-127789388 on Build GRCh38
rs757828715 Chr.9: 127789142-127789142 on Build GRCh38
rs572065012 Chr.9: 127789267-127789267 on Build GRCh38
rs780095447 Chr.9: 127789387-127789387 on Build GRCh38
rs775765477 Chr.9: 127789301-127789301 on Build GRCh38
rs777977604 Chr.9: 127789568-127789568 on Build GRCh38
rs3217748 Chr.9: 127789170-127789170 on Build GRCh38
rs760988808 Chr.9: 127789434-127789434 on Build GRCh38
rs754937212 Chr.9: 127789500-127789500 on Build GRCh38
rs769930999 Chr.9: 127789190-127789190 on Build GRCh38
rs745870791 Chr.9: 127789290-127789290 on Build GRCh38
rs765581062 Chr.9: 127789128-127789128 on Build GRCh38
rs141597514 Chr.9: 127789342-127789342 on Build GRCh38
rs1019575132 Chr.9: 127789544-127789544 on Build GRCh38
rs563333929 Chr.9: 127789548-127789548 on Build GRCh38
rs781196606 Chr.9: 127789286-127789286 on Build GRCh38
rs764624212 Chr.9: 127789139-127789139 on Build GRCh38
rs758446860 Chr.9: 127789567-127789567 on Build GRCh38
rs540712857 Chr.9: 127789282-127789282 on Build GRCh38
rs780150586 Chr.9: 127789496-127789496 on Build GRCh38
rs773619937 Chr.9: 127789322-127789322 on Build GRCh38
rs146248974 Chr.9: 127789360-127789360 on Build GRCh38
rs781588545 Chr.9: 127789582-127789582 on Build GRCh38
rs139560305 Chr.9: 127789162-127789162 on Build GRCh38
rs778462348 Chr.9: 127789246-127789246 on Build GRCh38
rs756164503 Chr.9: 127789386-127789386 on Build GRCh38
rs139856271 Chr.9: 127789336-127789336 on Build GRCh38
rs761228208 Chr.9: 127789329-127789329 on Build GRCh38
rs758087169 Chr.9: 127789272-127789272 on Build GRCh38
rs776385767 Chr.9: 127789184-127789184 on Build GRCh38
rs200376810 Chr.9: 127789484-127789484 on Build GRCh38
rs762456826 Chr.9: 127789460-127789460 on Build GRCh38
rs752742844 Chr.9: 127789561-127789561 on Build GRCh38
rs770616688 Chr.9: 127789486-127789486 on Build GRCh38
rs3217749 Chr.9: 127789432-127789432 on Build GRCh38
rs777252301 Chr.9: 127789273-127789273 on Build GRCh38
rs768690569 Chr.9: 127789232-127789232 on Build GRCh38
rs759675808 Chr.9: 127789189-127789189 on Build GRCh38
rs771708473 Chr.9: 127789165-127789165 on Build GRCh38
rs201475786 Chr.9: 127789526-127789526 on Build GRCh38
rs200559841 Chr.9: 127789510-127789510 on Build GRCh38
rs777924310 Chr.9: 127789505-127789505 on Build GRCh38
rs747730447 Chr.9: 127789164-127789164 on Build GRCh38
rs763329642 Chr.9: 127789197-127789197 on Build GRCh38
rs750834413 Chr.9: 127789228-127789228 on Build GRCh38
rs777679934 Chr.9: 127789167-127789167 on Build GRCh38
rs1004550576 Chr.9: 127789405-127789405 on Build GRCh38
rs755920229 Chr.9: 127789489-127789489 on Build GRCh38
rs368750293 Chr.9: 127789132-127789132 on Build GRCh38
rs139278925 Chr.9: 127789435-127789435 on Build GRCh38
rs35783224 Chr.9: 127789134-127789135 on Build GRCh38
rs781515520 Chr.9: 127789144-127789144 on Build GRCh38
rs149456430 Chr.9: 127789210-127789210 on Build GRCh38
rs777066206 Chr.9: 127789441-127789441 on Build GRCh38
rs556489645 Chr.9: 127789161-127789161 on Build GRCh38
rs753417800 Chr.9: 127789145-127789145 on Build GRCh38
rs559881586 Chr.9: 127789504-127789504 on Build GRCh38
rs763581624 Chr.9: 127789464-127789464 on Build GRCh38
rs766340319 Chr.9: 127789551-127789551 on Build GRCh38
rs376244572 Chr.9: 127789241-127789241 on Build GRCh38
rs763851751 Chr.9: 127789224-127789224 on Build GRCh38
rs778916606 Chr.9: 127789502-127789502 on Build GRCh38
rs201025809 Chr.9: 127789550-127789550 on Build GRCh38
rs145338923 Chr.9: 127789465-127789465 on Build GRCh38
rs773778565 Chr.9: 127789534-127789534 on Build GRCh38
rs376812602 Chr.9: 127789558-127789558 on Build GRCh38
rs774633888 Chr.9: 127789391-127789391 on Build GRCh38
rs954306428 Chr.9: 127789482-127789484 on Build GRCh38
rs748237241 Chr.9: 127789503-127789503 on Build GRCh38
rs886068205 Chr.9: 127789393-127789393 on Build GRCh38
rs752912301 Chr.9: 127789130-127789130 on Build GRCh38
rs868437001 Chr.9: 127789376-127789376 on Build GRCh38
rs756913144 Chr.9: 127789469-127789469 on Build GRCh38
rs377000414 Chr.9: 127789552-127789552 on Build GRCh38
rs775747149 Chr.9: 127789573-127789574 on Build GRCh38
rs961884432 Chr.9: 127789225-127789225 on Build GRCh38
rs201136315 Chr.9: 127789348-127789348 on Build GRCh38
rs779389613 Chr.9: 127789150-127789150 on Build GRCh38
rs776977894 Chr.9: 127789521-127789521 on Build GRCh38
rs760023385 Chr.9: 127789522-127789522 on Build GRCh38
rs749282983 Chr.9: 127789574-127789574 on Build GRCh38
rs374585241 Chr.9: 127789316-127789316 on Build GRCh38
rs150060991 Chr.9: 127789423-127789423 on Build GRCh38
rs746145143 Chr.9: 127789272-127789272 on Build GRCh38
rs766848600 Chr.9: 127789334-127789334 on Build GRCh38
rs145977259 Chr.9: 127789317-127789317 on Build GRCh38
rs771132092 Chr.9: 127789578-127789578 on Build GRCh38
rs774385639 Chr.9: 127789305-127789305 on Build GRCh38
rs202170960 Chr.9: 127789166-127789166 on Build GRCh38
rs761765306 Chr.9: 127789375-127789375 on Build GRCh38
rs1037994911 Chr.9: 127789163-127789163 on Build GRCh38
rs142704308 Chr.9: 127789426-127789426 on Build GRCh38
rs370938433 Chr.9: 127789274-127789274 on Build GRCh38
rs775710179 Chr.9: 127789196-127789196 on Build GRCh38
rs747301138 Chr.9: 127789570-127789570 on Build GRCh38
rs770113642 Chr.9: 127789502-127789502 on Build GRCh38
rs370044826 Chr.9: 127789285-127789285 on Build GRCh38
rs148443856 Chr.9: 127789390-127789390 on Build GRCh38
rs778878328 Chr.9: 127789560-127789560 on Build GRCh38
rs762641582 Chr.9: 127789175-127789175 on Build GRCh38
rs771220295 Chr.9: 127789512-127789512 on Build GRCh38
rs768949456 Chr.9: 127789303-127789303 on Build GRCh38
rs770932885 Chr.9: 127789179-127789179 on Build GRCh38
rs747387277 Chr.9: 127789508-127789508 on Build GRCh38
rs768088819 Chr.9: 127789227-127789227 on Build GRCh38
rs751991570 Chr.9: 127789211-127789211 on Build GRCh38
rs951146646 Chr.9: 127789450-127789450 on Build GRCh38
rs753758776 Chr.9: 127789497-127789497 on Build GRCh38
rs761350967 Chr.9: 127789535-127789535 on Build GRCh38
rs767296801 Chr.9: 127789537-127789537 on Build GRCh38
rs574082027 Chr.9: 127789300-127789300 on Build GRCh38
rs763822642 Chr.9: 127789340-127789340 on Build GRCh38
rs751860076 Chr.9: 127789141-127789141 on Build GRCh38

View Full Product Details