Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128169314-128169819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIZ1
Entrez Gene ID
25792
Gene Name
CDKN1A interacting zinc finger protein 1
Gene Aliases
LSFR1, NP94, ZNF356
UniGene
Hs.212395
Gene Chromosome Location
Chr.9: 128166065-128204383 on Build GRCh38
Transcripts
NM_001131015.1 NM_001131016.1 NM_001131017.1 NM_001131018.1 NM_001257975.1 NM_001257976.1 NM_012127.2 XM_005251888.3 XM_005251891.3 XM_005251892.3 XM_005251893.4 XM_006717039.3 XM_011518481.1 XM_011518482.2 XM_011518483.1 XM_017014594.1 XM_017014595.1 XM_017014596.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3654552 c.2131G>A p.D711N Chr.9: 128169420-128169420 on Build GRCh38
4905915 c.2209G>A p.D737N Chr.9: 128169420-128169420 on Build GRCh38
260115 c.2050C>T p.R684* Chr.9: 128169501-128169501 on Build GRCh38
5929110 c.2110-4C>T p.? Chr.9: 128169523-128169523 on Build GRCh38
4866781 c.2185C>T p.H729Y Chr.9: 128169444-128169444 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1035739679 Chr.9: 128169648-128169648 on Build GRCh38
rs988261485 Chr.9: 128169772-128169772 on Build GRCh38
rs769169316 Chr.9: 128169622-128169622 on Build GRCh38
rs550273493 Chr.9: 128169614-128169614 on Build GRCh38
rs574361051 Chr.9: 128169764-128169764 on Build GRCh38
rs762786694 Chr.9: 128169687-128169687 on Build GRCh38
rs749591724 Chr.9: 128169619-128169619 on Build GRCh38
rs552000697 Chr.9: 128169349-128169349 on Build GRCh38
rs759174857 Chr.9: 128169657-128169657 on Build GRCh38
rs764157910 Chr.9: 128169705-128169705 on Build GRCh38
rs761816343 Chr.9: 128169466-128169466 on Build GRCh38
rs759201456 Chr.9: 128169518-128169518 on Build GRCh38
rs778260735 Chr.9: 128169394-128169395 on Build GRCh38
rs754041232 Chr.9: 128169688-128169688 on Build GRCh38
rs761822318 Chr.9: 128169717-128169717 on Build GRCh38
rs767617433 Chr.9: 128169477-128169477 on Build GRCh38
rs748153430 Chr.9: 128169400-128169400 on Build GRCh38
rs775161523 Chr.9: 128169669-128169669 on Build GRCh38
rs776184434 Chr.9: 128169500-128169500 on Build GRCh38
rs572682745 Chr.9: 128169727-128169727 on Build GRCh38
rs45548032 Chr.9: 128169740-128169740 on Build GRCh38
rs966864725 Chr.9: 128169691-128169691 on Build GRCh38
rs376604922 Chr.9: 128169358-128169358 on Build GRCh38
rs1036933449 Chr.9: 128169565-128169565 on Build GRCh38
rs113839105 Chr.9: 128169620-128169620 on Build GRCh38
rs757369679 Chr.9: 128169597-128169597 on Build GRCh38
rs559214464 Chr.9: 128169363-128169363 on Build GRCh38
rs995333970 Chr.9: 128169611-128169611 on Build GRCh38
rs1005831570 Chr.9: 128169660-128169660 on Build GRCh38
rs539223689 Chr.9: 128169640-128169640 on Build GRCh38
rs764838835 Chr.9: 128169563-128169563 on Build GRCh38
rs371281592 Chr.9: 128169476-128169476 on Build GRCh38
rs758392645 Chr.9: 128169426-128169426 on Build GRCh38
rs747018616 Chr.9: 128169442-128169442 on Build GRCh38
rs201636309 Chr.9: 128169365-128169365 on Build GRCh38
rs1043696481 Chr.9: 128169606-128169606 on Build GRCh38
rs375899464 Chr.9: 128169487-128169487 on Build GRCh38
rs751585934 Chr.9: 128169709-128169709 on Build GRCh38
rs771862338 Chr.9: 128169553-128169554 on Build GRCh38
rs770096691 Chr.9: 128169472-128169472 on Build GRCh38
rs532154397 Chr.9: 128169344-128169344 on Build GRCh38
rs992394555 Chr.9: 128169667-128169667 on Build GRCh38
rs955442459 Chr.9: 128169768-128169768 on Build GRCh38
rs753683583 Chr.9: 128169384-128169384 on Build GRCh38
rs535299734 Chr.9: 128169751-128169751 on Build GRCh38
rs752622423 Chr.9: 128169420-128169420 on Build GRCh38
rs925482195 Chr.9: 128169720-128169720 on Build GRCh38
rs754683438 Chr.9: 128169385-128169385 on Build GRCh38
rs757566170 Chr.9: 128169446-128169446 on Build GRCh38
rs554981705 Chr.9: 128169757-128169757 on Build GRCh38
rs536284275 Chr.9: 128169610-128169610 on Build GRCh38
rs767552968 Chr.9: 128169733-128169733 on Build GRCh38
rs776187793 Chr.9: 128169655-128169655 on Build GRCh38
rs778061476 Chr.9: 128169436-128169436 on Build GRCh38
rs922759061 Chr.9: 128169730-128169730 on Build GRCh38
rs760124075 Chr.9: 128169490-128169490 on Build GRCh38
rs752528656 Chr.9: 128169529-128169529 on Build GRCh38
rs201094923 Chr.9: 128169393-128169393 on Build GRCh38
rs768452784 Chr.9: 128169346-128169346 on Build GRCh38
rs942892785 Chr.9: 128169458-128169458 on Build GRCh38
rs375729289 Chr.9: 128169382-128169382 on Build GRCh38
rs781303529 Chr.9: 128169609-128169609 on Build GRCh38
rs1025902834 Chr.9: 128169362-128169362 on Build GRCh38
rs944071383 Chr.9: 128169762-128169762 on Build GRCh38
rs373050955 Chr.9: 128169355-128169355 on Build GRCh38
rs186939440 Chr.9: 128169639-128169639 on Build GRCh38
rs559152606 Chr.9: 128169668-128169668 on Build GRCh38
rs543079803 Chr.9: 128169798-128169798 on Build GRCh38
rs984376900 Chr.9: 128169412-128169412 on Build GRCh38
rs762988791 Chr.9: 128169543-128169543 on Build GRCh38
rs764064184 Chr.9: 128169544-128169544 on Build GRCh38
rs772696737 Chr.9: 128169366-128169366 on Build GRCh38
rs1050156296 Chr.9: 128169433-128169433 on Build GRCh38
rs1026734811 Chr.9: 128169630-128169630 on Build GRCh38
rs766044318 Chr.9: 128169498-128169498 on Build GRCh38
rs766578350 Chr.9: 128169643-128169643 on Build GRCh38
rs959715447 Chr.9: 128169654-128169654 on Build GRCh38
rs370756371 Chr.9: 128169700-128169700 on Build GRCh38
rs139103399 Chr.9: 128169486-128169486 on Build GRCh38
rs764995298 Chr.9: 128169523-128169523 on Build GRCh38
rs951750818 Chr.9: 128169656-128169656 on Build GRCh38
rs370661541 Chr.9: 128169379-128169379 on Build GRCh38
rs946697075 Chr.9: 128169605-128169606 on Build GRCh38
rs781220647 Chr.9: 128169450-128169450 on Build GRCh38

View Full Product Details