Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128322199-128322710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ4
Entrez Gene ID
51117
Gene Name
coenzyme Q4
Gene Aliases
CGI-92, COQ10D7
UniGene
Hs.98541
Gene Chromosome Location
Chr.9: 128322486-128334072 on Build GRCh38
Transcripts
NM_001305942.1 NM_016035.4 XM_017014792.1 XM_017014793.1 NM_015679.1

Gene Symbol
TRUB2
Entrez Gene ID
26995
Gene Name
TruB pseudouridine synthase family member 2
Gene Aliases
CLONE24922
UniGene
Hs.632685
Gene Chromosome Location
Chr.9: 128309117-128322418 on Build GRCh38
Transcripts
NM_001305942.1 NM_016035.4 XM_017014792.1 XM_017014793.1 NM_015679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1660018 c.99A>C p.Q33H Chr.9: 128322310-128322310 on Build GRCh38
5410471 c.54C>T p.P18P Chr.9: 128322355-128322355 on Build GRCh38
3728623 c.25C>T p.L9L Chr.9: 128322384-128322384 on Build GRCh38
752634 c.105G>C p.L35L Chr.9: 128322304-128322304 on Build GRCh38
5000892 c.10G>T p.A4S Chr.9: 128322399-128322399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758685627 Chr.9: 128322451-128322451 on Build GRCh38
rs374714727 Chr.9: 128322417-128322417 on Build GRCh38
rs918202381 Chr.9: 128322639-128322639 on Build GRCh38
rs147400604 Chr.9: 128322224-128322224 on Build GRCh38
rs767463553 Chr.9: 128322295-128322295 on Build GRCh38
rs370791511 Chr.9: 128322258-128322258 on Build GRCh38
rs896661866 Chr.9: 128322264-128322264 on Build GRCh38
rs1027263331 Chr.9: 128322506-128322506 on Build GRCh38
rs756224366 Chr.9: 128322300-128322300 on Build GRCh38
rs753024337 Chr.9: 128322450-128322450 on Build GRCh38
rs755154929 Chr.9: 128322316-128322316 on Build GRCh38
rs747414475 Chr.9: 128322333-128322333 on Build GRCh38
rs1006716579 Chr.9: 128322496-128322496 on Build GRCh38
rs563966688 Chr.9: 128322394-128322394 on Build GRCh38
rs368089230 Chr.9: 128322438-128322438 on Build GRCh38
rs747615827 Chr.9: 128322388-128322388 on Build GRCh38
rs920092693 Chr.9: 128322226-128322226 on Build GRCh38
rs759886290 Chr.9: 128322549-128322551 on Build GRCh38
rs921882053 Chr.9: 128322677-128322677 on Build GRCh38
rs766547290 Chr.9: 128322370-128322370 on Build GRCh38
rs760518065 Chr.9: 128322368-128322368 on Build GRCh38
rs778427072 Chr.9: 128322329-128322329 on Build GRCh38
rs767234953 Chr.9: 128322365-128322365 on Build GRCh38
rs376308828 Chr.9: 128322270-128322270 on Build GRCh38
rs754147692 Chr.9: 128322313-128322313 on Build GRCh38
rs772679595 Chr.9: 128322267-128322267 on Build GRCh38
rs759569675 Chr.9: 128322432-128322432 on Build GRCh38
rs546796184 Chr.9: 128322652-128322652 on Build GRCh38
rs577886586 Chr.9: 128322480-128322480 on Build GRCh38
rs1027231977 Chr.9: 128322354-128322354 on Build GRCh38
rs766338065 Chr.9: 128322475-128322475 on Build GRCh38
rs992467630 Chr.9: 128322630-128322630 on Build GRCh38
rs1036285389 Chr.9: 128322557-128322557 on Build GRCh38
rs1052021122 Chr.9: 128322456-128322456 on Build GRCh38
rs947309098 Chr.9: 128322268-128322268 on Build GRCh38
rs200070066 Chr.9: 128322427-128322427 on Build GRCh38
rs779464432 Chr.9: 128322318-128322318 on Build GRCh38
rs188303869 Chr.9: 128322362-128322362 on Build GRCh38
rs201804445 Chr.9: 128322553-128322553 on Build GRCh38
rs890266790 Chr.9: 128322468-128322468 on Build GRCh38
rs948831104 Chr.9: 128322233-128322233 on Build GRCh38
rs758295180 Chr.9: 128322403-128322403 on Build GRCh38
rs978716253 Chr.9: 128322656-128322656 on Build GRCh38
rs968529726 Chr.9: 128322575-128322575 on Build GRCh38
rs771308348 Chr.9: 128322390-128322390 on Build GRCh38
rs529074080 Chr.9: 128322481-128322481 on Build GRCh38
rs772988201 Chr.9: 128322426-128322426 on Build GRCh38
rs776585923 Chr.9: 128322429-128322429 on Build GRCh38
rs752674420 Chr.9: 128322387-128322387 on Build GRCh38
rs190492755 Chr.9: 128322500-128322500 on Build GRCh38
rs750323144 Chr.9: 128322299-128322299 on Build GRCh38
rs573393921 Chr.9: 128322280-128322280 on Build GRCh38
rs988627392 Chr.9: 128322567-128322567 on Build GRCh38
rs778266043 Chr.9: 128322387-128322387 on Build GRCh38
rs1044439188 Chr.9: 128322245-128322245 on Build GRCh38
rs913310090 Chr.9: 128322601-128322601 on Build GRCh38
rs764589492 Chr.9: 128322457-128322457 on Build GRCh38
rs887887725 Chr.9: 128322485-128322485 on Build GRCh38
rs947106163 Chr.9: 128322399-128322399 on Build GRCh38
rs567883723 Chr.9: 128322294-128322294 on Build GRCh38
rs544236306 Chr.9: 128322375-128322375 on Build GRCh38
rs771913063 Chr.9: 128322424-128322424 on Build GRCh38
rs890183974 Chr.9: 128322303-128322303 on Build GRCh38
rs960411192 Chr.9: 128322383-128322383 on Build GRCh38
rs772518225 Chr.9: 128322323-128322323 on Build GRCh38
rs779663936 Chr.9: 128322392-128322392 on Build GRCh38
rs2231629 Chr.9: 128322225-128322225 on Build GRCh38
rs995765612 Chr.9: 128322353-128322353 on Build GRCh38
rs1021410495 Chr.9: 128322572-128322572 on Build GRCh38
rs758035132 Chr.9: 128322412-128322412 on Build GRCh38
rs200913132 Chr.9: 128322407-128322407 on Build GRCh38
rs1005058853 Chr.9: 128322471-128322471 on Build GRCh38
rs371765061 Chr.9: 128322380-128322380 on Build GRCh38
rs776332448 Chr.9: 128322269-128322269 on Build GRCh38
rs748769733 Chr.9: 128322405-128322405 on Build GRCh38
rs769117197 Chr.9: 128322274-128322274 on Build GRCh38
rs11539574 Chr.9: 128322374-128322374 on Build GRCh38
rs376651708 Chr.9: 128322546-128322546 on Build GRCh38
rs979829650 Chr.9: 128322416-128322416 on Build GRCh38
rs758845138 Chr.9: 128322382-128322382 on Build GRCh38
rs755280610 Chr.9: 128322376-128322376 on Build GRCh38
rs200866588 Chr.9: 128322278-128322278 on Build GRCh38
rs764012479 Chr.9: 128322281-128322281 on Build GRCh38
rs766208855 Chr.9: 128322428-128322428 on Build GRCh38
rs11539570 Chr.9: 128322349-128322349 on Build GRCh38
rs1053513471 Chr.9: 128322263-128322263 on Build GRCh38
rs749406355 Chr.9: 128322251-128322251 on Build GRCh38
rs548720657 Chr.9: 128322495-128322495 on Build GRCh38
rs748735739 Chr.9: 128322266-128322266 on Build GRCh38
rs1030217932 Chr.9: 128322526-128322526 on Build GRCh38
rs531538915 Chr.9: 128322525-128322525 on Build GRCh38
rs139996554 Chr.9: 128322527-128322527 on Build GRCh38
rs928761893 Chr.9: 128322373-128322373 on Build GRCh38
rs748504572 Chr.9: 128322321-128322321 on Build GRCh38
rs750374626 Chr.9: 128322367-128322367 on Build GRCh38
rs980215769 Chr.9: 128322243-128322243 on Build GRCh38
rs150001858 Chr.9: 128322646-128322646 on Build GRCh38
rs779940484 Chr.9: 128322310-128322310 on Build GRCh38
rs1006252550 Chr.9: 128322312-128322312 on Build GRCh38
rs928707950 Chr.9: 128322247-128322247 on Build GRCh38
rs989311226 Chr.9: 128322598-128322598 on Build GRCh38
rs751324221 Chr.9: 128322284-128322284 on Build GRCh38
rs368494818 Chr.9: 128322334-128322334 on Build GRCh38
rs142720314 Chr.9: 128322355-128322355 on Build GRCh38
rs757172611 Chr.9: 128322522-128322522 on Build GRCh38
rs914350659 Chr.9: 128322398-128322398 on Build GRCh38
rs377135272 Chr.9: 128322293-128322293 on Build GRCh38
rs898859830 Chr.9: 128322483-128322483 on Build GRCh38
rs116050651 Chr.9: 128322588-128322588 on Build GRCh38
rs769079958 Chr.9: 128322257-128322257 on Build GRCh38
rs773998115 Chr.9: 128322413-128322413 on Build GRCh38

View Full Product Details