Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128333598-128334094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ4
Entrez Gene ID
51117
Gene Name
coenzyme Q4
Gene Aliases
CGI-92, COQ10D7
UniGene
Hs.98541
Gene Chromosome Location
Chr.9: 128322486-128334072 on Build GRCh38
Transcripts
NM_001305942.1 NM_016035.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6033889 c.781G>A p.V261I Chr.9: 128333628-128333628 on Build GRCh38
1105545 c.780C>T p.H260H Chr.9: 128333627-128333627 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200203275 Chr.9: 128333627-128333627 on Build GRCh38
rs959950093 Chr.9: 128333849-128333849 on Build GRCh38
rs770009029 Chr.9: 128333962-128333962 on Build GRCh38
rs897232395 Chr.9: 128333774-128333774 on Build GRCh38
rs184775783 Chr.9: 128333911-128333911 on Build GRCh38
rs937992640 Chr.9: 128333914-128333914 on Build GRCh38
rs1026107657 Chr.9: 128333991-128333991 on Build GRCh38
rs780717789 Chr.9: 128333661-128333661 on Build GRCh38
rs193050173 Chr.9: 128333904-128333904 on Build GRCh38
rs576229714 Chr.9: 128333751-128333751 on Build GRCh38
rs769009364 Chr.9: 128333777-128333777 on Build GRCh38
rs944193359 Chr.9: 128333756-128333756 on Build GRCh38
rs756399844 Chr.9: 128333626-128333626 on Build GRCh38
rs997577163 Chr.9: 128334043-128334044 on Build GRCh38
rs1050411616 Chr.9: 128333735-128333735 on Build GRCh38
rs779317783 Chr.9: 128333650-128333650 on Build GRCh38
rs1029952136 Chr.9: 128334044-128334044 on Build GRCh38
rs111451353 Chr.9: 128333830-128333830 on Build GRCh38
rs149137752 Chr.9: 128333628-128333628 on Build GRCh38
rs912811706 Chr.9: 128334040-128334040 on Build GRCh38
rs1044173460 Chr.9: 128333894-128333894 on Build GRCh38
rs187424122 Chr.9: 128333866-128333866 on Build GRCh38
rs905856778 Chr.9: 128333811-128333811 on Build GRCh38
rs370347021 Chr.9: 128333683-128333683 on Build GRCh38
rs112374081 Chr.9: 128333892-128333892 on Build GRCh38
rs774771580 Chr.9: 128333983-128333983 on Build GRCh38
rs755325123 Chr.9: 128333633-128333633 on Build GRCh38
rs931633975 Chr.9: 128333990-128333990 on Build GRCh38
rs376033556 Chr.9: 128333982-128333982 on Build GRCh38
rs1037166994 Chr.9: 128333771-128333771 on Build GRCh38
rs989054825 Chr.9: 128333885-128333885 on Build GRCh38
rs756763729 Chr.9: 128333660-128333660 on Build GRCh38
rs927483514 Chr.9: 128333812-128333812 on Build GRCh38
rs753196999 Chr.9: 128333659-128333659 on Build GRCh38
rs779617452 Chr.9: 128333687-128333687 on Build GRCh38
rs745581380 Chr.9: 128333681-128333681 on Build GRCh38
rs921619901 Chr.9: 128333973-128333973 on Build GRCh38
rs896399753 Chr.9: 128333960-128333960 on Build GRCh38
rs914025502 Chr.9: 128333842-128333842 on Build GRCh38
rs750652321 Chr.9: 128333619-128333619 on Build GRCh38
rs951796217 Chr.9: 128333642-128333642 on Build GRCh38
rs995524836 Chr.9: 128333782-128333782 on Build GRCh38
rs887578603 Chr.9: 128333993-128333993 on Build GRCh38
rs371380712 Chr.9: 128333947-128333947 on Build GRCh38
rs1017278418 Chr.9: 128333704-128333704 on Build GRCh38
rs1005982460 Chr.9: 128333996-128333996 on Build GRCh38
rs993331434 Chr.9: 128333970-128333970 on Build GRCh38
rs964702249 Chr.9: 128333711-128333711 on Build GRCh38
rs932916857 Chr.9: 128333709-128333709 on Build GRCh38
rs189694101 Chr.9: 128334002-128334002 on Build GRCh38
rs779144812 Chr.9: 128333667-128333667 on Build GRCh38
rs934174740 Chr.9: 128333794-128333794 on Build GRCh38
rs145061462 Chr.9: 128334045-128334045 on Build GRCh38
rs1037622564 Chr.9: 128334065-128334065 on Build GRCh38
rs557576728 Chr.9: 128333933-128333933 on Build GRCh38
rs978493382 Chr.9: 128333927-128333927 on Build GRCh38
rs964431534 Chr.9: 128334004-128334005 on Build GRCh38
rs749113971 Chr.9: 128333695-128333695 on Build GRCh38
rs768392340 Chr.9: 128333697-128333697 on Build GRCh38
rs748169528 Chr.9: 128333673-128333676 on Build GRCh38
rs1026973587 Chr.9: 128333807-128333807 on Build GRCh38
rs113961503 Chr.9: 128334023-128334023 on Build GRCh38
rs985762050 Chr.9: 128334000-128334000 on Build GRCh38
rs746136470 Chr.9: 128333754-128333754 on Build GRCh38
rs951255812 Chr.9: 128333954-128333965 on Build GRCh38

View Full Product Details